Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 20860270-20860775 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCUN1D3
Entrez Gene ID
123879
Gene Name
defective in cullin neddylation 1 domain containing 3
Gene Aliases
44M2.4, SCCRO3
UniGene
Hs.101007
Gene Chromosome Location
Chr.16: 20854925-20900358 on Build GRCh38
Transcripts
NM_173475.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1678899 c.493C>T p.R165W Chr.16: 20860308-20860308 on Build GRCh38
3667808 c.462A>G p.I154M Chr.16: 20860339-20860339 on Build GRCh38
5800235 c.504C>G p.S168R Chr.16: 20860297-20860297 on Build GRCh38
7176149 c.525A>G p.Q175Q Chr.16: 20860276-20860276 on Build GRCh38
6838997 c.514G>A p.E172K Chr.16: 20860287-20860287 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778304055 Chr.16: 20860306-20860306 on Build GRCh38
rs1028914813 Chr.16: 20860515-20860515 on Build GRCh38
rs1022413407 Chr.16: 20860407-20860407 on Build GRCh38
rs367576234 Chr.16: 20860398-20860398 on Build GRCh38
rs544289895 Chr.16: 20860459-20860459 on Build GRCh38
rs766445392 Chr.16: 20860375-20860375 on Build GRCh38
rs35960243 Chr.16: 20860509-20860510 on Build GRCh38
rs147094254 Chr.16: 20860716-20860716 on Build GRCh38
rs940108190 Chr.16: 20860580-20860580 on Build GRCh38
rs202156477 Chr.16: 20860320-20860320 on Build GRCh38
rs999705102 Chr.16: 20860416-20860416 on Build GRCh38
rs182396062 Chr.16: 20860541-20860541 on Build GRCh38
rs902206070 Chr.16: 20860658-20860658 on Build GRCh38
rs77432343 Chr.16: 20860357-20860357 on Build GRCh38
rs61743475 Chr.16: 20860321-20860321 on Build GRCh38
rs895396223 Chr.16: 20860618-20860618 on Build GRCh38
rs902256700 Chr.16: 20860413-20860413 on Build GRCh38
rs199941022 Chr.16: 20860302-20860302 on Build GRCh38
rs1043379520 Chr.16: 20860648-20860648 on Build GRCh38
rs569490377 Chr.16: 20860609-20860609 on Build GRCh38
rs946996640 Chr.16: 20860620-20860620 on Build GRCh38
rs374366221 Chr.16: 20860297-20860297 on Build GRCh38
rs563767822 Chr.16: 20860519-20860519 on Build GRCh38
rs143835944 Chr.16: 20860307-20860307 on Build GRCh38
rs751680636 Chr.16: 20860382-20860382 on Build GRCh38
rs368597287 Chr.16: 20860338-20860338 on Build GRCh38
rs999254324 Chr.16: 20860675-20860675 on Build GRCh38
rs759736093 Chr.16: 20860386-20860386 on Build GRCh38
rs552447472 Chr.16: 20860600-20860600 on Build GRCh38
rs953991097 Chr.16: 20860533-20860533 on Build GRCh38
rs775124888 Chr.16: 20860333-20860333 on Build GRCh38
rs770510257 Chr.16: 20860313-20860313 on Build GRCh38
rs973244029 Chr.16: 20860586-20860586 on Build GRCh38
rs546625565 Chr.16: 20860730-20860730 on Build GRCh38
rs752894297 Chr.16: 20860410-20860410 on Build GRCh38
rs199765506 Chr.16: 20860336-20860336 on Build GRCh38
rs974632200 Chr.16: 20860494-20860494 on Build GRCh38
rs373730831 Chr.16: 20860373-20860373 on Build GRCh38
rs771889938 Chr.16: 20860330-20860330 on Build GRCh38
rs776561494 Chr.16: 20860344-20860344 on Build GRCh38
rs767734395 Chr.16: 20860392-20860392 on Build GRCh38
rs564464116 Chr.16: 20860693-20860693 on Build GRCh38
rs772772503 Chr.16: 20860717-20860717 on Build GRCh38
rs771694536 Chr.16: 20860606-20860606 on Build GRCh38

View Full Product Details