Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67529518-67530010 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM65A
Entrez Gene ID
79567
Gene Name
family with sequence similarity 65 member A
Gene Aliases
-
UniGene
Hs.152717
Gene Chromosome Location
Chr.16: 67528814-67546788 on Build GRCh38
Transcripts
NM_001193522.1 NM_001193523.1 NM_024519.3 XM_011523321.1 XM_011523325.1 XM_011523326.1

SNP Detail

SNP ID Chromosome Location
rs920367278 Chr.16: 67529926-67529926 on Build GRCh38
rs769624040 Chr.16: 67529886-67529886 on Build GRCh38
rs991433379 Chr.16: 67529556-67529556 on Build GRCh38
rs540759595 Chr.16: 67529956-67529956 on Build GRCh38
rs962928337 Chr.16: 67529859-67529859 on Build GRCh38
rs960977660 Chr.16: 67529827-67529827 on Build GRCh38
rs909696740 Chr.16: 67529927-67529927 on Build GRCh38
rs140179437 Chr.16: 67529593-67529593 on Build GRCh38
rs1011386817 Chr.16: 67529741-67529741 on Build GRCh38
rs1012507980 Chr.16: 67529836-67529836 on Build GRCh38
rs920231171 Chr.16: 67529589-67529589 on Build GRCh38
rs573215053 Chr.16: 67529940-67529940 on Build GRCh38
rs950361053 Chr.16: 67529937-67529937 on Build GRCh38
rs1047661157 Chr.16: 67529625-67529625 on Build GRCh38
rs983150511 Chr.16: 67529939-67529939 on Build GRCh38
rs191282321 Chr.16: 67529596-67529596 on Build GRCh38
rs1020316226 Chr.16: 67529622-67529622 on Build GRCh38
rs754398209 Chr.16: 67529788-67529788 on Build GRCh38
rs973178054 Chr.16: 67529585-67529585 on Build GRCh38
rs776819663 Chr.16: 67529987-67529987 on Build GRCh38
rs967583397 Chr.16: 67529640-67529640 on Build GRCh38
rs1039247001 Chr.16: 67529682-67529682 on Build GRCh38
rs911852179 Chr.16: 67529627-67529627 on Build GRCh38
rs944515651 Chr.16: 67529679-67529679 on Build GRCh38
rs779152844 Chr.16: 67529794-67529794 on Build GRCh38
rs1035231221 Chr.16: 67529784-67529784 on Build GRCh38
rs755482293 Chr.16: 67529789-67529789 on Build GRCh38
rs752780985 Chr.16: 67529812-67529812 on Build GRCh38
rs1024183702 Chr.16: 67529844-67529844 on Build GRCh38
rs976346433 Chr.16: 67529744-67529744 on Build GRCh38
rs910993905 Chr.16: 67529714-67529714 on Build GRCh38
rs747585064 Chr.16: 67529578-67529578 on Build GRCh38
rs758588544 Chr.16: 67529832-67529832 on Build GRCh38
rs372359867 Chr.16: 67529742-67529742 on Build GRCh38
rs554954059 Chr.16: 67529869-67529869 on Build GRCh38
rs915654344 Chr.16: 67529570-67529570 on Build GRCh38
rs911569120 Chr.16: 67529960-67529960 on Build GRCh38
rs902507486 Chr.16: 67529755-67529755 on Build GRCh38
rs36004773 Chr.16: 67529700-67529700 on Build GRCh38
rs745706458 Chr.16: 67529882-67529882 on Build GRCh38
rs1033174326 Chr.16: 67529655-67529655 on Build GRCh38
rs1000458619 Chr.16: 67529648-67529648 on Build GRCh38
rs888309866 Chr.16: 67529551-67529551 on Build GRCh38
rs777094999 Chr.16: 67529624-67529626 on Build GRCh38
rs1056305166 Chr.16: 67529573-67529573 on Build GRCh38
rs778032470 Chr.16: 67529834-67529834 on Build GRCh38
rs35227444 Chr.16: 67529554-67529554 on Build GRCh38
rs984041218 Chr.16: 67529884-67529884 on Build GRCh38
rs938384806 Chr.16: 67529667-67529667 on Build GRCh38
rs999503597 Chr.16: 67529779-67529779 on Build GRCh38

View Full Product Details