Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67538576-67539074 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM65A
Entrez Gene ID
79567
Gene Name
family with sequence similarity 65 member A
Gene Aliases
-
UniGene
Hs.152717
Gene Chromosome Location
Chr.16: 67528814-67546788 on Build GRCh38
Transcripts
NM_001193522.1 NM_001193523.1 NM_001193524.1 NM_024519.3 XM_011523321.1 XM_011523322.1 XM_011523324.2 XM_011523325.1 XM_011523326.1 XM_017023663.1 XM_017023664.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4061875 c.105-1G>A p.? Chr.16: 67538671-67538671 on Build GRCh38
6867070 c.336G>T p.L112F Chr.16: 67539068-67539068 on Build GRCh38
5157648 c.145delC p.P50fs*36 Chr.16: 67538700-67538700 on Build GRCh38
6659975 c.106C>T p.L36F Chr.16: 67538661-67538661 on Build GRCh38
972450 c.195C>T p.A65A Chr.16: 67538762-67538762 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375908811 Chr.16: 67538814-67538814 on Build GRCh38
rs754613083 Chr.16: 67538768-67538768 on Build GRCh38
rs920634056 Chr.16: 67538734-67538734 on Build GRCh38
rs774719944 Chr.16: 67538634-67538634 on Build GRCh38
rs775344220 Chr.16: 67538670-67538672 on Build GRCh38
rs906737629 Chr.16: 67538617-67538617 on Build GRCh38
rs957787396 Chr.16: 67538920-67538920 on Build GRCh38
rs772010535 Chr.16: 67538984-67538984 on Build GRCh38
rs199516887 Chr.16: 67538661-67538661 on Build GRCh38
rs751544853 Chr.16: 67538662-67538662 on Build GRCh38
rs371999360 Chr.16: 67539046-67539046 on Build GRCh38
rs556490179 Chr.16: 67538808-67538808 on Build GRCh38
rs758754618 Chr.16: 67538946-67538946 on Build GRCh38
rs200677001 Chr.16: 67538701-67538701 on Build GRCh38
rs548551482 Chr.16: 67538684-67538684 on Build GRCh38
rs774338405 Chr.16: 67538831-67538831 on Build GRCh38
rs745527672 Chr.16: 67538621-67538621 on Build GRCh38
rs767541942 Chr.16: 67538730-67538730 on Build GRCh38
rs773229607 Chr.16: 67538652-67538652 on Build GRCh38
rs759987634 Chr.16: 67538602-67538622 on Build GRCh38
rs528170969 Chr.16: 67538601-67538601 on Build GRCh38
rs949254274 Chr.16: 67538672-67538672 on Build GRCh38
rs958032753 Chr.16: 67538643-67538643 on Build GRCh38
rs1014790436 Chr.16: 67538938-67538938 on Build GRCh38
rs374771901 Chr.16: 67539038-67539038 on Build GRCh38
rs780781753 Chr.16: 67538616-67538616 on Build GRCh38
rs770920020 Chr.16: 67538798-67538798 on Build GRCh38
rs376326486 Chr.16: 67538641-67538641 on Build GRCh38
rs368689898 Chr.16: 67538855-67538855 on Build GRCh38
rs753113948 Chr.16: 67538945-67538945 on Build GRCh38
rs757578255 Chr.16: 67538732-67538732 on Build GRCh38
rs113573319 Chr.16: 67538718-67538718 on Build GRCh38
rs56968620 Chr.16: 67538840-67538840 on Build GRCh38
rs771239489 Chr.16: 67538712-67538712 on Build GRCh38
rs371921564 Chr.16: 67539041-67539041 on Build GRCh38
rs374261841 Chr.16: 67538746-67538746 on Build GRCh38
rs954804897 Chr.16: 67538927-67538927 on Build GRCh38
rs564723641 Chr.16: 67538609-67538609 on Build GRCh38
rs139082757 Chr.16: 67538695-67538695 on Build GRCh38
rs778043733 Chr.16: 67538947-67538947 on Build GRCh38
rs769292995 Chr.16: 67538622-67538622 on Build GRCh38
rs148612121 Chr.16: 67539017-67539017 on Build GRCh38
rs569084656 Chr.16: 67538658-67538658 on Build GRCh38
rs748477296 Chr.16: 67538775-67538775 on Build GRCh38
rs759736492 Chr.16: 67538723-67538723 on Build GRCh38
rs539860066 Chr.16: 67538996-67538996 on Build GRCh38
rs371498647 Chr.16: 67539018-67539018 on Build GRCh38
rs181876439 Chr.16: 67538971-67538971 on Build GRCh38
rs750011303 Chr.16: 67538735-67538735 on Build GRCh38
rs749456061 Chr.16: 67538977-67538977 on Build GRCh38
rs776153028 Chr.16: 67538712-67538712 on Build GRCh38
rs752347460 Chr.16: 67539025-67539025 on Build GRCh38
rs756885567 Chr.16: 67539026-67539026 on Build GRCh38
rs372941924 Chr.16: 67538726-67538726 on Build GRCh38
rs199987436 Chr.16: 67538844-67538844 on Build GRCh38
rs749732081 Chr.16: 67538697-67538697 on Build GRCh38
rs150369207 Chr.16: 67539031-67539031 on Build GRCh38
rs751348548 Chr.16: 67538728-67538728 on Build GRCh38
rs775964913 Chr.16: 67538625-67538625 on Build GRCh38
rs754624221 Chr.16: 67538863-67538863 on Build GRCh38
rs778235631 Chr.16: 67538793-67538793 on Build GRCh38
rs368350376 Chr.16: 67538953-67538953 on Build GRCh38
rs759030922 Chr.16: 67539004-67539004 on Build GRCh38
rs117467133 Chr.16: 67538631-67538631 on Build GRCh38
rs987711522 Chr.16: 67538696-67538696 on Build GRCh38
rs781526223 Chr.16: 67538760-67538760 on Build GRCh38
rs776712527 Chr.16: 67538716-67538716 on Build GRCh38
rs772525243 Chr.16: 67538706-67538706 on Build GRCh38
rs1028870125 Chr.16: 67538922-67538922 on Build GRCh38
rs998381973 Chr.16: 67538820-67538820 on Build GRCh38
rs765255033 Chr.16: 67539040-67539041 on Build GRCh38
rs201157660 Chr.16: 67538691-67538691 on Build GRCh38
rs575068102 Chr.16: 67538824-67538824 on Build GRCh38
rs746575229 Chr.16: 67538600-67538600 on Build GRCh38
rs201528706 Chr.16: 67538692-67538692 on Build GRCh38
rs772613318 Chr.16: 67538781-67538781 on Build GRCh38
rs368012173 Chr.16: 67538867-67538867 on Build GRCh38
rs753450193 Chr.16: 67538758-67538758 on Build GRCh38
rs996252151 Chr.16: 67538875-67538875 on Build GRCh38
rs747177030 Chr.16: 67538797-67538797 on Build GRCh38
rs974725133 Chr.16: 67538689-67538689 on Build GRCh38
rs755576251 Chr.16: 67538738-67538738 on Build GRCh38
rs539969878 Chr.16: 67538603-67538603 on Build GRCh38
rs760718190 Chr.16: 67538711-67538711 on Build GRCh38
rs749923451 Chr.16: 67538690-67538690 on Build GRCh38
rs550384936 Chr.16: 67538654-67538654 on Build GRCh38
rs770201656 Chr.16: 67538969-67538969 on Build GRCh38
rs761687448 Chr.16: 67538832-67538832 on Build GRCh38
rs760915192 Chr.16: 67538653-67538653 on Build GRCh38
rs1032123812 Chr.16: 67538638-67538638 on Build GRCh38
rs762638930 Chr.16: 67538660-67538660 on Build GRCh38
rs149487364 Chr.16: 67538748-67538748 on Build GRCh38
rs570973377 Chr.16: 67538707-67538707 on Build GRCh38
rs367981886 Chr.16: 67539044-67539044 on Build GRCh38
rs761883642 Chr.16: 67538729-67538729 on Build GRCh38
rs756822638 Chr.16: 67538614-67538614 on Build GRCh38
rs779284604 Chr.16: 67538774-67538774 on Build GRCh38
rs768910968 Chr.16: 67538699-67538699 on Build GRCh38
rs767629990 Chr.16: 67538836-67538836 on Build GRCh38
rs763705850 Chr.16: 67538711-67538712 on Build GRCh38
rs772544743 Chr.16: 67538645-67538648 on Build GRCh38
rs959284564 Chr.16: 67539033-67539033 on Build GRCh38
rs906784354 Chr.16: 67538791-67538791 on Build GRCh38
rs766312127 Chr.16: 67538998-67538998 on Build GRCh38
rs760403210 Chr.16: 67538700-67538700 on Build GRCh38
rs111681103 Chr.16: 67538934-67538934 on Build GRCh38
rs572541875 Chr.16: 67538916-67538916 on Build GRCh38
rs140402307 Chr.16: 67538703-67538703 on Build GRCh38
rs745940045 Chr.16: 67538959-67538959 on Build GRCh38
rs560970042 Chr.16: 67538668-67538668 on Build GRCh38
rs761701023 Chr.16: 67538837-67538838 on Build GRCh38
rs369978963 Chr.16: 67538628-67538628 on Build GRCh38
rs143856664 Chr.16: 67539003-67539003 on Build GRCh38

View Full Product Details