Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67539724-67540217 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM65A
Entrez Gene ID
79567
Gene Name
family with sequence similarity 65 member A
Gene Aliases
-
UniGene
Hs.152717
Gene Chromosome Location
Chr.16: 67528814-67546788 on Build GRCh38
Transcripts
NM_001193522.1 NM_001193523.1 NM_001193524.1 NM_024519.3 XM_011523321.1 XM_011523322.1 XM_011523324.2 XM_011523325.1 XM_011523326.1 XM_017023663.1 XM_017023664.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
972451 c.381C>T p.R127R Chr.16: 67539866-67539866 on Build GRCh38
1588485 c.393C>T p.R131R Chr.16: 67539866-67539866 on Build GRCh38
1206408 c.527G>A p.S176N Chr.16: 67540165-67540165 on Build GRCh38
7349422 c.407C>A p.A136D Chr.16: 67539892-67539892 on Build GRCh38
7283603 c.391C>T p.R131* Chr.16: 67539876-67539876 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770391113 Chr.16: 67540178-67540178 on Build GRCh38
rs542023070 Chr.16: 67539985-67539985 on Build GRCh38
rs377590115 Chr.16: 67539798-67539798 on Build GRCh38
rs369358400 Chr.16: 67539806-67539806 on Build GRCh38
rs371716961 Chr.16: 67540089-67540089 on Build GRCh38
rs761111085 Chr.16: 67539942-67539942 on Build GRCh38
rs138036670 Chr.16: 67539780-67539780 on Build GRCh38
rs779963301 Chr.16: 67539911-67539911 on Build GRCh38
rs752677665 Chr.16: 67540024-67540024 on Build GRCh38
rs758205404 Chr.16: 67540022-67540022 on Build GRCh38
rs761920550 Chr.16: 67539895-67539895 on Build GRCh38
rs768260871 Chr.16: 67539842-67539842 on Build GRCh38
rs1002049784 Chr.16: 67539983-67539983 on Build GRCh38
rs755304377 Chr.16: 67539799-67539799 on Build GRCh38
rs756397379 Chr.16: 67540117-67540117 on Build GRCh38
rs762438443 Chr.16: 67539838-67539838 on Build GRCh38
rs199511942 Chr.16: 67540192-67540192 on Build GRCh38
rs369864235 Chr.16: 67540120-67540120 on Build GRCh38
rs757983170 Chr.16: 67540153-67540153 on Build GRCh38
rs371091978 Chr.16: 67540193-67540193 on Build GRCh38
rs767706957 Chr.16: 67540191-67540191 on Build GRCh38
rs747592741 Chr.16: 67540055-67540055 on Build GRCh38
rs979689078 Chr.16: 67540108-67540108 on Build GRCh38
rs770761451 Chr.16: 67539817-67539817 on Build GRCh38
rs374336975 Chr.16: 67540086-67540086 on Build GRCh38
rs375437388 Chr.16: 67539987-67539987 on Build GRCh38
rs755077967 Chr.16: 67539894-67539894 on Build GRCh38
rs201004927 Chr.16: 67540184-67540184 on Build GRCh38
rs370845659 Chr.16: 67540038-67540038 on Build GRCh38
rs574578281 Chr.16: 67539927-67539927 on Build GRCh38
rs879148900 Chr.16: 67539816-67539816 on Build GRCh38
rs751794768 Chr.16: 67540111-67540111 on Build GRCh38
rs201301988 Chr.16: 67540119-67540119 on Build GRCh38
rs749156822 Chr.16: 67539914-67539914 on Build GRCh38
rs775856745 Chr.16: 67540179-67540179 on Build GRCh38
rs754026024 Chr.16: 67539889-67539889 on Build GRCh38
rs781070962 Chr.16: 67539906-67539906 on Build GRCh38
rs775021325 Chr.16: 67539837-67539837 on Build GRCh38
rs774376004 Chr.16: 67540182-67540182 on Build GRCh38
rs911648527 Chr.16: 67539839-67539839 on Build GRCh38
rs762065904 Chr.16: 67540183-67540183 on Build GRCh38
rs756092484 Chr.16: 67539910-67539910 on Build GRCh38
rs201474523 Chr.16: 67540141-67540141 on Build GRCh38
rs201137113 Chr.16: 67539818-67539818 on Build GRCh38
rs144382974 Chr.16: 67540163-67540163 on Build GRCh38
rs750276204 Chr.16: 67540026-67540026 on Build GRCh38
rs572431122 Chr.16: 67539772-67539772 on Build GRCh38
rs760118628 Chr.16: 67540091-67540091 on Build GRCh38
rs754994468 Chr.16: 67540137-67540137 on Build GRCh38
rs560905148 Chr.16: 67539994-67539997 on Build GRCh38
rs766973907 Chr.16: 67539879-67539879 on Build GRCh38
rs746194435 Chr.16: 67540172-67540172 on Build GRCh38
rs373261651 Chr.16: 67539877-67539877 on Build GRCh38
rs746589284 Chr.16: 67540066-67540066 on Build GRCh38
rs771594874 Chr.16: 67539932-67539932 on Build GRCh38
rs1054907137 Chr.16: 67540018-67540018 on Build GRCh38
rs905056600 Chr.16: 67539950-67539952 on Build GRCh38
rs372616105 Chr.16: 67540006-67540006 on Build GRCh38
rs773686729 Chr.16: 67539919-67539919 on Build GRCh38
rs764095596 Chr.16: 67540109-67540109 on Build GRCh38
rs146876313 Chr.16: 67540096-67540096 on Build GRCh38
rs764278310 Chr.16: 67540025-67540025 on Build GRCh38
rs767949905 Chr.16: 67539874-67539874 on Build GRCh38
rs372452161 Chr.16: 67540167-67540167 on Build GRCh38
rs761315124 Chr.16: 67539873-67539873 on Build GRCh38
rs370249978 Chr.16: 67539936-67539936 on Build GRCh38
rs201817788 Chr.16: 67539835-67539835 on Build GRCh38
rs183863939 Chr.16: 67539859-67539859 on Build GRCh38
rs560458559 Chr.16: 67540003-67540003 on Build GRCh38
rs373646662 Chr.16: 67539830-67539830 on Build GRCh38
rs911205573 Chr.16: 67539789-67539789 on Build GRCh38
rs771818233 Chr.16: 67540056-67540056 on Build GRCh38
rs753548388 Chr.16: 67540121-67540121 on Build GRCh38
rs561642006 Chr.16: 67539904-67539904 on Build GRCh38
rs763459821 Chr.16: 67540180-67540180 on Build GRCh38
rs780788822 Chr.16: 67539826-67539826 on Build GRCh38
rs749246863 Chr.16: 67540035-67540035 on Build GRCh38
rs777277666 Chr.16: 67540002-67540002 on Build GRCh38
rs767055726 Chr.16: 67539951-67539951 on Build GRCh38
rs374639736 Chr.16: 67540028-67540028 on Build GRCh38
rs778790010 Chr.16: 67539903-67539903 on Build GRCh38
rs149075175 Chr.16: 67539917-67539917 on Build GRCh38
rs770665491 Chr.16: 67540168-67540168 on Build GRCh38
rs746842326 Chr.16: 67539812-67539812 on Build GRCh38
rs750281897 Chr.16: 67539921-67539922 on Build GRCh38
rs376422701 Chr.16: 67540032-67540032 on Build GRCh38
rs747868860 Chr.16: 67539923-67539923 on Build GRCh38
rs139786497 Chr.16: 67540142-67540142 on Build GRCh38
rs765628387 Chr.16: 67540095-67540095 on Build GRCh38
rs564519127 Chr.16: 67540159-67540159 on Build GRCh38
rs772990866 Chr.16: 67540064-67540064 on Build GRCh38
rs761309186 Chr.16: 67539878-67539878 on Build GRCh38
rs781718325 Chr.16: 67540171-67540171 on Build GRCh38
rs34907450 Chr.16: 67539882-67539883 on Build GRCh38
rs939072983 Chr.16: 67540001-67540001 on Build GRCh38
rs370605093 Chr.16: 67539967-67539967 on Build GRCh38
rs778528746 Chr.16: 67540045-67540045 on Build GRCh38
rs750954506 Chr.16: 67539762-67539762 on Build GRCh38
rs907385728 Chr.16: 67539945-67539945 on Build GRCh38
rs745632140 Chr.16: 67539907-67539907 on Build GRCh38

View Full Product Details