Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67540193-67540691 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM65A
Entrez Gene ID
79567
Gene Name
family with sequence similarity 65 member A
Gene Aliases
-
UniGene
Hs.152717
Gene Chromosome Location
Chr.16: 67528814-67546788 on Build GRCh38
Transcripts
NM_001193522.1 NM_001193523.1 NM_001193524.1 NM_024519.3 XM_011523321.1 XM_011523322.1 XM_011523324.2 XM_011523325.1 XM_011523326.1 XM_017023663.1 XM_017023664.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1251942 c.609C>T p.G203G Chr.16: 67540341-67540341 on Build GRCh38
6659978 c.724C>T p.R242W Chr.16: 67540615-67540615 on Build GRCh38
6308251 c.751C>T p.Q251* Chr.16: 67540642-67540642 on Build GRCh38
5925864 c.622C>T p.R208* Chr.16: 67540354-67540354 on Build GRCh38
1679186 c.662G>A p.G221D Chr.16: 67540488-67540488 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753115937 Chr.16: 67540653-67540653 on Build GRCh38
rs994026740 Chr.16: 67540441-67540441 on Build GRCh38
rs200634890 Chr.16: 67540474-67540474 on Build GRCh38
rs777752546 Chr.16: 67540455-67540455 on Build GRCh38
rs764098878 Chr.16: 67540300-67540300 on Build GRCh38
rs749632668 Chr.16: 67540338-67540338 on Build GRCh38
rs140972331 Chr.16: 67540612-67540612 on Build GRCh38
rs773208639 Chr.16: 67540603-67540603 on Build GRCh38
rs773291011 Chr.16: 67540355-67540355 on Build GRCh38
rs944469953 Chr.16: 67540225-67540230 on Build GRCh38
rs778901617 Chr.16: 67540573-67540573 on Build GRCh38
rs965785285 Chr.16: 67540548-67540548 on Build GRCh38
rs779391264 Chr.16: 67540342-67540342 on Build GRCh38
rs771114467 Chr.16: 67540472-67540472 on Build GRCh38
rs371160500 Chr.16: 67540381-67540381 on Build GRCh38
rs761871726 Chr.16: 67540263-67540263 on Build GRCh38
rs746310107 Chr.16: 67540240-67540240 on Build GRCh38
rs780694572 Chr.16: 67540237-67540237 on Build GRCh38
rs770455471 Chr.16: 67540621-67540621 on Build GRCh38
rs752494009 Chr.16: 67540286-67540286 on Build GRCh38
rs746317359 Chr.16: 67540547-67540555 on Build GRCh38
rs757018751 Chr.16: 67540234-67540234 on Build GRCh38
rs199696724 Chr.16: 67540386-67540386 on Build GRCh38
rs767468922 Chr.16: 67540511-67540511 on Build GRCh38
rs763369738 Chr.16: 67540663-67540663 on Build GRCh38
rs754271577 Chr.16: 67540421-67540421 on Build GRCh38
rs376766206 Chr.16: 67540321-67540321 on Build GRCh38
rs777453357 Chr.16: 67540224-67540224 on Build GRCh38
rs751080715 Chr.16: 67540226-67540226 on Build GRCh38
rs747087160 Chr.16: 67540471-67540471 on Build GRCh38
rs766968628 Chr.16: 67540399-67540399 on Build GRCh38
rs928908105 Chr.16: 67540654-67540654 on Build GRCh38
rs771972828 Chr.16: 67540598-67540598 on Build GRCh38
rs747661303 Chr.16: 67540516-67540516 on Build GRCh38
rs769122522 Chr.16: 67540255-67540255 on Build GRCh38
rs747224418 Chr.16: 67540358-67540358 on Build GRCh38
rs374754244 Chr.16: 67540533-67540533 on Build GRCh38
rs757242419 Chr.16: 67540390-67540391 on Build GRCh38
rs773314808 Chr.16: 67540266-67540266 on Build GRCh38
rs748591674 Chr.16: 67540346-67540346 on Build GRCh38
rs967397231 Chr.16: 67540583-67540583 on Build GRCh38
rs749919579 Chr.16: 67540401-67540401 on Build GRCh38
rs764959997 Chr.16: 67540283-67540283 on Build GRCh38
rs774434442 Chr.16: 67540389-67540389 on Build GRCh38
rs756315543 Chr.16: 67540375-67540376 on Build GRCh38
rs780417375 Chr.16: 67540243-67540243 on Build GRCh38
rs937533615 Chr.16: 67540326-67540326 on Build GRCh38
rs151099148 Chr.16: 67540495-67540495 on Build GRCh38
rs372761052 Chr.16: 67540531-67540531 on Build GRCh38
rs201243978 Chr.16: 67540526-67540526 on Build GRCh38
rs1006645494 Chr.16: 67540514-67540514 on Build GRCh38
rs779194809 Chr.16: 67540430-67540430 on Build GRCh38
rs574495922 Chr.16: 67540532-67540532 on Build GRCh38
rs776074746 Chr.16: 67540622-67540622 on Build GRCh38
rs919015572 Chr.16: 67540577-67540577 on Build GRCh38
rs749827570 Chr.16: 67540252-67540252 on Build GRCh38
rs374445939 Chr.16: 67540276-67540276 on Build GRCh38
rs1038412350 Chr.16: 67540428-67540428 on Build GRCh38
rs771151604 Chr.16: 67540369-67540369 on Build GRCh38
rs755344344 Chr.16: 67540429-67540429 on Build GRCh38
rs746124772 Chr.16: 67540565-67540565 on Build GRCh38
rs746020180 Chr.16: 67540482-67540482 on Build GRCh38
rs368833107 Chr.16: 67540567-67540567 on Build GRCh38
rs753506741 Chr.16: 67540522-67540522 on Build GRCh38
rs537929456 Chr.16: 67540512-67540512 on Build GRCh38
rs75204333 Chr.16: 67540505-67540505 on Build GRCh38
rs573886629 Chr.16: 67540444-67540446 on Build GRCh38
rs759222975 Chr.16: 67540631-67540631 on Build GRCh38
rs201960687 Chr.16: 67540320-67540320 on Build GRCh38
rs573606224 Chr.16: 67540341-67540341 on Build GRCh38
rs369172714 Chr.16: 67540519-67540519 on Build GRCh38
rs751170704 Chr.16: 67540302-67540302 on Build GRCh38
rs752985038 Chr.16: 67540269-67540270 on Build GRCh38
rs754963653 Chr.16: 67540418-67540418 on Build GRCh38
rs749356795 Chr.16: 67540385-67540386 on Build GRCh38
rs766043766 Chr.16: 67540419-67540419 on Build GRCh38
rs367937282 Chr.16: 67540439-67540439 on Build GRCh38
rs751655133 Chr.16: 67540538-67540538 on Build GRCh38
rs765012104 Chr.16: 67540214-67540214 on Build GRCh38
rs758359380 Chr.16: 67540220-67540220 on Build GRCh38
rs201192844 Chr.16: 67540330-67540330 on Build GRCh38
rs763021708 Chr.16: 67540289-67540289 on Build GRCh38
rs774465587 Chr.16: 67540261-67540261 on Build GRCh38
rs138219494 Chr.16: 67540260-67540261 on Build GRCh38
rs748673074 Chr.16: 67540431-67540431 on Build GRCh38
rs757362192 Chr.16: 67540540-67540540 on Build GRCh38
rs565995739 Chr.16: 67540447-67540447 on Build GRCh38
rs770109930 Chr.16: 67540566-67540566 on Build GRCh38
rs777764862 Chr.16: 67540376-67540386 on Build GRCh38
rs532852286 Chr.16: 67540544-67540544 on Build GRCh38
rs780324728 Chr.16: 67540333-67540333 on Build GRCh38
rs764844466 Chr.16: 67540646-67540646 on Build GRCh38
rs550042454 Chr.16: 67540242-67540242 on Build GRCh38
rs747963585 Chr.16: 67540593-67540593 on Build GRCh38
rs896562610 Chr.16: 67540459-67540459 on Build GRCh38
rs981721414 Chr.16: 67540279-67540279 on Build GRCh38
rs777251833 Chr.16: 67540615-67540615 on Build GRCh38
rs761793745 Chr.16: 67540390-67540390 on Build GRCh38
rs772054482 Chr.16: 67540354-67540354 on Build GRCh38
rs199971815 Chr.16: 67540264-67540264 on Build GRCh38
rs752585570 Chr.16: 67540219-67540219 on Build GRCh38

View Full Product Details