Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67540437-67540946 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM65A
Entrez Gene ID
79567
Gene Name
family with sequence similarity 65 member A
Gene Aliases
-
UniGene
Hs.152717
Gene Chromosome Location
Chr.16: 67528814-67546788 on Build GRCh38
Transcripts
NM_001193522.1 NM_001193523.1 NM_001193524.1 NM_024519.3 XM_011523321.1 XM_011523322.1 XM_011523324.2 XM_011523325.1 XM_011523326.1 XM_017023663.1 XM_017023664.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6659978 c.724C>T p.R242W Chr.16: 67540615-67540615 on Build GRCh38
6308251 c.751C>T p.Q251* Chr.16: 67540642-67540642 on Build GRCh38
3377927 c.805T>C p.S269P Chr.16: 67540696-67540696 on Build GRCh38
1679186 c.662G>A p.G221D Chr.16: 67540488-67540488 on Build GRCh38
3377928 c.793T>C p.S265P Chr.16: 67540696-67540696 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753115937 Chr.16: 67540653-67540653 on Build GRCh38
rs35917599 Chr.16: 67540797-67540798 on Build GRCh38
rs546147585 Chr.16: 67540841-67540841 on Build GRCh38
rs200634890 Chr.16: 67540474-67540474 on Build GRCh38
rs776074746 Chr.16: 67540622-67540622 on Build GRCh38
rs574495922 Chr.16: 67540532-67540532 on Build GRCh38
rs919015572 Chr.16: 67540577-67540577 on Build GRCh38
rs893000247 Chr.16: 67540921-67540921 on Build GRCh38
rs560398662 Chr.16: 67540709-67540709 on Build GRCh38
rs918250330 Chr.16: 67540800-67540800 on Build GRCh38
rs757656565 Chr.16: 67540681-67540681 on Build GRCh38
rs746124772 Chr.16: 67540565-67540565 on Build GRCh38
rs546682231 Chr.16: 67540856-67540856 on Build GRCh38
rs773208639 Chr.16: 67540603-67540603 on Build GRCh38
rs140972331 Chr.16: 67540612-67540612 on Build GRCh38
rs368833107 Chr.16: 67540567-67540567 on Build GRCh38
rs746020180 Chr.16: 67540482-67540482 on Build GRCh38
rs1049782023 Chr.16: 67540904-67540904 on Build GRCh38
rs753506741 Chr.16: 67540522-67540522 on Build GRCh38
rs1038146289 Chr.16: 67540776-67540776 on Build GRCh38
rs772567030 Chr.16: 67540725-67540725 on Build GRCh38
rs746704806 Chr.16: 67540760-67540760 on Build GRCh38
rs537929456 Chr.16: 67540512-67540512 on Build GRCh38
rs368788053 Chr.16: 67540903-67540904 on Build GRCh38
rs75204333 Chr.16: 67540505-67540505 on Build GRCh38
rs759222975 Chr.16: 67540631-67540631 on Build GRCh38
rs780422097 Chr.16: 67540711-67540711 on Build GRCh38
rs778901617 Chr.16: 67540573-67540573 on Build GRCh38
rs965785285 Chr.16: 67540548-67540548 on Build GRCh38
rs1006827714 Chr.16: 67540886-67540886 on Build GRCh38
rs771114467 Chr.16: 67540472-67540472 on Build GRCh38
rs992042524 Chr.16: 67540691-67540691 on Build GRCh38
rs369172714 Chr.16: 67540519-67540519 on Build GRCh38
rs572367192 Chr.16: 67540807-67540807 on Build GRCh38
rs770455471 Chr.16: 67540621-67540621 on Build GRCh38
rs746317359 Chr.16: 67540547-67540555 on Build GRCh38
rs764298045 Chr.16: 67540674-67540674 on Build GRCh38
rs751655133 Chr.16: 67540538-67540538 on Build GRCh38
rs564459178 Chr.16: 67540881-67540881 on Build GRCh38
rs945219227 Chr.16: 67540816-67540816 on Build GRCh38
rs541612089 Chr.16: 67540675-67540675 on Build GRCh38
rs767468922 Chr.16: 67540511-67540511 on Build GRCh38
rs749406655 Chr.16: 67540746-67540746 on Build GRCh38
rs763369738 Chr.16: 67540663-67540663 on Build GRCh38
rs747087160 Chr.16: 67540471-67540471 on Build GRCh38
rs929677801 Chr.16: 67540806-67540806 on Build GRCh38
rs928908105 Chr.16: 67540654-67540654 on Build GRCh38
rs757362192 Chr.16: 67540540-67540540 on Build GRCh38
rs746712299 Chr.16: 67540748-67540757 on Build GRCh38
rs781462618 Chr.16: 67540685-67540685 on Build GRCh38
rs771972828 Chr.16: 67540598-67540598 on Build GRCh38
rs747661303 Chr.16: 67540516-67540516 on Build GRCh38
rs770109930 Chr.16: 67540566-67540566 on Build GRCh38
rs879013346 Chr.16: 67540767-67540767 on Build GRCh38
rs903966198 Chr.16: 67540860-67540860 on Build GRCh38
rs532852286 Chr.16: 67540544-67540544 on Build GRCh38
rs374754244 Chr.16: 67540533-67540533 on Build GRCh38
rs764844466 Chr.16: 67540646-67540646 on Build GRCh38
rs770468524 Chr.16: 67540875-67540875 on Build GRCh38
rs199541130 Chr.16: 67540755-67540755 on Build GRCh38
rs55659157 Chr.16: 67540747-67540747 on Build GRCh38
rs967397231 Chr.16: 67540583-67540583 on Build GRCh38
rs1039913770 Chr.16: 67540910-67540910 on Build GRCh38
rs750509807 Chr.16: 67540686-67540686 on Build GRCh38
rs747963585 Chr.16: 67540593-67540593 on Build GRCh38
rs896562610 Chr.16: 67540459-67540459 on Build GRCh38
rs151099148 Chr.16: 67540495-67540495 on Build GRCh38
rs771918099 Chr.16: 67540750-67540750 on Build GRCh38
rs777251833 Chr.16: 67540615-67540615 on Build GRCh38
rs372761052 Chr.16: 67540531-67540531 on Build GRCh38
rs775990086 Chr.16: 67540743-67540753 on Build GRCh38
rs201243978 Chr.16: 67540526-67540526 on Build GRCh38
rs1006645494 Chr.16: 67540514-67540514 on Build GRCh38

View Full Product Details