Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67663336-67663843 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENKD1
Entrez Gene ID
84080
Gene Name
enkurin domain containing 1
Gene Aliases
C16orf48, DAKV6410
UniGene
Hs.729159
Gene Chromosome Location
Chr.16: 67662947-67666725 on Build GRCh38
Transcripts
NM_032140.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5363740 c.586G>T p.G196C Chr.16: 67663814-67663814 on Build GRCh38
972549 c.832C>T p.R278C Chr.16: 67663468-67663468 on Build GRCh38
5363500 c.704A>T p.E235V Chr.16: 67663696-67663696 on Build GRCh38
7366250 c.607C>T p.R203C Chr.16: 67663793-67663793 on Build GRCh38
5093189 c.833G>A p.R278H Chr.16: 67663467-67663467 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759144896 Chr.16: 67663638-67663638 on Build GRCh38
rs766430251 Chr.16: 67663553-67663553 on Build GRCh38
rs758467673 Chr.16: 67663567-67663567 on Build GRCh38
rs771789935 Chr.16: 67663764-67663764 on Build GRCh38
rs1033532423 Chr.16: 67663465-67663465 on Build GRCh38
rs746262565 Chr.16: 67663757-67663758 on Build GRCh38
rs974054244 Chr.16: 67663576-67663576 on Build GRCh38
rs111825527 Chr.16: 67663448-67663448 on Build GRCh38
rs142623402 Chr.16: 67663455-67663455 on Build GRCh38
rs760477593 Chr.16: 67663409-67663409 on Build GRCh38
rs755353862 Chr.16: 67663630-67663630 on Build GRCh38
rs376082091 Chr.16: 67663639-67663639 on Build GRCh38
rs771877570 Chr.16: 67663697-67663697 on Build GRCh38
rs777709753 Chr.16: 67663578-67663598 on Build GRCh38
rs755981923 Chr.16: 67663656-67663656 on Build GRCh38
rs1020569460 Chr.16: 67663425-67663425 on Build GRCh38
rs750493946 Chr.16: 67663816-67663816 on Build GRCh38
rs753786845 Chr.16: 67663759-67663759 on Build GRCh38
rs970647067 Chr.16: 67663445-67663445 on Build GRCh38
rs754923936 Chr.16: 67663760-67663760 on Build GRCh38
rs897815631 Chr.16: 67663780-67663780 on Build GRCh38
rs746266622 Chr.16: 67663383-67663383 on Build GRCh38
rs777549648 Chr.16: 67663781-67663781 on Build GRCh38
rs758022748 Chr.16: 67663758-67663758 on Build GRCh38
rs763840775 Chr.16: 67663507-67663507 on Build GRCh38
rs369557268 Chr.16: 67663438-67663438 on Build GRCh38
rs75786533 Chr.16: 67663793-67663793 on Build GRCh38
rs754425398 Chr.16: 67663594-67663594 on Build GRCh38
rs972760084 Chr.16: 67663503-67663503 on Build GRCh38
rs778814794 Chr.16: 67663479-67663479 on Build GRCh38
rs1056823894 Chr.16: 67663633-67663636 on Build GRCh38
rs763488576 Chr.16: 67663450-67663450 on Build GRCh38
rs776536067 Chr.16: 67663487-67663487 on Build GRCh38
rs115927593 Chr.16: 67663688-67663688 on Build GRCh38
rs770929055 Chr.16: 67663411-67663411 on Build GRCh38
rs922654494 Chr.16: 67663703-67663703 on Build GRCh38
rs747991422 Chr.16: 67663762-67663762 on Build GRCh38
rs1016758002 Chr.16: 67663683-67663683 on Build GRCh38
rs144134135 Chr.16: 67663467-67663467 on Build GRCh38
rs765586823 Chr.16: 67663632-67663632 on Build GRCh38
rs571882950 Chr.16: 67663619-67663619 on Build GRCh38
rs752341319 Chr.16: 67663443-67663443 on Build GRCh38
rs893517115 Chr.16: 67663382-67663382 on Build GRCh38
rs913381993 Chr.16: 67663774-67663774 on Build GRCh38
rs368974361 Chr.16: 67663373-67663373 on Build GRCh38
rs962349174 Chr.16: 67663556-67663556 on Build GRCh38
rs371042455 Chr.16: 67663641-67663641 on Build GRCh38
rs534928998 Chr.16: 67663491-67663491 on Build GRCh38
rs779768082 Chr.16: 67663759-67663760 on Build GRCh38
rs373115208 Chr.16: 67663612-67663612 on Build GRCh38
rs920810005 Chr.16: 67663584-67663584 on Build GRCh38
rs761443530 Chr.16: 67663598-67663598 on Build GRCh38
rs761400328 Chr.16: 67663512-67663512 on Build GRCh38
rs746722191 Chr.16: 67663785-67663785 on Build GRCh38
rs767108756 Chr.16: 67663599-67663599 on Build GRCh38
rs1008826364 Chr.16: 67663513-67663513 on Build GRCh38
rs374065667 Chr.16: 67663708-67663708 on Build GRCh38
rs371955623 Chr.16: 67663614-67663614 on Build GRCh38
rs749115303 Chr.16: 67663589-67663589 on Build GRCh38
rs774040013 Chr.16: 67663595-67663595 on Build GRCh38
rs376060707 Chr.16: 67663545-67663545 on Build GRCh38
rs79956490 Chr.16: 67663518-67663518 on Build GRCh38
rs777188035 Chr.16: 67663709-67663709 on Build GRCh38
rs537102528 Chr.16: 67663449-67663449 on Build GRCh38
rs760037096 Chr.16: 67663792-67663792 on Build GRCh38
rs768497566 Chr.16: 67663680-67663680 on Build GRCh38
rs911502527 Chr.16: 67663747-67663747 on Build GRCh38
rs760001026 Chr.16: 67663711-67663711 on Build GRCh38
rs368877167 Chr.16: 67663387-67663387 on Build GRCh38
rs767749879 Chr.16: 67663496-67663497 on Build GRCh38
rs772512087 Chr.16: 67663760-67663760 on Build GRCh38
rs752933352 Chr.16: 67663633-67663633 on Build GRCh38
rs201476428 Chr.16: 67663724-67663724 on Build GRCh38
rs781364008 Chr.16: 67663374-67663374 on Build GRCh38
rs1027874646 Chr.16: 67663803-67663803 on Build GRCh38
rs539432820 Chr.16: 67663672-67663672 on Build GRCh38
rs757817401 Chr.16: 67663654-67663658 on Build GRCh38
rs761221282 Chr.16: 67663615-67663615 on Build GRCh38
rs776337430 Chr.16: 67663431-67663431 on Build GRCh38
rs201008671 Chr.16: 67663700-67663700 on Build GRCh38
rs115455791 Chr.16: 67663742-67663742 on Build GRCh38
rs770618888 Chr.16: 67663705-67663705 on Build GRCh38
rs776053333 Chr.16: 67663787-67663787 on Build GRCh38
rs148702803 Chr.16: 67663519-67663519 on Build GRCh38
rs900514350 Chr.16: 67663524-67663524 on Build GRCh38
rs141246944 Chr.16: 67663531-67663531 on Build GRCh38
rs149787550 Chr.16: 67663730-67663730 on Build GRCh38
rs200361187 Chr.16: 67663588-67663588 on Build GRCh38
rs768616764 Chr.16: 67663407-67663407 on Build GRCh38
rs762699410 Chr.16: 67663388-67663394 on Build GRCh38
rs940114667 Chr.16: 67663472-67663472 on Build GRCh38
rs749719131 Chr.16: 67663528-67663528 on Build GRCh38
rs777407203 Chr.16: 67663372-67663372 on Build GRCh38
rs750192334 Chr.16: 67663387-67663388 on Build GRCh38
rs764306733 Chr.16: 67663736-67663736 on Build GRCh38
rs745781474 Chr.16: 67663583-67663583 on Build GRCh38
rs777870769 Chr.16: 67663569-67663569 on Build GRCh38
rs750759275 Chr.16: 67663468-67663468 on Build GRCh38
rs774533267 Chr.16: 67663408-67663408 on Build GRCh38
rs756224421 Chr.16: 67663524-67663538 on Build GRCh38
rs938293723 Chr.16: 67663466-67663466 on Build GRCh38
rs754257906 Chr.16: 67663527-67663527 on Build GRCh38
rs769744244 Chr.16: 67663586-67663586 on Build GRCh38
rs770711938 Chr.16: 67663481-67663481 on Build GRCh38
rs766830869 Chr.16: 67663523-67663523 on Build GRCh38
rs757832516 Chr.16: 67663360-67663360 on Build GRCh38
rs772946321 Chr.16: 67663402-67663402 on Build GRCh38
rs751776684 Chr.16: 67663809-67663809 on Build GRCh38
rs377099080 Chr.16: 67663404-67663404 on Build GRCh38
rs747782761 Chr.16: 67663691-67663691 on Build GRCh38
rs769545525 Chr.16: 67663496-67663496 on Build GRCh38
rs938347686 Chr.16: 67663608-67663608 on Build GRCh38
rs373303243 Chr.16: 67663681-67663681 on Build GRCh38
rs752804327 Chr.16: 67663514-67663519 on Build GRCh38
rs369090695 Chr.16: 67663591-67663591 on Build GRCh38
rs200151482 Chr.16: 67663470-67663470 on Build GRCh38
rs763121148 Chr.16: 67663732-67663732 on Build GRCh38
rs760272639 Chr.16: 67663702-67663702 on Build GRCh38
rs549077304 Chr.16: 67663469-67663469 on Build GRCh38
rs765074393 Chr.16: 67663439-67663439 on Build GRCh38
rs748373274 Chr.16: 67663546-67663546 on Build GRCh38
rs768942945 Chr.16: 67663478-67663478 on Build GRCh38
rs773668421 Chr.16: 67663508-67663508 on Build GRCh38
rs780440789 Chr.16: 67663398-67663398 on Build GRCh38
rs757666851 Chr.16: 67663462-67663462 on Build GRCh38
rs780054404 Chr.16: 67663660-67663660 on Build GRCh38
rs754088989 Chr.16: 67663621-67663621 on Build GRCh38
rs775312149 Chr.16: 67663497-67663497 on Build GRCh38
rs779460377 Chr.16: 67663663-67663663 on Build GRCh38
rs755087145 Chr.16: 67663530-67663530 on Build GRCh38
rs762465679 Chr.16: 67663499-67663499 on Build GRCh38
rs770075014 Chr.16: 67663392-67663392 on Build GRCh38

View Full Product Details