Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67663648-67664154 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENKD1
Entrez Gene ID
84080
Gene Name
enkurin domain containing 1
Gene Aliases
C16orf48, DAKV6410
UniGene
Hs.729159
Gene Chromosome Location
Chr.16: 67662947-67666725 on Build GRCh38
Transcripts
NM_032140.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5363740 c.586G>T p.G196C Chr.16: 67663814-67663814 on Build GRCh38
5363500 c.704A>T p.E235V Chr.16: 67663696-67663696 on Build GRCh38
6425630 c.478G>T p.E160* Chr.16: 67664038-67664038 on Build GRCh38
7366250 c.607C>T p.R203C Chr.16: 67663793-67663793 on Build GRCh38
6276100 c.530C>A p.P177Q Chr.16: 67663986-67663986 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371856578 Chr.16: 67663879-67663879 on Build GRCh38
rs1027926981 Chr.16: 67664058-67664058 on Build GRCh38
rs762942318 Chr.16: 67664032-67664032 on Build GRCh38
rs756867040 Chr.16: 67663984-67663984 on Build GRCh38
rs771789935 Chr.16: 67663764-67663764 on Build GRCh38
rs766545107 Chr.16: 67663946-67663946 on Build GRCh38
rs202225875 Chr.16: 67663837-67663837 on Build GRCh38
rs567255541 Chr.16: 67663839-67663839 on Build GRCh38
rs746262565 Chr.16: 67663757-67663758 on Build GRCh38
rs200478905 Chr.16: 67664002-67664002 on Build GRCh38
rs751431968 Chr.16: 67663980-67663980 on Build GRCh38
rs368923424 Chr.16: 67663887-67663887 on Build GRCh38
rs200396232 Chr.16: 67663825-67663825 on Build GRCh38
rs752648263 Chr.16: 67663961-67663961 on Build GRCh38
rs28484968 Chr.16: 67664127-67664127 on Build GRCh38
rs751310345 Chr.16: 67664062-67664062 on Build GRCh38
rs771877570 Chr.16: 67663697-67663697 on Build GRCh38
rs750493946 Chr.16: 67663816-67663816 on Build GRCh38
rs753786845 Chr.16: 67663759-67663759 on Build GRCh38
rs373980533 Chr.16: 67664008-67664008 on Build GRCh38
rs754923936 Chr.16: 67663760-67663760 on Build GRCh38
rs897815631 Chr.16: 67663780-67663780 on Build GRCh38
rs750815774 Chr.16: 67663866-67663866 on Build GRCh38
rs777549648 Chr.16: 67663781-67663781 on Build GRCh38
rs939094574 Chr.16: 67664054-67664054 on Build GRCh38
rs758022748 Chr.16: 67663758-67663758 on Build GRCh38
rs755841094 Chr.16: 67664110-67664110 on Build GRCh38
rs894831166 Chr.16: 67664064-67664064 on Build GRCh38
rs75786533 Chr.16: 67663793-67663793 on Build GRCh38
rs770279049 Chr.16: 67663886-67663886 on Build GRCh38
rs750014672 Chr.16: 67664109-67664109 on Build GRCh38
rs961213972 Chr.16: 67663879-67663879 on Build GRCh38
rs995216467 Chr.16: 67664052-67664052 on Build GRCh38
rs780711476 Chr.16: 67663856-67663856 on Build GRCh38
rs750401873 Chr.16: 67664071-67664071 on Build GRCh38
rs779014313 Chr.16: 67664087-67664087 on Build GRCh38
rs866031813 Chr.16: 67664038-67664038 on Build GRCh38
rs1050248355 Chr.16: 67663919-67663919 on Build GRCh38
rs115927593 Chr.16: 67663688-67663688 on Build GRCh38
rs993969734 Chr.16: 67663881-67663881 on Build GRCh38
rs746572747 Chr.16: 67663825-67663826 on Build GRCh38
rs375932308 Chr.16: 67663863-67663863 on Build GRCh38
rs922654494 Chr.16: 67663703-67663703 on Build GRCh38
rs776871003 Chr.16: 67664031-67664031 on Build GRCh38
rs761854141 Chr.16: 67663981-67663981 on Build GRCh38
rs747991422 Chr.16: 67663762-67663762 on Build GRCh38
rs148941300 Chr.16: 67663994-67663994 on Build GRCh38
rs1016758002 Chr.16: 67663683-67663683 on Build GRCh38
rs913381993 Chr.16: 67663774-67663774 on Build GRCh38
rs35368900 Chr.16: 67664060-67664060 on Build GRCh38
rs779768082 Chr.16: 67663759-67663760 on Build GRCh38
rs746722191 Chr.16: 67663785-67663785 on Build GRCh38
rs940998132 Chr.16: 67664073-67664073 on Build GRCh38
rs763931231 Chr.16: 67663975-67663975 on Build GRCh38
rs549040365 Chr.16: 67663951-67663951 on Build GRCh38
rs140310273 Chr.16: 67663945-67663945 on Build GRCh38
rs374065667 Chr.16: 67663708-67663708 on Build GRCh38
rs201039452 Chr.16: 67664009-67664009 on Build GRCh38
rs1005451035 Chr.16: 67663850-67663850 on Build GRCh38
rs200352952 Chr.16: 67664055-67664055 on Build GRCh38
rs1020421551 Chr.16: 67664133-67664133 on Build GRCh38
rs760037096 Chr.16: 67663792-67663792 on Build GRCh38
rs777188035 Chr.16: 67663709-67663709 on Build GRCh38
rs768497566 Chr.16: 67663680-67663680 on Build GRCh38
rs769175759 Chr.16: 67664007-67664007 on Build GRCh38
rs778726623 Chr.16: 67663826-67663826 on Build GRCh38
rs911502527 Chr.16: 67663747-67663747 on Build GRCh38
rs760001026 Chr.16: 67663711-67663711 on Build GRCh38
rs372850599 Chr.16: 67663838-67663838 on Build GRCh38
rs752738083 Chr.16: 67663830-67663830 on Build GRCh38
rs762050371 Chr.16: 67663914-67663914 on Build GRCh38
rs767413576 Chr.16: 67664089-67664089 on Build GRCh38
rs772512087 Chr.16: 67663760-67663760 on Build GRCh38
rs201476428 Chr.16: 67663724-67663724 on Build GRCh38
rs1027874646 Chr.16: 67663803-67663803 on Build GRCh38
rs539432820 Chr.16: 67663672-67663672 on Build GRCh38
rs768272937 Chr.16: 67663887-67663887 on Build GRCh38
rs112756488 Chr.16: 67663878-67663879 on Build GRCh38
rs201008671 Chr.16: 67663700-67663700 on Build GRCh38
rs3833057 Chr.16: 67663886-67663886 on Build GRCh38
rs115455791 Chr.16: 67663742-67663742 on Build GRCh38
rs780723103 Chr.16: 67663986-67663986 on Build GRCh38
rs957881942 Chr.16: 67663918-67663918 on Build GRCh38
rs143645135 Chr.16: 67664019-67664019 on Build GRCh38
rs770618888 Chr.16: 67663705-67663705 on Build GRCh38
rs776053333 Chr.16: 67663787-67663787 on Build GRCh38
rs555422369 Chr.16: 67664128-67664128 on Build GRCh38
rs557367249 Chr.16: 67663879-67663880 on Build GRCh38
rs368355254 Chr.16: 67664104-67664104 on Build GRCh38
rs775985524 Chr.16: 67663886-67663887 on Build GRCh38
rs149787550 Chr.16: 67663730-67663730 on Build GRCh38
rs749775704 Chr.16: 67664003-67664003 on Build GRCh38
rs537793832 Chr.16: 67663949-67663949 on Build GRCh38
rs560723939 Chr.16: 67663845-67663845 on Build GRCh38
rs959807552 Chr.16: 67663849-67663849 on Build GRCh38
rs758457952 Chr.16: 67663963-67663963 on Build GRCh38
rs748469532 Chr.16: 67664015-67664015 on Build GRCh38
rs764306733 Chr.16: 67663736-67663736 on Build GRCh38
rs756882411 Chr.16: 67663855-67663855 on Build GRCh38
rs571907003 Chr.16: 67664016-67664016 on Build GRCh38
rs981534223 Chr.16: 67663998-67663998 on Build GRCh38
rs766541463 Chr.16: 67664026-67664026 on Build GRCh38
rs528670686 Chr.16: 67663995-67663995 on Build GRCh38
rs769758516 Chr.16: 67664082-67664083 on Build GRCh38
rs768979251 Chr.16: 67663894-67663894 on Build GRCh38
rs377698422 Chr.16: 67664065-67664065 on Build GRCh38
rs774749832 Chr.16: 67663911-67663911 on Build GRCh38
rs751776684 Chr.16: 67663809-67663809 on Build GRCh38
rs747782761 Chr.16: 67663691-67663691 on Build GRCh38
rs760995389 Chr.16: 67664020-67664020 on Build GRCh38
rs143646110 Chr.16: 67663883-67663883 on Build GRCh38
rs373303243 Chr.16: 67663681-67663681 on Build GRCh38
rs763121148 Chr.16: 67663732-67663732 on Build GRCh38
rs760272639 Chr.16: 67663702-67663702 on Build GRCh38
rs756445330 Chr.16: 67663885-67663885 on Build GRCh38
rs955439345 Chr.16: 67663884-67663884 on Build GRCh38
rs1038299458 Chr.16: 67664079-67664079 on Build GRCh38
rs990435497 Chr.16: 67663922-67663922 on Build GRCh38
rs182223360 Chr.16: 67663882-67663882 on Build GRCh38

View Full Product Details