Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67674817-67675314 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GFOD2
Entrez Gene ID
81577
Gene Name
glucose-fructose oxidoreductase domain containing 2
Gene Aliases
-
UniGene
Hs.307084
Gene Chromosome Location
Chr.16: 67674533-67719370 on Build GRCh38
Transcripts
NM_030819.3 NR_027398.1 XM_006721288.3 XM_017023738.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4061953 c.1036A>G p.S346G Chr.16: 67675277-67675277 on Build GRCh38
4061952 c.1144C>T p.R382W Chr.16: 67675169-67675169 on Build GRCh38
1609530 c.1116C>T p.T372T Chr.16: 67675197-67675197 on Build GRCh38
5479662 c.1011C>T p.S337S Chr.16: 67675302-67675302 on Build GRCh38
5933445 c.1070C>T p.S357F Chr.16: 67675243-67675243 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372735909 Chr.16: 67675115-67675115 on Build GRCh38
rs958098738 Chr.16: 67675160-67675160 on Build GRCh38
rs749564867 Chr.16: 67675158-67675158 on Build GRCh38
rs746912827 Chr.16: 67675153-67675153 on Build GRCh38
rs771890577 Chr.16: 67675203-67675203 on Build GRCh38
rs772842221 Chr.16: 67675205-67675205 on Build GRCh38
rs1055914697 Chr.16: 67674991-67674991 on Build GRCh38
rs914148210 Chr.16: 67674929-67674929 on Build GRCh38
rs953445878 Chr.16: 67674909-67674909 on Build GRCh38
rs190910234 Chr.16: 67675195-67675195 on Build GRCh38
rs559084882 Chr.16: 67674992-67674992 on Build GRCh38
rs542481975 Chr.16: 67675048-67675048 on Build GRCh38
rs201012397 Chr.16: 67675296-67675296 on Build GRCh38
rs1030140969 Chr.16: 67675064-67675064 on Build GRCh38
rs868441305 Chr.16: 67674958-67674958 on Build GRCh38
rs995564540 Chr.16: 67675175-67675175 on Build GRCh38
rs986598441 Chr.16: 67674910-67674910 on Build GRCh38
rs1022886329 Chr.16: 67674912-67674912 on Build GRCh38
rs748147787 Chr.16: 67675251-67675251 on Build GRCh38
rs768505813 Chr.16: 67675191-67675191 on Build GRCh38
rs772899473 Chr.16: 67675120-67675120 on Build GRCh38
rs982330251 Chr.16: 67675192-67675192 on Build GRCh38
rs200247678 Chr.16: 67675169-67675169 on Build GRCh38
rs113170495 Chr.16: 67675174-67675174 on Build GRCh38
rs897735785 Chr.16: 67675029-67675029 on Build GRCh38
rs764611800 Chr.16: 67675240-67675240 on Build GRCh38
rs373575636 Chr.16: 67675212-67675212 on Build GRCh38
rs560441468 Chr.16: 67675071-67675071 on Build GRCh38
rs1050989422 Chr.16: 67675074-67675074 on Build GRCh38
rs548207678 Chr.16: 67674943-67674943 on Build GRCh38
rs12449157 Chr.16: 67674994-67674994 on Build GRCh38
rs754353968 Chr.16: 67675147-67675147 on Build GRCh38
rs777556002 Chr.16: 67675253-67675253 on Build GRCh38
rs749180448 Chr.16: 67675184-67675184 on Build GRCh38
rs751748640 Chr.16: 67675241-67675241 on Build GRCh38
rs202206587 Chr.16: 67675131-67675131 on Build GRCh38
rs912419025 Chr.16: 67675077-67675077 on Build GRCh38
rs781291314 Chr.16: 67675244-67675244 on Build GRCh38
rs754653511 Chr.16: 67674914-67674914 on Build GRCh38
rs201851129 Chr.16: 67675206-67675206 on Build GRCh38
rs151146759 Chr.16: 67675282-67675282 on Build GRCh38
rs768896072 Chr.16: 67675168-67675168 on Build GRCh38
rs747914627 Chr.16: 67675202-67675202 on Build GRCh38
rs532200147 Chr.16: 67674919-67674919 on Build GRCh38
rs184046652 Chr.16: 67674939-67674939 on Build GRCh38
rs753300498 Chr.16: 67675226-67675226 on Build GRCh38
rs113387693 Chr.16: 67674844-67674844 on Build GRCh38
rs1016074260 Chr.16: 67674861-67674861 on Build GRCh38
rs949559779 Chr.16: 67674961-67674961 on Build GRCh38
rs566848470 Chr.16: 67674954-67674954 on Build GRCh38
rs564805607 Chr.16: 67675295-67675295 on Build GRCh38
rs765867545 Chr.16: 67675145-67675145 on Build GRCh38
rs558969157 Chr.16: 67675112-67675112 on Build GRCh38
rs760042816 Chr.16: 67675218-67675218 on Build GRCh38
rs753490040 Chr.16: 67674847-67674847 on Build GRCh38
rs770740515 Chr.16: 67675219-67675220 on Build GRCh38
rs975882277 Chr.16: 67674845-67674845 on Build GRCh38
rs761655859 Chr.16: 67675213-67675213 on Build GRCh38
rs767371058 Chr.16: 67675247-67675247 on Build GRCh38
rs191760265 Chr.16: 67674935-67674935 on Build GRCh38
rs897474299 Chr.16: 67675255-67675255 on Build GRCh38
rs758886337 Chr.16: 67675163-67675163 on Build GRCh38
rs147248628 Chr.16: 67675152-67675152 on Build GRCh38
rs769822147 Chr.16: 67675089-67675089 on Build GRCh38
rs142050875 Chr.16: 67675242-67675242 on Build GRCh38
rs745949154 Chr.16: 67675073-67675073 on Build GRCh38
rs905336292 Chr.16: 67674978-67674978 on Build GRCh38
rs771837402 Chr.16: 67675252-67675252 on Build GRCh38
rs763406154 Chr.16: 67675227-67675227 on Build GRCh38
rs939865962 Chr.16: 67675093-67675093 on Build GRCh38
rs187455264 Chr.16: 67675013-67675013 on Build GRCh38
rs760198233 Chr.16: 67675125-67675125 on Build GRCh38
rs537956509 Chr.16: 67675012-67675012 on Build GRCh38
rs1030404286 Chr.16: 67674858-67674858 on Build GRCh38
rs745342956 Chr.16: 67675275-67675275 on Build GRCh38
rs756235582 Chr.16: 67675246-67675246 on Build GRCh38
rs761413494 Chr.16: 67675109-67675109 on Build GRCh38
rs568964703 Chr.16: 67674937-67674937 on Build GRCh38
rs534218494 Chr.16: 67674988-67674988 on Build GRCh38
rs769496087 Chr.16: 67675283-67675283 on Build GRCh38
rs759968429 Chr.16: 67675150-67675150 on Build GRCh38
rs1051597653 Chr.16: 67675291-67675291 on Build GRCh38
rs936634875 Chr.16: 67674920-67674920 on Build GRCh38
rs746635131 Chr.16: 67675256-67675256 on Build GRCh38
rs765821476 Chr.16: 67675221-67675221 on Build GRCh38
rs1046582337 Chr.16: 67674967-67674967 on Build GRCh38
rs542016152 Chr.16: 67674888-67674888 on Build GRCh38
rs750489849 Chr.16: 67675245-67675245 on Build GRCh38
rs776398926 Chr.16: 67675274-67675274 on Build GRCh38
rs778110641 Chr.16: 67675167-67675167 on Build GRCh38
rs45616637 Chr.16: 67674928-67674928 on Build GRCh38
rs770513696 Chr.16: 67675273-67675273 on Build GRCh38
rs142847654 Chr.16: 67675041-67675041 on Build GRCh38
rs781146345 Chr.16: 67675177-67675177 on Build GRCh38

View Full Product Details