Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67831138-67831641 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPT
Entrez Gene ID
80152
Gene Name
centromere protein T
Gene Aliases
C16orf56, CENP-T
UniGene
Hs.288382
Gene Chromosome Location
Chr.16: 67828157-67847458 on Build GRCh38
Transcripts
NM_025082.3 XM_006721277.2 XM_011523344.1 XM_011523345.1 XM_017023712.1 XM_017023713.1 XM_017023714.1 XM_017023715.1 XM_017023716.1 XM_017023717.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6008175 c.391C>T p.P131S Chr.16: 67831219-67831219 on Build GRCh38
1609534 c.670C>T p.R224W Chr.16: 67831249-67831249 on Build GRCh38
4061988 c.625C>T p.R209C Chr.16: 67831294-67831294 on Build GRCh38
5983804 c.220C>A p.Q74K Chr.16: 67831607-67831607 on Build GRCh38
6626699 c.371G>T p.R124L Chr.16: 67831239-67831239 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747011534 Chr.16: 67831357-67831357 on Build GRCh38
rs757441860 Chr.16: 67831248-67831248 on Build GRCh38
rs116478426 Chr.16: 67831162-67831162 on Build GRCh38
rs1047082915 Chr.16: 67831212-67831212 on Build GRCh38
rs1041135051 Chr.16: 67831524-67831524 on Build GRCh38
rs1028931361 Chr.16: 67831281-67831281 on Build GRCh38
rs758338398 Chr.16: 67831269-67831269 on Build GRCh38
rs762559337 Chr.16: 67831309-67831309 on Build GRCh38
rs376945315 Chr.16: 67831541-67831541 on Build GRCh38
rs374376997 Chr.16: 67831249-67831249 on Build GRCh38
rs923641449 Chr.16: 67831338-67831338 on Build GRCh38
rs776160973 Chr.16: 67831213-67831213 on Build GRCh38
rs997486445 Chr.16: 67831544-67831544 on Build GRCh38
rs769411999 Chr.16: 67831289-67831289 on Build GRCh38
rs777605471 Chr.16: 67831350-67831350 on Build GRCh38
rs8056617 Chr.16: 67831596-67831596 on Build GRCh38
rs781123654 Chr.16: 67831583-67831583 on Build GRCh38
rs748074207 Chr.16: 67831268-67831268 on Build GRCh38
rs758285395 Chr.16: 67831336-67831336 on Build GRCh38
rs772950169 Chr.16: 67831178-67831178 on Build GRCh38
rs774689384 Chr.16: 67831531-67831531 on Build GRCh38
rs529898028 Chr.16: 67831498-67831498 on Build GRCh38
rs766381827 Chr.16: 67831332-67831332 on Build GRCh38
rs781095735 Chr.16: 67831380-67831380 on Build GRCh38
rs373811070 Chr.16: 67831528-67831528 on Build GRCh38
rs764883695 Chr.16: 67831225-67831225 on Build GRCh38
rs750254179 Chr.16: 67831616-67831616 on Build GRCh38
rs369461786 Chr.16: 67831319-67831319 on Build GRCh38
rs763607981 Chr.16: 67831231-67831231 on Build GRCh38
rs370602284 Chr.16: 67831613-67831613 on Build GRCh38
rs879242757 Chr.16: 67831200-67831200 on Build GRCh38
rs1047046407 Chr.16: 67831431-67831431 on Build GRCh38
rs944084305 Chr.16: 67831452-67831452 on Build GRCh38
rs776120648 Chr.16: 67831279-67831279 on Build GRCh38
rs139395964 Chr.16: 67831176-67831176 on Build GRCh38
rs759152872 Chr.16: 67831219-67831219 on Build GRCh38
rs762431803 Chr.16: 67831532-67831532 on Build GRCh38
rs770507082 Chr.16: 67831198-67831198 on Build GRCh38
rs751442028 Chr.16: 67831599-67831599 on Build GRCh38
rs753929412 Chr.16: 67831335-67831335 on Build GRCh38
rs767964076 Chr.16: 67831311-67831311 on Build GRCh38
rs377663014 Chr.16: 67831255-67831255 on Build GRCh38
rs983484970 Chr.16: 67831570-67831570 on Build GRCh38
rs760831431 Chr.16: 67831554-67831554 on Build GRCh38
rs375826833 Chr.16: 67831169-67831169 on Build GRCh38
rs748154725 Chr.16: 67831332-67831332 on Build GRCh38
rs941224431 Chr.16: 67831265-67831265 on Build GRCh38
rs757282392 Chr.16: 67831608-67831608 on Build GRCh38
rs764520926 Chr.16: 67831294-67831294 on Build GRCh38
rs560392321 Chr.16: 67831218-67831218 on Build GRCh38
rs866348769 Chr.16: 67831222-67831222 on Build GRCh38
rs780167096 Chr.16: 67831264-67831264 on Build GRCh38
rs962513990 Chr.16: 67831605-67831605 on Build GRCh38
rs956652139 Chr.16: 67831375-67831375 on Build GRCh38
rs750631410 Chr.16: 67831314-67831314 on Build GRCh38
rs748827853 Chr.16: 67831406-67831406 on Build GRCh38
rs371104081 Chr.16: 67831563-67831563 on Build GRCh38
rs763483728 Chr.16: 67831293-67831293 on Build GRCh38
rs746805575 Chr.16: 67831275-67831275 on Build GRCh38
rs8052287 Chr.16: 67831170-67831170 on Build GRCh38
rs995480853 Chr.16: 67831171-67831171 on Build GRCh38
rs1030765295 Chr.16: 67831557-67831557 on Build GRCh38
rs371547144 Chr.16: 67831167-67831167 on Build GRCh38
rs774727424 Chr.16: 67831297-67831297 on Build GRCh38
rs150039991 Chr.16: 67831361-67831361 on Build GRCh38
rs1011792369 Chr.16: 67831441-67831441 on Build GRCh38
rs1008348261 Chr.16: 67831239-67831239 on Build GRCh38
rs756984464 Chr.16: 67831367-67831367 on Build GRCh38
rs756154039 Chr.16: 67831258-67831258 on Build GRCh38
rs777567442 Chr.16: 67831272-67831272 on Build GRCh38
rs998620367 Chr.16: 67831401-67831401 on Build GRCh38
rs751872005 Chr.16: 67831245-67831245 on Build GRCh38
rs374147555 Chr.16: 67831195-67831195 on Build GRCh38
rs775204811 Chr.16: 67831292-67831292 on Build GRCh38
rs887141351 Chr.16: 67831434-67831434 on Build GRCh38
rs8052191 Chr.16: 67831384-67831384 on Build GRCh38
rs764546512 Chr.16: 67831241-67831241 on Build GRCh38
rs773571241 Chr.16: 67831552-67831552 on Build GRCh38
rs113675768 Chr.16: 67831413-67831413 on Build GRCh38
rs920166787 Chr.16: 67831410-67831410 on Build GRCh38
rs918040235 Chr.16: 67831394-67831394 on Build GRCh38
rs200745931 Chr.16: 67831404-67831404 on Build GRCh38
rs762115986 Chr.16: 67831300-67831300 on Build GRCh38
rs776114944 Chr.16: 67831230-67831230 on Build GRCh38
rs370743685 Chr.16: 67831403-67831403 on Build GRCh38
rs770581188 Chr.16: 67831278-67831278 on Build GRCh38

View Full Product Details