Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67831652-67832162 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPT
Entrez Gene ID
80152
Gene Name
centromere protein T
Gene Aliases
C16orf56, CENP-T
UniGene
Hs.288382
Gene Chromosome Location
Chr.16: 67828157-67847458 on Build GRCh38
Transcripts
NM_025082.3 XM_006721277.2 XM_011523344.1 XM_017023712.1 XM_017023713.1 XM_017023714.1 XM_017023715.1 XM_017023716.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6025761 c.68G>A p.S23N Chr.16: 67832021-67832021 on Build GRCh38
6025760 c.377G>A p.S126N Chr.16: 67832021-67832021 on Build GRCh38
6002360 c.9G>A p.E3E Chr.16: 67832080-67832080 on Build GRCh38
4129223 c.303C>G p.S101S Chr.16: 67832095-67832095 on Build GRCh38
1679191 c.523G>T p.E175* Chr.16: 67831754-67831754 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749517327 Chr.16: 67832015-67832015 on Build GRCh38
rs764975014 Chr.16: 67831885-67831885 on Build GRCh38
rs746006421 Chr.16: 67831858-67831858 on Build GRCh38
rs750201233 Chr.16: 67831778-67831778 on Build GRCh38
rs1016238405 Chr.16: 67832127-67832127 on Build GRCh38
rs764309153 Chr.16: 67831862-67831862 on Build GRCh38
rs903696623 Chr.16: 67832053-67832053 on Build GRCh38
rs762530607 Chr.16: 67832035-67832035 on Build GRCh38
rs144356336 Chr.16: 67831718-67831718 on Build GRCh38
rs760752826 Chr.16: 67831866-67831866 on Build GRCh38
rs780043405 Chr.16: 67831787-67831787 on Build GRCh38
rs775380845 Chr.16: 67831861-67831861 on Build GRCh38
rs200384622 Chr.16: 67831770-67831770 on Build GRCh38
rs191010035 Chr.16: 67831927-67831927 on Build GRCh38
rs969948769 Chr.16: 67831982-67831982 on Build GRCh38
rs375155332 Chr.16: 67831719-67831719 on Build GRCh38
rs535231412 Chr.16: 67831820-67831820 on Build GRCh38
rs778229845 Chr.16: 67831814-67831814 on Build GRCh38
rs748536924 Chr.16: 67831715-67831715 on Build GRCh38
rs759013748 Chr.16: 67832030-67832030 on Build GRCh38
rs761062377 Chr.16: 67831726-67831726 on Build GRCh38
rs749520828 Chr.16: 67831915-67831915 on Build GRCh38
rs1052780187 Chr.16: 67832016-67832016 on Build GRCh38
rs572614065 Chr.16: 67831908-67831908 on Build GRCh38
rs756547928 Chr.16: 67831764-67831770 on Build GRCh38
rs752335935 Chr.16: 67831887-67831887 on Build GRCh38
rs566246287 Chr.16: 67831767-67831767 on Build GRCh38
rs778225207 Chr.16: 67831851-67831851 on Build GRCh38
rs755374051 Chr.16: 67832119-67832119 on Build GRCh38
rs187620725 Chr.16: 67832037-67832037 on Build GRCh38
rs766211190 Chr.16: 67831870-67831870 on Build GRCh38
rs765757879 Chr.16: 67832121-67832121 on Build GRCh38
rs753681583 Chr.16: 67831872-67831872 on Build GRCh38
rs112697867 Chr.16: 67831965-67831965 on Build GRCh38
rs773260273 Chr.16: 67831865-67831865 on Build GRCh38
rs558638104 Chr.16: 67831679-67831679 on Build GRCh38
rs372047486 Chr.16: 67831890-67831890 on Build GRCh38
rs969575353 Chr.16: 67831863-67831863 on Build GRCh38
rs372795270 Chr.16: 67831963-67831963 on Build GRCh38
rs760632199 Chr.16: 67831935-67831935 on Build GRCh38
rs769637001 Chr.16: 67832078-67832078 on Build GRCh38
rs773967973 Chr.16: 67832100-67832100 on Build GRCh38
rs774840496 Chr.16: 67832095-67832095 on Build GRCh38
rs771362580 Chr.16: 67831929-67831929 on Build GRCh38
rs778386288 Chr.16: 67831796-67831797 on Build GRCh38
rs771666658 Chr.16: 67832029-67832030 on Build GRCh38
rs761279209 Chr.16: 67832040-67832040 on Build GRCh38
rs143748923 Chr.16: 67831986-67831986 on Build GRCh38
rs554491310 Chr.16: 67831893-67831893 on Build GRCh38
rs766983092 Chr.16: 67832041-67832041 on Build GRCh38
rs183190026 Chr.16: 67832021-67832021 on Build GRCh38
rs765224013 Chr.16: 67831745-67831745 on Build GRCh38
rs756645512 Chr.16: 67832048-67832048 on Build GRCh38
rs780339159 Chr.16: 67832013-67832013 on Build GRCh38
rs750927777 Chr.16: 67832047-67832047 on Build GRCh38
rs146813402 Chr.16: 67832025-67832025 on Build GRCh38
rs749694330 Chr.16: 67831856-67831857 on Build GRCh38
rs11558536 Chr.16: 67831845-67831845 on Build GRCh38
rs753419395 Chr.16: 67831803-67831803 on Build GRCh38
rs201330281 Chr.16: 67832038-67832038 on Build GRCh38
rs759863422 Chr.16: 67831740-67831740 on Build GRCh38
rs778711412 Chr.16: 67831972-67831975 on Build GRCh38
rs202192558 Chr.16: 67832017-67832017 on Build GRCh38
rs928576455 Chr.16: 67831995-67831995 on Build GRCh38
rs556225780 Chr.16: 67831786-67831786 on Build GRCh38
rs185855516 Chr.16: 67831704-67831704 on Build GRCh38
rs768476304 Chr.16: 67832096-67832096 on Build GRCh38
rs376460419 Chr.16: 67832005-67832005 on Build GRCh38
rs747717633 Chr.16: 67831825-67831825 on Build GRCh38
rs963435382 Chr.16: 67831948-67831950 on Build GRCh38
rs534238356 Chr.16: 67831678-67831678 on Build GRCh38
rs200881668 Chr.16: 67832118-67832118 on Build GRCh38
rs749906158 Chr.16: 67832105-67832105 on Build GRCh38
rs201102788 Chr.16: 67831924-67831924 on Build GRCh38
rs8057789 Chr.16: 67831993-67831993 on Build GRCh38
rs752967024 Chr.16: 67832125-67832125 on Build GRCh38
rs570946463 Chr.16: 67831698-67831698 on Build GRCh38
rs764993047 Chr.16: 67831964-67831964 on Build GRCh38
rs11558535 Chr.16: 67831859-67831859 on Build GRCh38
rs367603713 Chr.16: 67831920-67831920 on Build GRCh38
rs12102580 Chr.16: 67832054-67832054 on Build GRCh38
rs776816049 Chr.16: 67831732-67831732 on Build GRCh38
rs757884730 Chr.16: 67831889-67831889 on Build GRCh38
rs758793161 Chr.16: 67831850-67831850 on Build GRCh38
rs762764659 Chr.16: 67832086-67832086 on Build GRCh38
rs754540431 Chr.16: 67831874-67831874 on Build GRCh38
rs748432943 Chr.16: 67832072-67832072 on Build GRCh38
rs766442339 Chr.16: 67831939-67831939 on Build GRCh38
rs746263011 Chr.16: 67831902-67831902 on Build GRCh38
rs542018475 Chr.16: 67831838-67831838 on Build GRCh38
rs776251728 Chr.16: 67831962-67831962 on Build GRCh38
rs769331079 Chr.16: 67832130-67832130 on Build GRCh38
rs562051979 Chr.16: 67831857-67831857 on Build GRCh38
rs191893158 Chr.16: 67832010-67832010 on Build GRCh38
rs771328291 Chr.16: 67831728-67831728 on Build GRCh38
rs201699312 Chr.16: 67832106-67832106 on Build GRCh38
rs745499844 Chr.16: 67831977-67831978 on Build GRCh38
rs754277492 Chr.16: 67832058-67832058 on Build GRCh38
rs760885247 Chr.16: 67831724-67831724 on Build GRCh38
rs766827504 Chr.16: 67832115-67832115 on Build GRCh38
rs934338833 Chr.16: 67831753-67831753 on Build GRCh38
rs973911359 Chr.16: 67831950-67831950 on Build GRCh38
rs778553356 Chr.16: 67831713-67831713 on Build GRCh38
rs772181925 Chr.16: 67832075-67832075 on Build GRCh38
rs768592676 Chr.16: 67831948-67831948 on Build GRCh38
rs769630043 Chr.16: 67832033-67832033 on Build GRCh38
rs747514427 Chr.16: 67831891-67831891 on Build GRCh38
rs747085598 Chr.16: 67832138-67832138 on Build GRCh38
rs767048036 Chr.16: 67832103-67832103 on Build GRCh38
rs200046488 Chr.16: 67831806-67831806 on Build GRCh38
rs758117492 Chr.16: 67831979-67831979 on Build GRCh38
rs777855520 Chr.16: 67832133-67832133 on Build GRCh38
rs757708857 Chr.16: 67832001-67832001 on Build GRCh38
rs928278410 Chr.16: 67831735-67831739 on Build GRCh38
rs779247083 Chr.16: 67832068-67832068 on Build GRCh38
rs375024438 Chr.16: 67831900-67831900 on Build GRCh38
rs755212510 Chr.16: 67832061-67832061 on Build GRCh38
rs371678424 Chr.16: 67831856-67831856 on Build GRCh38
rs11558533 Chr.16: 67832034-67832034 on Build GRCh38

View Full Product Details