Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67989960-67990454 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPEP2
Entrez Gene ID
64174
Gene Name
dipeptidase 2
Gene Aliases
MBD2
UniGene
Hs.372633
Gene Chromosome Location
Chr.16: 67987390-68000635 on Build GRCh38
Transcripts
NM_001324159.1 NM_022355.3 NR_136706.1 XM_005256090.4 XM_011523266.1 XM_011523268.2 XM_011523271.2 XM_011523272.2 XM_011523273.2 XM_011523274.2 XM_017023541.1 XM_017023542.1 XM_017023543.1 XM_017023544.1 XM_017023545.1 XM_017023546.1 XM_017023547.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6364237 c.989T>C p.V330A Chr.16: 67990052-67990052 on Build GRCh38
1379052 c.927C>T p.V309V Chr.16: 67990114-67990114 on Build GRCh38
7210759 c.977A>G p.N326S Chr.16: 67990064-67990064 on Build GRCh38
3421100 c.944C>T p.S315F Chr.16: 67990097-67990097 on Build GRCh38
5957587 c.948G>A p.M316I Chr.16: 67990093-67990093 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs964313821 Chr.16: 67990284-67990284 on Build GRCh38
rs1017236441 Chr.16: 67990275-67990275 on Build GRCh38
rs761554456 Chr.16: 67990145-67990145 on Build GRCh38
rs575456132 Chr.16: 67990123-67990123 on Build GRCh38
rs372357723 Chr.16: 67990119-67990119 on Build GRCh38
rs115340171 Chr.16: 67990170-67990170 on Build GRCh38
rs565033787 Chr.16: 67990187-67990187 on Build GRCh38
rs774514632 Chr.16: 67990032-67990032 on Build GRCh38
rs367913444 Chr.16: 67990038-67990038 on Build GRCh38
rs978314016 Chr.16: 67990414-67990414 on Build GRCh38
rs773942656 Chr.16: 67989983-67989983 on Build GRCh38
rs989639747 Chr.16: 67990300-67990300 on Build GRCh38
rs759538140 Chr.16: 67990038-67990038 on Build GRCh38
rs780818900 Chr.16: 67990085-67990085 on Build GRCh38
rs753249157 Chr.16: 67990124-67990126 on Build GRCh38
rs889715692 Chr.16: 67990318-67990318 on Build GRCh38
rs146608913 Chr.16: 67990271-67990271 on Build GRCh38
rs751211654 Chr.16: 67990081-67990081 on Build GRCh38
rs773317563 Chr.16: 67990042-67990042 on Build GRCh38
rs779298810 Chr.16: 67990113-67990113 on Build GRCh38
rs746356309 Chr.16: 67990006-67990006 on Build GRCh38
rs751440469 Chr.16: 67990142-67990142 on Build GRCh38
rs113027359 Chr.16: 67990353-67990353 on Build GRCh38
rs928237943 Chr.16: 67990069-67990069 on Build GRCh38
rs992793933 Chr.16: 67990268-67990268 on Build GRCh38
rs146119054 Chr.16: 67990082-67990082 on Build GRCh38
rs550738160 Chr.16: 67990301-67990301 on Build GRCh38
rs1026751378 Chr.16: 67990431-67990431 on Build GRCh38
rs201800946 Chr.16: 67990089-67990089 on Build GRCh38
rs775921681 Chr.16: 67990008-67990008 on Build GRCh38
rs145887676 Chr.16: 67990122-67990122 on Build GRCh38
rs765551221 Chr.16: 67990143-67990143 on Build GRCh38
rs980651675 Chr.16: 67990191-67990191 on Build GRCh38
rs770243820 Chr.16: 67990007-67990007 on Build GRCh38
rs927533103 Chr.16: 67989984-67989984 on Build GRCh38
rs62059928 Chr.16: 67990140-67990140 on Build GRCh38
rs141329738 Chr.16: 67990117-67990117 on Build GRCh38
rs368738582 Chr.16: 67990076-67990076 on Build GRCh38
rs201088768 Chr.16: 67990179-67990179 on Build GRCh38
rs140744195 Chr.16: 67990116-67990116 on Build GRCh38
rs376520005 Chr.16: 67990202-67990202 on Build GRCh38
rs368353371 Chr.16: 67990111-67990111 on Build GRCh38
rs891891700 Chr.16: 67990051-67990051 on Build GRCh38
rs767907951 Chr.16: 67990306-67990306 on Build GRCh38
rs767337674 Chr.16: 67990147-67990147 on Build GRCh38
rs370250083 Chr.16: 67990057-67990057 on Build GRCh38
rs1055153934 Chr.16: 67990120-67990120 on Build GRCh38
rs774510909 Chr.16: 67990004-67990005 on Build GRCh38
rs915037777 Chr.16: 67990309-67990309 on Build GRCh38
rs778105355 Chr.16: 67990193-67990193 on Build GRCh38
rs751666681 Chr.16: 67990230-67990230 on Build GRCh38
rs754276928 Chr.16: 67990178-67990178 on Build GRCh38
rs936536963 Chr.16: 67990307-67990307 on Build GRCh38
rs372203515 Chr.16: 67990129-67990129 on Build GRCh38
rs966856794 Chr.16: 67990404-67990404 on Build GRCh38
rs946032359 Chr.16: 67990280-67990280 on Build GRCh38
rs767211201 Chr.16: 67990114-67990114 on Build GRCh38
rs138542230 Chr.16: 67990398-67990398 on Build GRCh38
rs73596566 Chr.16: 67990031-67990031 on Build GRCh38
rs182720433 Chr.16: 67990075-67990075 on Build GRCh38
rs1008187519 Chr.16: 67990383-67990383 on Build GRCh38
rs374262446 Chr.16: 67990064-67990064 on Build GRCh38
rs753082525 Chr.16: 67990389-67990389 on Build GRCh38
rs767561904 Chr.16: 67990040-67990040 on Build GRCh38
rs994218995 Chr.16: 67990418-67990418 on Build GRCh38
rs759509901 Chr.16: 67990045-67990045 on Build GRCh38
rs200198109 Chr.16: 67990141-67990141 on Build GRCh38
rs928100287 Chr.16: 67990428-67990428 on Build GRCh38
rs780357249 Chr.16: 67990095-67990095 on Build GRCh38
rs763983825 Chr.16: 67990078-67990078 on Build GRCh38
rs750023132 Chr.16: 67990088-67990088 on Build GRCh38
rs749830324 Chr.16: 67990099-67990099 on Build GRCh38
rs756792012 Chr.16: 67990004-67990004 on Build GRCh38
rs746666950 Chr.16: 67989998-67989998 on Build GRCh38
rs372211741 Chr.16: 67990077-67990077 on Build GRCh38
rs376566235 Chr.16: 67990152-67990152 on Build GRCh38
rs1043178092 Chr.16: 67990283-67990283 on Build GRCh38
rs750105740 Chr.16: 67990149-67990149 on Build GRCh38
rs768141175 Chr.16: 67990073-67990073 on Build GRCh38
rs960244830 Chr.16: 67990235-67990235 on Build GRCh38

View Full Product Details