Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 69153493-69153991 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UTP4
Entrez Gene ID
84916
Gene Name
UTP4, small subunit processome component
Gene Aliases
CIRH1A, CIRHIN, NAIC, TEX292
UniGene
Hs.461113
Gene Chromosome Location
Chr.16: 69132596-69169034 on Build GRCh38
Transcripts
NM_001318391.1 NM_032830.2 XM_005256205.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6860803 c.1099+8delG p.? Chr.16: 69153688-69153688 on Build GRCh38
4443887 c.1036C>A p.L346I Chr.16: 69153617-69153617 on Build GRCh38
3387555 c.1057C>T p.H353Y Chr.16: 69153638-69153638 on Build GRCh38
5897111 c.1076G>T p.R359L Chr.16: 69153657-69153657 on Build GRCh38
3272106 c.1042C>A p.L348I Chr.16: 69153623-69153623 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1035916003 Chr.16: 69153704-69153704 on Build GRCh38
rs773445419 Chr.16: 69153573-69153573 on Build GRCh38
rs954342562 Chr.16: 69153777-69153777 on Build GRCh38
rs953351860 Chr.16: 69153780-69153780 on Build GRCh38
rs368750280 Chr.16: 69153675-69153675 on Build GRCh38
rs1002882497 Chr.16: 69153687-69153687 on Build GRCh38
rs1007330926 Chr.16: 69153783-69153783 on Build GRCh38
rs774862302 Chr.16: 69153554-69153554 on Build GRCh38
rs940641349 Chr.16: 69153813-69153813 on Build GRCh38
rs567757035 Chr.16: 69153859-69153859 on Build GRCh38
rs189588544 Chr.16: 69153517-69153517 on Build GRCh38
rs748756687 Chr.16: 69153669-69153669 on Build GRCh38
rs1044465919 Chr.16: 69153916-69153916 on Build GRCh38
rs1030634133 Chr.16: 69153762-69153762 on Build GRCh38
rs374553550 Chr.16: 69153821-69153821 on Build GRCh38
rs913319979 Chr.16: 69153826-69153826 on Build GRCh38
rs779769091 Chr.16: 69153752-69153752 on Build GRCh38
rs1028701206 Chr.16: 69153760-69153760 on Build GRCh38
rs761858247 Chr.16: 69153728-69153728 on Build GRCh38
rs374770632 Chr.16: 69153544-69153544 on Build GRCh38
rs184771566 Chr.16: 69153631-69153631 on Build GRCh38
rs2288040 Chr.16: 69153793-69153793 on Build GRCh38
rs887520741 Chr.16: 69153713-69153713 on Build GRCh38
rs780780392 Chr.16: 69153536-69153536 on Build GRCh38
rs997209428 Chr.16: 69153751-69153751 on Build GRCh38
rs967761289 Chr.16: 69153857-69153857 on Build GRCh38
rs527709611 Chr.16: 69153521-69153525 on Build GRCh38
rs181556648 Chr.16: 69153893-69153893 on Build GRCh38
rs766690535 Chr.16: 69153587-69153587 on Build GRCh38
rs990161898 Chr.16: 69153813-69153813 on Build GRCh38
rs755803540 Chr.16: 69153642-69153642 on Build GRCh38
rs191103964 Chr.16: 69153810-69153810 on Build GRCh38
rs771423860 Chr.16: 69153905-69153905 on Build GRCh38
rs778329470 Chr.16: 69153668-69153668 on Build GRCh38
rs752701954 Chr.16: 69153720-69153720 on Build GRCh38
rs755924666 Chr.16: 69153745-69153745 on Build GRCh38
rs778363534 Chr.16: 69153710-69153710 on Build GRCh38
rs532732531 Chr.16: 69153681-69153681 on Build GRCh38
rs181963483 Chr.16: 69153622-69153622 on Build GRCh38
rs372301646 Chr.16: 69153553-69153553 on Build GRCh38
rs372632310 Chr.16: 69153731-69153731 on Build GRCh38
rs138151655 Chr.16: 69153595-69153595 on Build GRCh38
rs754013896 Chr.16: 69153594-69153594 on Build GRCh38
rs750334379 Chr.16: 69153739-69153739 on Build GRCh38
rs1036121386 Chr.16: 69153738-69153738 on Build GRCh38
rs775029545 Chr.16: 69153550-69153550 on Build GRCh38
rs367934569 Chr.16: 69153549-69153549 on Build GRCh38
rs773679108 Chr.16: 69153679-69153679 on Build GRCh38
rs771385144 Chr.16: 69153695-69153695 on Build GRCh38
rs777003791 Chr.16: 69153696-69153696 on Build GRCh38
rs751516872 Chr.16: 69153610-69153610 on Build GRCh38
rs142180446 Chr.16: 69153951-69153951 on Build GRCh38
rs907760026 Chr.16: 69153809-69153809 on Build GRCh38
rs905824336 Chr.16: 69153676-69153676 on Build GRCh38
rs750161529 Chr.16: 69153638-69153638 on Build GRCh38
rs754713932 Chr.16: 69153804-69153804 on Build GRCh38
rs768585959 Chr.16: 69153718-69153719 on Build GRCh38
rs779710114 Chr.16: 69153649-69153649 on Build GRCh38
rs765347755 Chr.16: 69153599-69153599 on Build GRCh38
rs567689512 Chr.16: 69153865-69153865 on Build GRCh38
rs762171150 Chr.16: 69153555-69153555 on Build GRCh38
rs377096317 Chr.16: 69153706-69153706 on Build GRCh38
rs1037558947 Chr.16: 69153645-69153645 on Build GRCh38
rs907791057 Chr.16: 69153614-69153614 on Build GRCh38
rs374852903 Chr.16: 69153566-69153566 on Build GRCh38
rs35733601 Chr.16: 69153942-69153943 on Build GRCh38
rs34747774 Chr.16: 69153664-69153664 on Build GRCh38
rs761020024 Chr.16: 69153578-69153578 on Build GRCh38
rs549163190 Chr.16: 69153785-69153785 on Build GRCh38
rs116469888 Chr.16: 69153715-69153715 on Build GRCh38
rs777073323 Chr.16: 69153965-69153965 on Build GRCh38
rs779063441 Chr.16: 69153685-69153685 on Build GRCh38
rs188746466 Chr.16: 69153659-69153659 on Build GRCh38
rs547849720 Chr.16: 69153656-69153656 on Build GRCh38
rs553690827 Chr.16: 69153895-69153895 on Build GRCh38
rs200662372 Chr.16: 69153657-69153657 on Build GRCh38
rs920472964 Chr.16: 69153889-69153889 on Build GRCh38

View Full Product Details