Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 69154151-69154649 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UTP4
Entrez Gene ID
84916
Gene Name
UTP4, small subunit processome component
Gene Aliases
CIRH1A, CIRHIN, NAIC, TEX292
UniGene
Hs.461113
Gene Chromosome Location
Chr.16: 69132596-69169034 on Build GRCh38
Transcripts
NM_001318391.1 NM_032830.2 XM_005256205.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6464469 c.1104G>A p.K368K Chr.16: 69154397-69154397 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs750284461 Chr.16: 69154220-69154220 on Build GRCh38
rs563129103 Chr.16: 69154203-69154203 on Build GRCh38
rs1051859732 Chr.16: 69154286-69154286 on Build GRCh38
rs371376413 Chr.16: 69154466-69154466 on Build GRCh38
rs753617639 Chr.16: 69154420-69154420 on Build GRCh38
rs542111255 Chr.16: 69154430-69154430 on Build GRCh38
rs969183508 Chr.16: 69154235-69154235 on Build GRCh38
rs554731009 Chr.16: 69154491-69154491 on Build GRCh38
rs191124564 Chr.16: 69154198-69154198 on Build GRCh38
rs757821823 Chr.16: 69154448-69154448 on Build GRCh38
rs757871274 Chr.16: 69154440-69154440 on Build GRCh38
rs760541143 Chr.16: 69154418-69154418 on Build GRCh38
rs747527844 Chr.16: 69154553-69154553 on Build GRCh38
rs772678195 Chr.16: 69154358-69154358 on Build GRCh38
rs1042703474 Chr.16: 69154618-69154618 on Build GRCh38
rs200109466 Chr.16: 69154395-69154395 on Build GRCh38
rs980619377 Chr.16: 69154236-69154236 on Build GRCh38
rs761980560 Chr.16: 69154365-69154365 on Build GRCh38
rs751521157 Chr.16: 69154194-69154194 on Build GRCh38
rs1057440366 Chr.16: 69154262-69154262 on Build GRCh38
rs77012617 Chr.16: 69154492-69154492 on Build GRCh38
rs549246365 Chr.16: 69154175-69154175 on Build GRCh38
rs571924487 Chr.16: 69154445-69154445 on Build GRCh38
rs777267915 Chr.16: 69154444-69154444 on Build GRCh38
rs115501719 Chr.16: 69154595-69154595 on Build GRCh38
rs761681831 Chr.16: 69154205-69154205 on Build GRCh38
rs759892228 Chr.16: 69154383-69154383 on Build GRCh38
rs766281940 Chr.16: 69154507-69154507 on Build GRCh38
rs973305986 Chr.16: 69154199-69154199 on Build GRCh38
rs536681319 Chr.16: 69154482-69154482 on Build GRCh38
rs752357214 Chr.16: 69154435-69154435 on Build GRCh38
rs776454832 Chr.16: 69154508-69154508 on Build GRCh38
rs919468470 Chr.16: 69154475-69154475 on Build GRCh38
rs187936276 Chr.16: 69154463-69154463 on Build GRCh38
rs943043421 Chr.16: 69154339-69154339 on Build GRCh38
rs61737990 Chr.16: 69154452-69154452 on Build GRCh38
rs779803024 Chr.16: 69154312-69154312 on Build GRCh38
rs972287021 Chr.16: 69154250-69154250 on Build GRCh38
rs1016511587 Chr.16: 69154439-69154439 on Build GRCh38
rs528665197 Chr.16: 69154354-69154354 on Build GRCh38
rs552756963 Chr.16: 69154540-69154540 on Build GRCh38
rs915154601 Chr.16: 69154589-69154589 on Build GRCh38
rs942147447 Chr.16: 69154249-69154249 on Build GRCh38
rs201981850 Chr.16: 69154476-69154476 on Build GRCh38
rs754678886 Chr.16: 69154428-69154428 on Build GRCh38
rs913274006 Chr.16: 69154263-69154263 on Build GRCh38
rs879066538 Chr.16: 69154573-69154573 on Build GRCh38
rs2288039 Chr.16: 69154209-69154209 on Build GRCh38
rs201502851 Chr.16: 69154410-69154410 on Build GRCh38
rs373128799 Chr.16: 69154415-69154415 on Build GRCh38
rs775678795 Chr.16: 69154394-69154394 on Build GRCh38
rs35783297 Chr.16: 69154327-69154327 on Build GRCh38
rs764769648 Chr.16: 69154431-69154431 on Build GRCh38
rs112053857 Chr.16: 69154465-69154465 on Build GRCh38
rs375258733 Chr.16: 69154353-69154353 on Build GRCh38
rs776733926 Chr.16: 69154380-69154380 on Build GRCh38
rs918984095 Chr.16: 69154254-69154254 on Build GRCh38
rs766230520 Chr.16: 69154419-69154419 on Build GRCh38
rs145910976 Chr.16: 69154183-69154183 on Build GRCh38
rs183222059 Chr.16: 69154214-69154214 on Build GRCh38
rs369181053 Chr.16: 69154386-69154386 on Build GRCh38
rs371000792 Chr.16: 69154468-69154468 on Build GRCh38
rs753494696 Chr.16: 69154345-69154345 on Build GRCh38
rs138499836 Chr.16: 69154272-69154272 on Build GRCh38
rs534463609 Chr.16: 69154510-69154510 on Build GRCh38
rs771424086 Chr.16: 69154375-69154375 on Build GRCh38
rs369518930 Chr.16: 69154359-69154359 on Build GRCh38
rs754621922 Chr.16: 69154351-69154351 on Build GRCh38
rs373007792 Chr.16: 69154376-69154376 on Build GRCh38
rs760664078 Chr.16: 69154502-69154502 on Build GRCh38
rs1043468178 Chr.16: 69154572-69154572 on Build GRCh38
rs963647109 Chr.16: 69154446-69154446 on Build GRCh38
rs1014621301 Chr.16: 69154176-69154176 on Build GRCh38

View Full Product Details