Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 70250835-70251378 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOSC6
Entrez Gene ID
118460
Gene Name
exosome component 6
Gene Aliases
EAP4, MTR3, Mtr3p, hMtr3p, p11
UniGene
Hs.660633
Gene Chromosome Location
Chr.16: 70250231-70251930 on Build GRCh38
Transcripts
NM_058219.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4439548 c.815C>T p.P272L Chr.16: 70251086-70251086 on Build GRCh38
5687515 c.765G>A p.L255L Chr.16: 70251136-70251136 on Build GRCh38
6398151 c.760G>A p.V254M Chr.16: 70251141-70251141 on Build GRCh38
7332178 c.715G>A p.E239K Chr.16: 70251186-70251186 on Build GRCh38
7259445 c.810C>A p.A270A Chr.16: 70251091-70251091 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771795328 Chr.16: 70251043-70251043 on Build GRCh38
rs542140946 Chr.16: 70251211-70251211 on Build GRCh38
rs898287444 Chr.16: 70250893-70250893 on Build GRCh38
rs924009696 Chr.16: 70251004-70251004 on Build GRCh38
rs746393319 Chr.16: 70251194-70251195 on Build GRCh38
rs562892611 Chr.16: 70250868-70250868 on Build GRCh38
rs772148824 Chr.16: 70251119-70251119 on Build GRCh38
rs995311097 Chr.16: 70250896-70250896 on Build GRCh38
rs1022088055 Chr.16: 70251325-70251325 on Build GRCh38
rs771101366 Chr.16: 70251270-70251270 on Build GRCh38
rs761978743 Chr.16: 70251117-70251117 on Build GRCh38
rs1026419695 Chr.16: 70251005-70251005 on Build GRCh38
rs777228220 Chr.16: 70251157-70251157 on Build GRCh38
rs373258652 Chr.16: 70251150-70251150 on Build GRCh38
rs1047742206 Chr.16: 70251299-70251299 on Build GRCh38
rs923921443 Chr.16: 70251247-70251247 on Build GRCh38
rs751208133 Chr.16: 70251160-70251160 on Build GRCh38
rs773488876 Chr.16: 70251126-70251126 on Build GRCh38
rs1029261091 Chr.16: 70250995-70250995 on Build GRCh38
rs769808459 Chr.16: 70251181-70251181 on Build GRCh38
rs899462834 Chr.16: 70251105-70251105 on Build GRCh38
rs992201823 Chr.16: 70251262-70251262 on Build GRCh38
rs760884693 Chr.16: 70251195-70251195 on Build GRCh38
rs779799095 Chr.16: 70251251-70251251 on Build GRCh38
rs968185908 Chr.16: 70251001-70251001 on Build GRCh38
rs975360000 Chr.16: 70250971-70250971 on Build GRCh38
rs1019731071 Chr.16: 70250954-70250954 on Build GRCh38
rs867320742 Chr.16: 70251210-70251210 on Build GRCh38
rs752107411 Chr.16: 70251143-70251143 on Build GRCh38
rs779073866 Chr.16: 70250859-70250859 on Build GRCh38
rs879520262 Chr.16: 70251330-70251330 on Build GRCh38
rs758712071 Chr.16: 70251223-70251223 on Build GRCh38
rs548411175 Chr.16: 70250890-70250890 on Build GRCh38
rs531713550 Chr.16: 70251339-70251339 on Build GRCh38
rs1047341591 Chr.16: 70251173-70251173 on Build GRCh38
rs201922942 Chr.16: 70251035-70251035 on Build GRCh38
rs1007834985 Chr.16: 70250933-70250933 on Build GRCh38
rs904104986 Chr.16: 70251285-70251285 on Build GRCh38
rs376301785 Chr.16: 70251130-70251130 on Build GRCh38
rs778794234 Chr.16: 70251102-70251116 on Build GRCh38
rs756533043 Chr.16: 70251074-70251074 on Build GRCh38
rs1008431909 Chr.16: 70251097-70251097 on Build GRCh38
rs968320169 Chr.16: 70251148-70251148 on Build GRCh38
rs11859730 Chr.16: 70251199-70251199 on Build GRCh38
rs903686665 Chr.16: 70251336-70251336 on Build GRCh38
rs764254073 Chr.16: 70251060-70251060 on Build GRCh38
rs776842991 Chr.16: 70251282-70251282 on Build GRCh38
rs770176935 Chr.16: 70251304-70251304 on Build GRCh38
rs900213082 Chr.16: 70251323-70251323 on Build GRCh38
rs562829040 Chr.16: 70251197-70251197 on Build GRCh38
rs886880658 Chr.16: 70250917-70250917 on Build GRCh38
rs776710679 Chr.16: 70251056-70251056 on Build GRCh38
rs911719890 Chr.16: 70250862-70250862 on Build GRCh38
rs761913644 Chr.16: 70251070-70251070 on Build GRCh38
rs745793316 Chr.16: 70251107-70251107 on Build GRCh38
rs757035062 Chr.16: 70251165-70251165 on Build GRCh38
rs769894271 Chr.16: 70251109-70251109 on Build GRCh38
rs931263458 Chr.16: 70251225-70251225 on Build GRCh38
rs996544191 Chr.16: 70251123-70251123 on Build GRCh38
rs772206637 Chr.16: 70251188-70251188 on Build GRCh38
rs546267057 Chr.16: 70251271-70251271 on Build GRCh38
rs1031216495 Chr.16: 70251177-70251177 on Build GRCh38
rs895296969 Chr.16: 70250946-70250946 on Build GRCh38
rs930181761 Chr.16: 70251295-70251295 on Build GRCh38
rs939435225 Chr.16: 70250915-70250915 on Build GRCh38
rs1040057585 Chr.16: 70251313-70251313 on Build GRCh38
rs777362624 Chr.16: 70250937-70250937 on Build GRCh38
rs558409992 Chr.16: 70251142-70251142 on Build GRCh38
rs980818074 Chr.16: 70251220-70251220 on Build GRCh38
rs550888671 Chr.16: 70251017-70251017 on Build GRCh38
rs772859029 Chr.16: 70251047-70251047 on Build GRCh38
rs770859907 Chr.16: 70251102-70251102 on Build GRCh38
rs543834231 Chr.16: 70251182-70251182 on Build GRCh38
rs763651944 Chr.16: 70251217-70251217 on Build GRCh38
rs188117178 Chr.16: 70250906-70250906 on Build GRCh38
rs923945041 Chr.16: 70251086-70251086 on Build GRCh38
rs745709536 Chr.16: 70251175-70251175 on Build GRCh38
rs780472355 Chr.16: 70251256-70251256 on Build GRCh38
rs1056949727 Chr.16: 70251095-70251095 on Build GRCh38
rs918023476 Chr.16: 70251206-70251206 on Build GRCh38
rs138696758 Chr.16: 70251324-70251324 on Build GRCh38
rs978406322 Chr.16: 70251244-70251244 on Build GRCh38
rs572974845 Chr.16: 70251093-70251093 on Build GRCh38
rs370392673 Chr.16: 70251226-70251226 on Build GRCh38
rs746841607 Chr.16: 70251053-70251053 on Build GRCh38
rs1041168637 Chr.16: 70251253-70251253 on Build GRCh38
rs773952157 Chr.16: 70251355-70251355 on Build GRCh38
rs765596883 Chr.16: 70251205-70251205 on Build GRCh38
rs774642702 Chr.16: 70251067-70251067 on Build GRCh38
rs538281243 Chr.16: 70251002-70251002 on Build GRCh38
rs536434410 Chr.16: 70251085-70251085 on Build GRCh38
rs948211782 Chr.16: 70251111-70251111 on Build GRCh38
rs527761151 Chr.16: 70250965-70250965 on Build GRCh38
rs528661450 Chr.16: 70251329-70251329 on Build GRCh38
rs944342016 Chr.16: 70251311-70251311 on Build GRCh38
rs747112995 Chr.16: 70251265-70251265 on Build GRCh38
rs530211058 Chr.16: 70250876-70250876 on Build GRCh38
rs1038464466 Chr.16: 70251284-70251284 on Build GRCh38
rs1056602465 Chr.16: 70250935-70250935 on Build GRCh38
rs570781513 Chr.16: 70250994-70250994 on Build GRCh38
rs935438972 Chr.16: 70251012-70251012 on Build GRCh38
rs950460670 Chr.16: 70251026-70251026 on Build GRCh38
rs770734017 Chr.16: 70251055-70251055 on Build GRCh38
rs967923525 Chr.16: 70251243-70251243 on Build GRCh38
rs748109812 Chr.16: 70251187-70251187 on Build GRCh38
rs759924078 Chr.16: 70251202-70251202 on Build GRCh38
rs767833065 Chr.16: 70251072-70251072 on Build GRCh38
rs969768544 Chr.16: 70251214-70251214 on Build GRCh38
rs552396154 Chr.16: 70250988-70250988 on Build GRCh38
rs781189003 Chr.16: 70251104-70251104 on Build GRCh38
rs756560156 Chr.16: 70251031-70251031 on Build GRCh38
rs533845250 Chr.16: 70251141-70251141 on Build GRCh38
rs144728576 Chr.16: 70251232-70251232 on Build GRCh38
rs558035411 Chr.16: 70251279-70251279 on Build GRCh38
rs996904563 Chr.16: 70251268-70251268 on Build GRCh38
rs908344423 Chr.16: 70251229-70251229 on Build GRCh38
rs777512251 Chr.16: 70251094-70251094 on Build GRCh38
rs760891355 Chr.16: 70251128-70251128 on Build GRCh38
rs754938376 Chr.16: 70251260-70251260 on Build GRCh38
rs764555739 Chr.16: 70251344-70251344 on Build GRCh38
rs948007586 Chr.16: 70250872-70250872 on Build GRCh38
rs202233631 Chr.16: 70251264-70251264 on Build GRCh38
rs746173498 Chr.16: 70251300-70251300 on Build GRCh38
rs754490384 Chr.16: 70251041-70251041 on Build GRCh38
rs773224272 Chr.16: 70251191-70251191 on Build GRCh38
rs28619873 Chr.16: 70250881-70250881 on Build GRCh38
rs781105156 Chr.16: 70251171-70251171 on Build GRCh38
rs746711549 Chr.16: 70251098-70251098 on Build GRCh38
rs776930904 Chr.16: 70251200-70251200 on Build GRCh38
rs149061783 Chr.16: 70251087-70251087 on Build GRCh38
rs763064788 Chr.16: 70251057-70251057 on Build GRCh38
rs199955950 Chr.16: 70251042-70251042 on Build GRCh38
rs749237213 Chr.16: 70251254-70251254 on Build GRCh38
rs774275507 Chr.16: 70251116-70251116 on Build GRCh38
rs766805718 Chr.16: 70251079-70251079 on Build GRCh38
rs373161644 Chr.16: 70251088-70251088 on Build GRCh38
rs1031323781 Chr.16: 70251343-70251343 on Build GRCh38
rs750709481 Chr.16: 70251073-70251073 on Build GRCh38
rs868351357 Chr.16: 70251236-70251236 on Build GRCh38
rs1042778321 Chr.16: 70251276-70251276 on Build GRCh38

View Full Product Details