Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 70251235-70251728 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOSC6
Entrez Gene ID
118460
Gene Name
exosome component 6
Gene Aliases
EAP4, MTR3, Mtr3p, hMtr3p, p11
UniGene
Hs.660633
Gene Chromosome Location
Chr.16: 70250231-70251930 on Build GRCh38
Transcripts
NM_058219.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6499428 c.301C>T p.R101C Chr.16: 70251600-70251600 on Build GRCh38
5683623 c.399T>A p.A133A Chr.16: 70251502-70251502 on Build GRCh38
5687516 c.627C>A p.A209A Chr.16: 70251274-70251274 on Build GRCh38
6375097 c.664G>T p.V222L Chr.16: 70251237-70251237 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs753618366 Chr.16: 70251429-70251429 on Build GRCh38
rs935100295 Chr.16: 70251621-70251621 on Build GRCh38
rs867670680 Chr.16: 70251472-70251472 on Build GRCh38
rs570247550 Chr.16: 70251515-70251515 on Build GRCh38
rs766195473 Chr.16: 70251424-70251424 on Build GRCh38
rs778744923 Chr.16: 70251442-70251442 on Build GRCh38
rs912737642 Chr.16: 70251576-70251576 on Build GRCh38
rs1022088055 Chr.16: 70251325-70251325 on Build GRCh38
rs771101366 Chr.16: 70251270-70251270 on Build GRCh38
rs761440746 Chr.16: 70251372-70251372 on Build GRCh38
rs1020141310 Chr.16: 70251700-70251700 on Build GRCh38
rs918298675 Chr.16: 70251663-70251674 on Build GRCh38
rs529988120 Chr.16: 70251415-70251415 on Build GRCh38
rs1052665567 Chr.16: 70251623-70251623 on Build GRCh38
rs1047742206 Chr.16: 70251299-70251299 on Build GRCh38
rs138696758 Chr.16: 70251324-70251324 on Build GRCh38
rs1043426494 Chr.16: 70251612-70251612 on Build GRCh38
rs555713582 Chr.16: 70251531-70251531 on Build GRCh38
rs896304486 Chr.16: 70251627-70251627 on Build GRCh38
rs925001579 Chr.16: 70251613-70251613 on Build GRCh38
rs200071202 Chr.16: 70251497-70251497 on Build GRCh38
rs975660016 Chr.16: 70251590-70251590 on Build GRCh38
rs747021297 Chr.16: 70251471-70251471 on Build GRCh38
rs754893211 Chr.16: 70251430-70251430 on Build GRCh38
rs931624269 Chr.16: 70251610-70251610 on Build GRCh38
rs969339236 Chr.16: 70251549-70251549 on Build GRCh38
rs767212857 Chr.16: 70251408-70251408 on Build GRCh38
rs757338425 Chr.16: 70251493-70251493 on Build GRCh38
rs992201823 Chr.16: 70251262-70251262 on Build GRCh38
rs537566605 Chr.16: 70251525-70251525 on Build GRCh38
rs773952157 Chr.16: 70251355-70251355 on Build GRCh38
rs972485339 Chr.16: 70251400-70251414 on Build GRCh38
rs974071262 Chr.16: 70251561-70251561 on Build GRCh38
rs528661450 Chr.16: 70251329-70251329 on Build GRCh38
rs548191419 Chr.16: 70251422-70251422 on Build GRCh38
rs879520262 Chr.16: 70251330-70251330 on Build GRCh38
rs944342016 Chr.16: 70251311-70251311 on Build GRCh38
rs747112995 Chr.16: 70251265-70251265 on Build GRCh38
rs866196318 Chr.16: 70251659-70251659 on Build GRCh38
rs531713550 Chr.16: 70251339-70251339 on Build GRCh38
rs1038464466 Chr.16: 70251284-70251284 on Build GRCh38
rs755891883 Chr.16: 70251423-70251423 on Build GRCh38
rs921537944 Chr.16: 70251604-70251604 on Build GRCh38
rs568478509 Chr.16: 70251409-70251409 on Build GRCh38
rs904104986 Chr.16: 70251285-70251285 on Build GRCh38
rs1023544274 Chr.16: 70251544-70251544 on Build GRCh38
rs983314707 Chr.16: 70251574-70251574 on Build GRCh38
rs1026962344 Chr.16: 70251681-70251681 on Build GRCh38
rs920032321 Chr.16: 70251434-70251434 on Build GRCh38
rs558035411 Chr.16: 70251279-70251279 on Build GRCh38
rs903686665 Chr.16: 70251336-70251336 on Build GRCh38
rs996904563 Chr.16: 70251268-70251268 on Build GRCh38
rs960671206 Chr.16: 70251374-70251374 on Build GRCh38
rs558600389 Chr.16: 70251502-70251502 on Build GRCh38
rs769948694 Chr.16: 70251498-70251498 on Build GRCh38
rs776842991 Chr.16: 70251282-70251282 on Build GRCh38
rs754938376 Chr.16: 70251260-70251260 on Build GRCh38
rs775899457 Chr.16: 70251505-70251505 on Build GRCh38
rs764555739 Chr.16: 70251344-70251344 on Build GRCh38
rs867003845 Chr.16: 70251567-70251567 on Build GRCh38
rs202233631 Chr.16: 70251264-70251264 on Build GRCh38
rs944305625 Chr.16: 70251466-70251466 on Build GRCh38
rs770176935 Chr.16: 70251304-70251304 on Build GRCh38
rs912782022 Chr.16: 70251456-70251456 on Build GRCh38
rs900213082 Chr.16: 70251323-70251323 on Build GRCh38
rs746173498 Chr.16: 70251300-70251300 on Build GRCh38
rs574100322 Chr.16: 70251589-70251589 on Build GRCh38
rs969412851 Chr.16: 70251412-70251412 on Build GRCh38
rs1013879484 Chr.16: 70251654-70251654 on Build GRCh38
rs948859671 Chr.16: 70251592-70251592 on Build GRCh38
rs995430218 Chr.16: 70251665-70251666 on Build GRCh38
rs550232370 Chr.16: 70251386-70251386 on Build GRCh38
rs546267057 Chr.16: 70251271-70251271 on Build GRCh38
rs951518012 Chr.16: 70251693-70251693 on Build GRCh38
rs1004288370 Chr.16: 70251695-70251695 on Build GRCh38
rs982982435 Chr.16: 70251450-70251450 on Build GRCh38
rs1031323781 Chr.16: 70251343-70251343 on Build GRCh38
rs930181761 Chr.16: 70251295-70251295 on Build GRCh38
rs1040057585 Chr.16: 70251313-70251313 on Build GRCh38
rs900160805 Chr.16: 70251474-70251474 on Build GRCh38
rs1042778321 Chr.16: 70251276-70251276 on Build GRCh38

View Full Product Details