Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 70496181-70496681 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COG4
Entrez Gene ID
25839
Gene Name
component of oligomeric golgi complex 4
Gene Aliases
CDG2J, COD1
UniGene
Hs.208680
Gene Chromosome Location
Chr.16: 70480569-70523554 on Build GRCh38
Transcripts
NM_001195139.1 NM_015386.2 XM_011522981.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1709335 c.1520_1521CC>TT p.A507V Chr.16: 70496392-70496393 on Build GRCh38
6994584 c.1563G>T p.V521V Chr.16: 70496350-70496350 on Build GRCh38
7328933 c.1498A>C p.K500Q Chr.16: 70496415-70496415 on Build GRCh38
3937143 c.1611C>A p.G537G Chr.16: 70496302-70496302 on Build GRCh38
6632221 c.1615G>A p.E539K Chr.16: 70496298-70496298 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs986626381 Chr.16: 70496570-70496570 on Build GRCh38
rs374027719 Chr.16: 70496225-70496225 on Build GRCh38
rs377490069 Chr.16: 70496215-70496215 on Build GRCh38
rs35525850 Chr.16: 70496432-70496432 on Build GRCh38
rs115668774 Chr.16: 70496353-70496353 on Build GRCh38
rs986699941 Chr.16: 70496282-70496282 on Build GRCh38
rs779983714 Chr.16: 70496332-70496332 on Build GRCh38
rs755998441 Chr.16: 70496320-70496320 on Build GRCh38
rs376657474 Chr.16: 70496369-70496369 on Build GRCh38
rs751237267 Chr.16: 70496308-70496308 on Build GRCh38
rs529186566 Chr.16: 70496245-70496245 on Build GRCh38
rs757054202 Chr.16: 70496309-70496309 on Build GRCh38
rs767223671 Chr.16: 70496402-70496402 on Build GRCh38
rs1032393639 Chr.16: 70496538-70496538 on Build GRCh38
rs760158081 Chr.16: 70496296-70496296 on Build GRCh38
rs770348849 Chr.16: 70496473-70496473 on Build GRCh38
rs183710443 Chr.16: 70496553-70496553 on Build GRCh38
rs771255200 Chr.16: 70496281-70496281 on Build GRCh38
rs1010132702 Chr.16: 70496586-70496586 on Build GRCh38
rs975192824 Chr.16: 70496542-70496542 on Build GRCh38
rs777590106 Chr.16: 70496238-70496238 on Build GRCh38
rs777169655 Chr.16: 70496289-70496289 on Build GRCh38
rs541274302 Chr.16: 70496299-70496299 on Build GRCh38
rs781576955 Chr.16: 70496465-70496465 on Build GRCh38
rs755910468 Chr.16: 70496436-70496436 on Build GRCh38
rs893205800 Chr.16: 70496376-70496376 on Build GRCh38
rs940522796 Chr.16: 70496396-70496396 on Build GRCh38
rs147508980 Chr.16: 70496389-70496389 on Build GRCh38
rs886052258 Chr.16: 70496282-70496282 on Build GRCh38
rs1052580926 Chr.16: 70496370-70496392 on Build GRCh38
rs1025254451 Chr.16: 70496593-70496593 on Build GRCh38
rs752940850 Chr.16: 70496397-70496397 on Build GRCh38
rs981024001 Chr.16: 70496613-70496613 on Build GRCh38
rs745843280 Chr.16: 70496246-70496246 on Build GRCh38
rs748162218 Chr.16: 70496335-70496335 on Build GRCh38
rs773314806 Chr.16: 70496265-70496265 on Build GRCh38
rs1000953792 Chr.16: 70496531-70496531 on Build GRCh38
rs751308615 Chr.16: 70496239-70496239 on Build GRCh38
rs767744080 Chr.16: 70496243-70496243 on Build GRCh38
rs545837302 Chr.16: 70496266-70496266 on Build GRCh38
rs747240944 Chr.16: 70496352-70496352 on Build GRCh38
rs758545148 Chr.16: 70496336-70496336 on Build GRCh38
rs761333204 Chr.16: 70496394-70496394 on Build GRCh38
rs777750953 Chr.16: 70496454-70496454 on Build GRCh38
rs1000920120 Chr.16: 70496215-70496215 on Build GRCh38
rs763735488 Chr.16: 70496306-70496306 on Build GRCh38
rs778924738 Chr.16: 70496257-70496257 on Build GRCh38
rs766304115 Chr.16: 70496439-70496439 on Build GRCh38
rs902785478 Chr.16: 70496495-70496495 on Build GRCh38
rs746254288 Chr.16: 70496365-70496365 on Build GRCh38
rs750251710 Chr.16: 70496318-70496318 on Build GRCh38
rs759099941 Chr.16: 70496218-70496218 on Build GRCh38
rs2303793 Chr.16: 70496456-70496456 on Build GRCh38
rs1044924146 Chr.16: 70496429-70496429 on Build GRCh38
rs768691305 Chr.16: 70496253-70496253 on Build GRCh38
rs1028090974 Chr.16: 70496549-70496549 on Build GRCh38
rs971400789 Chr.16: 70496601-70496601 on Build GRCh38
rs746167747 Chr.16: 70496471-70496471 on Build GRCh38
rs934637366 Chr.16: 70496610-70496610 on Build GRCh38
rs374414938 Chr.16: 70496450-70496450 on Build GRCh38
rs951045514 Chr.16: 70496209-70496209 on Build GRCh38
rs11641683 Chr.16: 70496644-70496644 on Build GRCh38
rs371160353 Chr.16: 70496370-70496370 on Build GRCh38
rs926950737 Chr.16: 70496305-70496305 on Build GRCh38
rs924515792 Chr.16: 70496609-70496609 on Build GRCh38
rs769237568 Chr.16: 70496483-70496483 on Build GRCh38
rs187838403 Chr.16: 70496216-70496216 on Build GRCh38
rs78104579 Chr.16: 70496217-70496217 on Build GRCh38
rs199758667 Chr.16: 70496493-70496493 on Build GRCh38
rs755746795 Chr.16: 70496433-70496434 on Build GRCh38
rs767256716 Chr.16: 70496316-70496316 on Build GRCh38
rs745586634 Chr.16: 70496258-70496259 on Build GRCh38
rs190357875 Chr.16: 70496408-70496408 on Build GRCh38
rs765873830 Chr.16: 70496298-70496298 on Build GRCh38
rs889095149 Chr.16: 70496522-70496522 on Build GRCh38
rs200438500 Chr.16: 70496458-70496458 on Build GRCh38
rs368765825 Chr.16: 70496452-70496452 on Build GRCh38
rs367863540 Chr.16: 70496518-70496518 on Build GRCh38
rs778046649 Chr.16: 70496340-70496340 on Build GRCh38
rs762564242 Chr.16: 70496302-70496302 on Build GRCh38
rs774025415 Chr.16: 70496391-70496391 on Build GRCh38
rs772226017 Chr.16: 70496261-70496261 on Build GRCh38
rs758081308 Chr.16: 70496234-70496234 on Build GRCh38
rs1034643668 Chr.16: 70496206-70496206 on Build GRCh38
rs770365416 Chr.16: 70496368-70496368 on Build GRCh38
rs530493728 Chr.16: 70496491-70496491 on Build GRCh38
rs567643441 Chr.16: 70496529-70496529 on Build GRCh38
rs57301833 Chr.16: 70496481-70496481 on Build GRCh38
rs759717884 Chr.16: 70496226-70496226 on Build GRCh38
rs147242136 Chr.16: 70496532-70496532 on Build GRCh38

View Full Product Details