Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 71285568-71286066 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMTR2
Entrez Gene ID
55783
Gene Name
cap methyltransferase 2
Gene Aliases
AFT, FTSJD1, HMTr2, MTr2
UniGene
Hs.72782
Gene Chromosome Location
Chr.16: 71282300-71290178 on Build GRCh38
Transcripts
NM_001099642.1 NM_001324374.1 NM_001324377.1 NM_001324378.1 NM_001324379.1 NM_018348.5 XM_011523228.1 XM_011523229.1 XM_017023444.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6538572 c.232A>C p.S78R Chr.16: 71285689-71285689 on Build GRCh38
1519667 c.82G>A p.E28K Chr.16: 71285839-71285839 on Build GRCh38
1644626 c.344G>A p.W115* Chr.16: 71285577-71285577 on Build GRCh38
4649564 c.217G>T p.V73L Chr.16: 71285704-71285704 on Build GRCh38
5833167 c.6_7insT p.K3fs*1 Chr.16: 71285914-71285915 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200569499 Chr.16: 71285874-71285874 on Build GRCh38
rs563690149 Chr.16: 71285888-71285888 on Build GRCh38
rs754383285 Chr.16: 71285898-71285898 on Build GRCh38
rs549288798 Chr.16: 71286029-71286029 on Build GRCh38
rs750274474 Chr.16: 71285682-71285682 on Build GRCh38
rs1037133804 Chr.16: 71285876-71285876 on Build GRCh38
rs903324605 Chr.16: 71285685-71285685 on Build GRCh38
rs545597868 Chr.16: 71285878-71285878 on Build GRCh38
rs747945522 Chr.16: 71285804-71285804 on Build GRCh38
rs762200646 Chr.16: 71285771-71285771 on Build GRCh38
rs139187106 Chr.16: 71285756-71285756 on Build GRCh38
rs749038805 Chr.16: 71285717-71285717 on Build GRCh38
rs773678121 Chr.16: 71285873-71285873 on Build GRCh38
rs772545362 Chr.16: 71285696-71285697 on Build GRCh38
rs371216040 Chr.16: 71285605-71285605 on Build GRCh38
rs147189785 Chr.16: 71285839-71285839 on Build GRCh38
rs541012112 Chr.16: 71285859-71285859 on Build GRCh38
rs1019670545 Chr.16: 71286011-71286011 on Build GRCh38
rs769922175 Chr.16: 71285748-71285748 on Build GRCh38
rs768151348 Chr.16: 71285773-71285773 on Build GRCh38
rs531238347 Chr.16: 71285986-71285986 on Build GRCh38
rs749959745 Chr.16: 71285686-71285686 on Build GRCh38
rs751429535 Chr.16: 71285760-71285760 on Build GRCh38
rs866089679 Chr.16: 71285806-71285806 on Build GRCh38
rs764200259 Chr.16: 71285762-71285762 on Build GRCh38
rs953824017 Chr.16: 71285988-71285988 on Build GRCh38
rs1026838167 Chr.16: 71285867-71285867 on Build GRCh38
rs774015734 Chr.16: 71285620-71285620 on Build GRCh38
rs746303020 Chr.16: 71285607-71285607 on Build GRCh38
rs753262010 Chr.16: 71285792-71285792 on Build GRCh38
rs765446460 Chr.16: 71285638-71285638 on Build GRCh38
rs767424783 Chr.16: 71285632-71285632 on Build GRCh38
rs775688863 Chr.16: 71285830-71285830 on Build GRCh38
rs764960244 Chr.16: 71285909-71285909 on Build GRCh38
rs947167977 Chr.16: 71286007-71286007 on Build GRCh38
rs751023866 Chr.16: 71285775-71285775 on Build GRCh38
rs759568448 Chr.16: 71285637-71285637 on Build GRCh38
rs538853572 Chr.16: 71285665-71285665 on Build GRCh38
rs746784861 Chr.16: 71285958-71285958 on Build GRCh38
rs3826247 Chr.16: 71285636-71285636 on Build GRCh38
rs868480726 Chr.16: 71285734-71285734 on Build GRCh38
rs763071162 Chr.16: 71285661-71285661 on Build GRCh38
rs752904742 Chr.16: 71285654-71285654 on Build GRCh38
rs192739719 Chr.16: 71285812-71285812 on Build GRCh38
rs12599703 Chr.16: 71285987-71285987 on Build GRCh38
rs747017344 Chr.16: 71285825-71285825 on Build GRCh38
rs201357878 Chr.16: 71285856-71285856 on Build GRCh38
rs778821297 Chr.16: 71285730-71285730 on Build GRCh38
rs148074898 Chr.16: 71285678-71285678 on Build GRCh38
rs754676757 Chr.16: 71285796-71285796 on Build GRCh38
rs954107128 Chr.16: 71285627-71285627 on Build GRCh38
rs912645448 Chr.16: 71285653-71285653 on Build GRCh38
rs768166688 Chr.16: 71285724-71285724 on Build GRCh38
rs963412578 Chr.16: 71285848-71285848 on Build GRCh38
rs761853994 Chr.16: 71285622-71285622 on Build GRCh38
rs199611664 Chr.16: 71285853-71285853 on Build GRCh38
rs143775335 Chr.16: 71285783-71285783 on Build GRCh38
rs756681901 Chr.16: 71285776-71285776 on Build GRCh38
rs897056709 Chr.16: 71285892-71285892 on Build GRCh38
rs758182966 Chr.16: 71285707-71285707 on Build GRCh38
rs770744374 Chr.16: 71285826-71285826 on Build GRCh38
rs778393775 Chr.16: 71285798-71285798 on Build GRCh38
rs747893947 Chr.16: 71285735-71285735 on Build GRCh38
rs772025244 Chr.16: 71285739-71285739 on Build GRCh38
rs572908241 Chr.16: 71285765-71285765 on Build GRCh38
rs759629413 Chr.16: 71285744-71285744 on Build GRCh38
rs777533581 Chr.16: 71285823-71285823 on Build GRCh38
rs764585691 Chr.16: 71285664-71285664 on Build GRCh38
rs373868631 Chr.16: 71285957-71285957 on Build GRCh38
rs749459132 Chr.16: 71285958-71285961 on Build GRCh38
rs775715839 Chr.16: 71285752-71285758 on Build GRCh38
rs578011693 Chr.16: 71285875-71285875 on Build GRCh38
rs781088912 Chr.16: 71285864-71285864 on Build GRCh38
rs752408477 Chr.16: 71285922-71285922 on Build GRCh38
rs913453963 Chr.16: 71285687-71285687 on Build GRCh38
rs755629456 Chr.16: 71285693-71285693 on Build GRCh38
rs912474785 Chr.16: 71285974-71285974 on Build GRCh38
rs189345363 Chr.16: 71285745-71285745 on Build GRCh38
rs899580225 Chr.16: 71285640-71285640 on Build GRCh38
rs140513334 Chr.16: 71285896-71285896 on Build GRCh38
rs868735846 Chr.16: 71285901-71285901 on Build GRCh38
rs943723625 Chr.16: 71285982-71285982 on Build GRCh38
rs3096380 Chr.16: 71285743-71285743 on Build GRCh38
rs34629717 Chr.16: 71285607-71285608 on Build GRCh38
rs758044072 Chr.16: 71285946-71285946 on Build GRCh38
rs9928675 Chr.16: 71285997-71285997 on Build GRCh38
rs749052074 Chr.16: 71285594-71285594 on Build GRCh38
rs757241661 Chr.16: 71285964-71285964 on Build GRCh38

View Full Product Details