Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 71358628-71359134 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALB2
Entrez Gene ID
794
Gene Name
calbindin 2
Gene Aliases
CAB29, CAL2, CR
UniGene
Hs.106857
Gene Chromosome Location
Chr.16: 71358713-71390438 on Build GRCh38
Transcripts
NM_001740.4 NM_007088.3 NR_027910.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
704250 c.43G>A p.E15K Chr.16: 71358835-71358835 on Build GRCh38
3717054 c.3G>T p.M1I Chr.16: 71358795-71358795 on Build GRCh38
6839143 c.52G>A p.A18T Chr.16: 71358844-71358844 on Build GRCh38
4608629 c.52G>T p.A18S Chr.16: 71358844-71358844 on Build GRCh38
6079693 c.93C>G p.D31E Chr.16: 71358885-71358885 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs956010800 Chr.16: 71359106-71359106 on Build GRCh38
rs1022278376 Chr.16: 71358669-71358669 on Build GRCh38
rs777053099 Chr.16: 71358884-71358884 on Build GRCh38
rs760077786 Chr.16: 71358777-71358777 on Build GRCh38
rs200609559 Chr.16: 71359030-71359030 on Build GRCh38
rs772451161 Chr.16: 71358846-71358846 on Build GRCh38
rs1030008510 Chr.16: 71358790-71358790 on Build GRCh38
rs886732312 Chr.16: 71358865-71358865 on Build GRCh38
rs888728257 Chr.16: 71359016-71359016 on Build GRCh38
rs764722881 Chr.16: 71358780-71358780 on Build GRCh38
rs750207842 Chr.16: 71358924-71358924 on Build GRCh38
rs916972222 Chr.16: 71358723-71358723 on Build GRCh38
rs886613124 Chr.16: 71358665-71358665 on Build GRCh38
rs1046406594 Chr.16: 71358717-71358717 on Build GRCh38
rs749667135 Chr.16: 71359059-71359059 on Build GRCh38
rs747418228 Chr.16: 71358876-71358876 on Build GRCh38
rs374258247 Chr.16: 71358922-71358922 on Build GRCh38
rs1039889728 Chr.16: 71359084-71359084 on Build GRCh38
rs916693316 Chr.16: 71358664-71358664 on Build GRCh38
rs756943994 Chr.16: 71358813-71358813 on Build GRCh38
rs575232191 Chr.16: 71358994-71358994 on Build GRCh38
rs989705224 Chr.16: 71358656-71358656 on Build GRCh38
rs567635737 Chr.16: 71358666-71358666 on Build GRCh38
rs957936886 Chr.16: 71358886-71358886 on Build GRCh38
rs763750106 Chr.16: 71358907-71358907 on Build GRCh38
rs1047384868 Chr.16: 71358736-71358736 on Build GRCh38
rs762410004 Chr.16: 71358899-71358899 on Build GRCh38
rs1048009705 Chr.16: 71358864-71358864 on Build GRCh38
rs964859289 Chr.16: 71358668-71358668 on Build GRCh38
rs989341744 Chr.16: 71358890-71358890 on Build GRCh38
rs770265641 Chr.16: 71358892-71358892 on Build GRCh38
rs369277856 Chr.16: 71359110-71359110 on Build GRCh38
rs766918913 Chr.16: 71358771-71358771 on Build GRCh38
rs948521047 Chr.16: 71358724-71358724 on Build GRCh38
rs751387727 Chr.16: 71358810-71358810 on Build GRCh38
rs371484836 Chr.16: 71358914-71358914 on Build GRCh38
rs775004919 Chr.16: 71358896-71358896 on Build GRCh38
rs888694248 Chr.16: 71358751-71358751 on Build GRCh38
rs751227285 Chr.16: 71358909-71358909 on Build GRCh38
rs899968124 Chr.16: 71359091-71359091 on Build GRCh38
rs906498789 Chr.16: 71358727-71358727 on Build GRCh38
rs542857916 Chr.16: 71359047-71359047 on Build GRCh38
rs1019445751 Chr.16: 71358667-71358667 on Build GRCh38
rs557047288 Chr.16: 71358936-71358936 on Build GRCh38
rs748418782 Chr.16: 71358756-71358756 on Build GRCh38
rs760003388 Chr.16: 71358887-71358887 on Build GRCh38
rs778071408 Chr.16: 71358848-71358848 on Build GRCh38
rs777985465 Chr.16: 71358953-71358953 on Build GRCh38
rs1044429101 Chr.16: 71358726-71358726 on Build GRCh38
rs929999694 Chr.16: 71358952-71358952 on Build GRCh38
rs534959638 Chr.16: 71358746-71358746 on Build GRCh38
rs930215552 Chr.16: 71358732-71358732 on Build GRCh38
rs771261646 Chr.16: 71358879-71358879 on Build GRCh38
rs975138330 Chr.16: 71358669-71358670 on Build GRCh38
rs968030519 Chr.16: 71358940-71358941 on Build GRCh38
rs1009474049 Chr.16: 71358875-71358875 on Build GRCh38
rs776949800 Chr.16: 71358775-71358775 on Build GRCh38
rs924414571 Chr.16: 71358696-71358696 on Build GRCh38
rs538251781 Chr.16: 71358933-71358933 on Build GRCh38
rs1031541094 Chr.16: 71359052-71359052 on Build GRCh38
rs752173653 Chr.16: 71358781-71358781 on Build GRCh38
rs780925338 Chr.16: 71358743-71358743 on Build GRCh38
rs756045977 Chr.16: 71358834-71358834 on Build GRCh38
rs773731138 Chr.16: 71358760-71358760 on Build GRCh38
rs763688995 Chr.16: 71358793-71358793 on Build GRCh38
rs745857934 Chr.16: 71358829-71358829 on Build GRCh38
rs954413837 Chr.16: 71358835-71358835 on Build GRCh38
rs1047075706 Chr.16: 71359014-71359014 on Build GRCh38
rs774567031 Chr.16: 71358750-71358750 on Build GRCh38
rs760417141 Chr.16: 71358758-71358780 on Build GRCh38
rs1000031506 Chr.16: 71358951-71358951 on Build GRCh38
rs948111085 Chr.16: 71358686-71358686 on Build GRCh38
rs748187694 Chr.16: 71358838-71358838 on Build GRCh38
rs968583217 Chr.16: 71358674-71358674 on Build GRCh38
rs552552534 Chr.16: 71358827-71358827 on Build GRCh38
rs779181390 Chr.16: 71358837-71358837 on Build GRCh38
rs780095422 Chr.16: 71358935-71358935 on Build GRCh38
rs898842960 Chr.16: 71358748-71358748 on Build GRCh38
rs998358194 Chr.16: 71359098-71359098 on Build GRCh38
rs934492340 Chr.16: 71358710-71358710 on Build GRCh38
rs769940416 Chr.16: 71358927-71358927 on Build GRCh38
rs760903738 Chr.16: 71358762-71358762 on Build GRCh38
rs768443898 Chr.16: 71358763-71358764 on Build GRCh38

View Full Product Details