Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 71374505-71375005 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALB2
Entrez Gene ID
794
Gene Name
calbindin 2
Gene Aliases
CAB29, CAL2, CR
UniGene
Hs.106857
Gene Chromosome Location
Chr.16: 71358713-71390438 on Build GRCh38
Transcripts
NM_001740.4 NM_007088.3 NR_027910.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4845334 c.180G>C p.K60N Chr.16: 71374753-71374753 on Build GRCh38
219210 c.226G>A p.D76N Chr.16: 71374799-71374799 on Build GRCh38
4062523 c.242G>T p.G81V Chr.16: 71374815-71374815 on Build GRCh38
3511820 c.213C>T p.F71F Chr.16: 71374786-71374786 on Build GRCh38
5722470 c.208G>A p.E70K Chr.16: 71374781-71374781 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs535776697 Chr.16: 71374898-71374898 on Build GRCh38
rs759366481 Chr.16: 71374781-71374781 on Build GRCh38
rs962831758 Chr.16: 71374581-71374581 on Build GRCh38
rs1045944552 Chr.16: 71374567-71374567 on Build GRCh38
rs778381159 Chr.16: 71374931-71374931 on Build GRCh38
rs755530471 Chr.16: 71374830-71374830 on Build GRCh38
rs771666794 Chr.16: 71374759-71374759 on Build GRCh38
rs369408428 Chr.16: 71374838-71374838 on Build GRCh38
rs541446343 Chr.16: 71374881-71374881 on Build GRCh38
rs776592931 Chr.16: 71374777-71374777 on Build GRCh38
rs914934791 Chr.16: 71374652-71374652 on Build GRCh38
rs755307344 Chr.16: 71374858-71374858 on Build GRCh38
rs200083611 Chr.16: 71374720-71374720 on Build GRCh38
rs545598992 Chr.16: 71374921-71374921 on Build GRCh38
rs375145613 Chr.16: 71374849-71374849 on Build GRCh38
rs749603957 Chr.16: 71374733-71374733 on Build GRCh38
rs748741756 Chr.16: 71374859-71374859 on Build GRCh38
rs1003649674 Chr.16: 71374704-71374704 on Build GRCh38
rs867857228 Chr.16: 71374822-71374822 on Build GRCh38
rs952461569 Chr.16: 71374617-71374617 on Build GRCh38
rs12444419 Chr.16: 71374572-71374572 on Build GRCh38
rs915077197 Chr.16: 71374961-71374961 on Build GRCh38
rs1005297133 Chr.16: 71374622-71374622 on Build GRCh38
rs779407086 Chr.16: 71374743-71374743 on Build GRCh38
rs918586282 Chr.16: 71374606-71374606 on Build GRCh38
rs750478171 Chr.16: 71374841-71374841 on Build GRCh38
rs758740208 Chr.16: 71374594-71374594 on Build GRCh38
rs546231096 Chr.16: 71374590-71374590 on Build GRCh38
rs372727389 Chr.16: 71374712-71374712 on Build GRCh38
rs959519305 Chr.16: 71374560-71374560 on Build GRCh38
rs1005328362 Chr.16: 71374906-71374906 on Build GRCh38
rs548694562 Chr.16: 71374535-71374535 on Build GRCh38
rs781329253 Chr.16: 71374801-71374801 on Build GRCh38
rs963689049 Chr.16: 71374635-71374635 on Build GRCh38
rs8046337 Chr.16: 71374624-71374624 on Build GRCh38
rs142502192 Chr.16: 71374834-71374834 on Build GRCh38
rs878939001 Chr.16: 71374700-71374700 on Build GRCh38
rs766545830 Chr.16: 71374848-71374848 on Build GRCh38
rs571782306 Chr.16: 71374904-71374904 on Build GRCh38
rs770688056 Chr.16: 71374775-71374775 on Build GRCh38
rs765340622 Chr.16: 71374582-71374582 on Build GRCh38
rs756297028 Chr.16: 71374714-71374714 on Build GRCh38
rs577621467 Chr.16: 71374649-71374649 on Build GRCh38
rs766822511 Chr.16: 71374619-71374619 on Build GRCh38
rs755507380 Chr.16: 71374741-71374741 on Build GRCh38
rs775441278 Chr.16: 71374832-71374832 on Build GRCh38
rs1025835339 Chr.16: 71374602-71374602 on Build GRCh38
rs764781594 Chr.16: 71374725-71374725 on Build GRCh38
rs371362570 Chr.16: 71374651-71374651 on Build GRCh38
rs371863812 Chr.16: 71374738-71374738 on Build GRCh38
rs147510392 Chr.16: 71374613-71374613 on Build GRCh38
rs867998895 Chr.16: 71374680-71374680 on Build GRCh38
rs943154569 Chr.16: 71374623-71374623 on Build GRCh38
rs375167687 Chr.16: 71374837-71374837 on Build GRCh38
rs747621062 Chr.16: 71374756-71374756 on Build GRCh38
rs1014501868 Chr.16: 71374563-71374563 on Build GRCh38
rs757295115 Chr.16: 71374697-71374697 on Build GRCh38
rs756350024 Chr.16: 71374845-71374845 on Build GRCh38
rs375554739 Chr.16: 71374611-71374611 on Build GRCh38
rs369615740 Chr.16: 71374701-71374701 on Build GRCh38
rs964625544 Chr.16: 71374926-71374926 on Build GRCh38
rs946493670 Chr.16: 71374657-71374657 on Build GRCh38
rs974539114 Chr.16: 71374946-71374946 on Build GRCh38
rs148393734 Chr.16: 71374774-71374774 on Build GRCh38
rs779129616 Chr.16: 71374875-71374875 on Build GRCh38
rs556576170 Chr.16: 71374764-71374764 on Build GRCh38
rs375585271 Chr.16: 71374703-71374703 on Build GRCh38
rs765093163 Chr.16: 71374786-71374786 on Build GRCh38
rs976692497 Chr.16: 71374673-71374673 on Build GRCh38

View Full Product Details