Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7390083-7390579 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLSCR3
Entrez Gene ID
57048
Gene Name
phospholipid scramblase 3
Gene Aliases
-
UniGene
Hs.534591
Gene Chromosome Location
Chr.17: 7389727-7394843 on Build GRCh38
Transcripts
NM_001201576.1 NM_020360.3 NR_037719.1

Gene Symbol
TMEM256-PLSCR3
Entrez Gene ID
100529211
Gene Name
TMEM256-PLSCR3 readthrough (NMD candidate)
Gene Aliases
C17orf61-PLSCR3
UniGene
Hs.534591
Gene Chromosome Location
Chr.17: 7389727-7404131 on Build GRCh38
Transcripts
NM_001201576.1 NM_020360.3 NR_037719.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7222272 c.877G>A p.V293I Chr.17: 7390396-7390396 on Build GRCh38
5430783 c.861C>T p.G287G Chr.17: 7390412-7390412 on Build GRCh38
6283050 c.832-1G>A p.? Chr.17: 7390442-7390442 on Build GRCh38
6283049 c.1045-1G>A p.? Chr.17: 7390442-7390442 on Build GRCh38
5430782 c.1074C>T p.G358G Chr.17: 7390412-7390412 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755389796 Chr.17: 7390466-7390466 on Build GRCh38
rs771850451 Chr.17: 7390533-7390533 on Build GRCh38
rs758694945 Chr.17: 7390375-7390375 on Build GRCh38
rs77407748 Chr.17: 7390169-7390169 on Build GRCh38
rs892895172 Chr.17: 7390239-7390239 on Build GRCh38
rs930437106 Chr.17: 7390123-7390123 on Build GRCh38
rs1048891232 Chr.17: 7390157-7390157 on Build GRCh38
rs535806752 Chr.17: 7390397-7390397 on Build GRCh38
rs34961966 Chr.17: 7390433-7390433 on Build GRCh38
rs866207735 Chr.17: 7390405-7390405 on Build GRCh38
rs909166766 Chr.17: 7390218-7390218 on Build GRCh38
rs202149802 Chr.17: 7390369-7390369 on Build GRCh38
rs74395124 Chr.17: 7390366-7390366 on Build GRCh38
rs765489424 Chr.17: 7390435-7390435 on Build GRCh38
rs781672746 Chr.17: 7390469-7390469 on Build GRCh38
rs377553710 Chr.17: 7390411-7390411 on Build GRCh38
rs1011071259 Chr.17: 7390265-7390266 on Build GRCh38
rs3744549 Chr.17: 7390396-7390396 on Build GRCh38
rs537146529 Chr.17: 7390256-7390256 on Build GRCh38
rs558397818 Chr.17: 7390266-7390266 on Build GRCh38
rs148500270 Chr.17: 7390437-7390437 on Build GRCh38
rs575594227 Chr.17: 7390432-7390432 on Build GRCh38
rs751983446 Chr.17: 7390463-7390463 on Build GRCh38
rs749450214 Chr.17: 7390119-7390119 on Build GRCh38
rs748939643 Chr.17: 7390507-7390507 on Build GRCh38
rs375375075 Chr.17: 7390420-7390420 on Build GRCh38
rs774319540 Chr.17: 7390414-7390414 on Build GRCh38
rs373690880 Chr.17: 7390158-7390158 on Build GRCh38
rs763365463 Chr.17: 7390449-7390449 on Build GRCh38
rs1016173616 Chr.17: 7390549-7390549 on Build GRCh38
rs776807170 Chr.17: 7390421-7390421 on Build GRCh38
rs370775245 Chr.17: 7390412-7390412 on Build GRCh38
rs770730812 Chr.17: 7390528-7390528 on Build GRCh38
rs1031402582 Chr.17: 7390228-7390228 on Build GRCh38
rs376839053 Chr.17: 7390544-7390544 on Build GRCh38
rs766704085 Chr.17: 7390456-7390456 on Build GRCh38
rs919272078 Chr.17: 7390312-7390312 on Build GRCh38
rs745701003 Chr.17: 7390532-7390532 on Build GRCh38
rs777403711 Chr.17: 7390406-7390406 on Build GRCh38
rs1039313046 Chr.17: 7390477-7390500 on Build GRCh38
rs553857643 Chr.17: 7390538-7390538 on Build GRCh38
rs189301132 Chr.17: 7390105-7390105 on Build GRCh38
rs998536618 Chr.17: 7390227-7390227 on Build GRCh38
rs752066148 Chr.17: 7390388-7390388 on Build GRCh38
rs960668365 Chr.17: 7390358-7390358 on Build GRCh38
rs930765760 Chr.17: 7390383-7390383 on Build GRCh38
rs770058593 Chr.17: 7390547-7390547 on Build GRCh38
rs573350814 Chr.17: 7390529-7390529 on Build GRCh38
rs1035289667 Chr.17: 7390353-7390353 on Build GRCh38
rs771523917 Chr.17: 7390490-7390490 on Build GRCh38
rs754478715 Chr.17: 7390389-7390389 on Build GRCh38
rs762090319 Chr.17: 7390355-7390355 on Build GRCh38
rs752135002 Chr.17: 7390473-7390473 on Build GRCh38
rs986444247 Chr.17: 7390217-7390217 on Build GRCh38
rs192298510 Chr.17: 7390179-7390179 on Build GRCh38
rs1030114382 Chr.17: 7390517-7390517 on Build GRCh38
rs780609079 Chr.17: 7390386-7390386 on Build GRCh38
rs769378024 Chr.17: 7390403-7390403 on Build GRCh38
rs568083105 Chr.17: 7390495-7390496 on Build GRCh38
rs867627072 Chr.17: 7390453-7390453 on Build GRCh38
rs114731648 Chr.17: 7390292-7390292 on Build GRCh38
rs759434956 Chr.17: 7390417-7390417 on Build GRCh38
rs953243230 Chr.17: 7390198-7390198 on Build GRCh38
rs772081469 Chr.17: 7390419-7390419 on Build GRCh38
rs949474701 Chr.17: 7390321-7390321 on Build GRCh38
rs755683283 Chr.17: 7390479-7390479 on Build GRCh38
rs997290164 Chr.17: 7390175-7390175 on Build GRCh38

View Full Product Details