Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7425877-7426367 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf74
Entrez Gene ID
201243
Gene Name
chromosome 17 open reading frame 74
Gene Aliases
-
UniGene
Hs.380704
Gene Chromosome Location
Chr.17: 7424874-7427568 on Build GRCh38
Transcripts
NM_175734.4 XM_006721471.3 XM_011523722.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3522090 c.199G>A p.E67K Chr.17: 7426190-7426190 on Build GRCh38
3522091 c.321C>T p.F107F Chr.17: 7426312-7426312 on Build GRCh38
6198184 c.299G>A p.R100Q Chr.17: 7426290-7426290 on Build GRCh38
3522094 c.376C>T p.R126C Chr.17: 7426367-7426367 on Build GRCh38
6442469 c.323C>T p.S108F Chr.17: 7426314-7426314 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs891053869 Chr.17: 7426134-7426134 on Build GRCh38
rs763523156 Chr.17: 7426057-7426057 on Build GRCh38
rs978932807 Chr.17: 7425936-7425936 on Build GRCh38
rs767198056 Chr.17: 7426078-7426078 on Build GRCh38
rs762324717 Chr.17: 7425953-7425953 on Build GRCh38
rs562221750 Chr.17: 7425942-7425942 on Build GRCh38
rs918836507 Chr.17: 7425989-7425989 on Build GRCh38
rs902357224 Chr.17: 7426130-7426130 on Build GRCh38
rs752912269 Chr.17: 7426296-7426296 on Build GRCh38
rs201123413 Chr.17: 7426169-7426169 on Build GRCh38
rs746634106 Chr.17: 7426333-7426333 on Build GRCh38
rs974244108 Chr.17: 7425960-7425960 on Build GRCh38
rs373853895 Chr.17: 7425963-7425963 on Build GRCh38
rs753844132 Chr.17: 7426004-7426004 on Build GRCh38
rs376834840 Chr.17: 7425926-7425926 on Build GRCh38
rs778216192 Chr.17: 7426179-7426179 on Build GRCh38
rs112559835 Chr.17: 7426060-7426060 on Build GRCh38
rs760346583 Chr.17: 7426081-7426081 on Build GRCh38
rs757683230 Chr.17: 7426312-7426312 on Build GRCh38
rs578139792 Chr.17: 7426290-7426290 on Build GRCh38
rs201970262 Chr.17: 7426332-7426332 on Build GRCh38
rs987773699 Chr.17: 7425925-7425925 on Build GRCh38
rs761616719 Chr.17: 7426346-7426346 on Build GRCh38
rs752555933 Chr.17: 7425971-7425971 on Build GRCh38
rs773555949 Chr.17: 7426056-7426056 on Build GRCh38
rs1029498836 Chr.17: 7426121-7426121 on Build GRCh38
rs745414742 Chr.17: 7426045-7426045 on Build GRCh38
rs775056247 Chr.17: 7426217-7426217 on Build GRCh38
rs1029632071 Chr.17: 7425924-7425924 on Build GRCh38
rs765310990 Chr.17: 7426269-7426269 on Build GRCh38
rs369509500 Chr.17: 7426014-7426014 on Build GRCh38
rs762402607 Chr.17: 7425944-7425944 on Build GRCh38
rs567515569 Chr.17: 7426141-7426141 on Build GRCh38
rs531443696 Chr.17: 7426101-7426101 on Build GRCh38
rs959095071 Chr.17: 7426276-7426276 on Build GRCh38
rs370832954 Chr.17: 7426284-7426284 on Build GRCh38
rs765699691 Chr.17: 7425952-7425952 on Build GRCh38
rs999854123 Chr.17: 7425913-7425913 on Build GRCh38
rs777996653 Chr.17: 7426297-7426297 on Build GRCh38
rs369819820 Chr.17: 7425955-7425955 on Build GRCh38
rs371484318 Chr.17: 7425954-7425954 on Build GRCh38
rs773910509 Chr.17: 7426072-7426072 on Build GRCh38
rs753658255 Chr.17: 7426092-7426092 on Build GRCh38
rs771536180 Chr.17: 7426197-7426197 on Build GRCh38
rs777522039 Chr.17: 7425988-7425988 on Build GRCh38
rs1015081069 Chr.17: 7425939-7425939 on Build GRCh38
rs561423308 Chr.17: 7426025-7426025 on Build GRCh38
rs996693465 Chr.17: 7426110-7426110 on Build GRCh38
rs533796911 Chr.17: 7426098-7426098 on Build GRCh38
rs115375487 Chr.17: 7425927-7425927 on Build GRCh38
rs781659846 Chr.17: 7426032-7426032 on Build GRCh38
rs930203884 Chr.17: 7425991-7425991 on Build GRCh38
rs115992886 Chr.17: 7426013-7426013 on Build GRCh38
rs755224918 Chr.17: 7426147-7426147 on Build GRCh38
rs75175048 Chr.17: 7426042-7426042 on Build GRCh38
rs370485691 Chr.17: 7426080-7426080 on Build GRCh38
rs763953202 Chr.17: 7426082-7426082 on Build GRCh38
rs760388271 Chr.17: 7426239-7426239 on Build GRCh38
rs773820752 Chr.17: 7426201-7426201 on Build GRCh38
rs1018303450 Chr.17: 7426242-7426242 on Build GRCh38
rs376929151 Chr.17: 7426178-7426178 on Build GRCh38
rs556644443 Chr.17: 7426252-7426252 on Build GRCh38
rs373604431 Chr.17: 7426117-7426117 on Build GRCh38
rs375353507 Chr.17: 7426336-7426336 on Build GRCh38
rs1009161442 Chr.17: 7426162-7426162 on Build GRCh38
rs370306548 Chr.17: 7426184-7426184 on Build GRCh38
rs756014820 Chr.17: 7425987-7425987 on Build GRCh38
rs761621618 Chr.17: 7426267-7426267 on Build GRCh38
rs773879220 Chr.17: 7425967-7425968 on Build GRCh38
rs962806932 Chr.17: 7426294-7426294 on Build GRCh38
rs375487223 Chr.17: 7426212-7426212 on Build GRCh38
rs775241919 Chr.17: 7426096-7426096 on Build GRCh38
rs748416641 Chr.17: 7426175-7426175 on Build GRCh38
rs561499391 Chr.17: 7425900-7425900 on Build GRCh38
rs749703438 Chr.17: 7426311-7426311 on Build GRCh38
rs769062909 Chr.17: 7426028-7426028 on Build GRCh38
rs955381974 Chr.17: 7426231-7426231 on Build GRCh38
rs759185960 Chr.17: 7426076-7426076 on Build GRCh38
rs770221506 Chr.17: 7426051-7426051 on Build GRCh38
rs13290 Chr.17: 7426313-7426313 on Build GRCh38
rs899701861 Chr.17: 7426091-7426091 on Build GRCh38
rs771630138 Chr.17: 7426047-7426047 on Build GRCh38
rs750403753 Chr.17: 7426271-7426271 on Build GRCh38
rs772867396 Chr.17: 7426055-7426055 on Build GRCh38
rs755073547 Chr.17: 7426277-7426277 on Build GRCh38
rs751725202 Chr.17: 7426142-7426142 on Build GRCh38
rs759096278 Chr.17: 7426203-7426203 on Build GRCh38
rs991703006 Chr.17: 7425957-7425957 on Build GRCh38
rs967818023 Chr.17: 7426186-7426186 on Build GRCh38
rs917514416 Chr.17: 7425984-7425984 on Build GRCh38
rs1007330050 Chr.17: 7426240-7426240 on Build GRCh38
rs999422540 Chr.17: 7426174-7426174 on Build GRCh38
rs939039489 Chr.17: 7426038-7426038 on Build GRCh38
rs200975825 Chr.17: 7426015-7426015 on Build GRCh38
rs763781973 Chr.17: 7426246-7426246 on Build GRCh38
rs550194923 Chr.17: 7425931-7425931 on Build GRCh38

View Full Product Details