Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7427233-7427725 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf74
Entrez Gene ID
201243
Gene Name
chromosome 17 open reading frame 74
Gene Aliases
-
UniGene
Hs.380704
Gene Chromosome Location
Chr.17: 7424874-7427568 on Build GRCh38
Transcripts
NM_175734.4 XM_006721471.3 XM_011523722.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6220438 c.1348C>T p.R450W Chr.17: 7427339-7427339 on Build GRCh38
7347968 c.1384G>A p.D462N Chr.17: 7427375-7427375 on Build GRCh38
5182843 c.1323C>T p.P441P Chr.17: 7427314-7427314 on Build GRCh38
7412346 c.1445C>T p.P482L Chr.17: 7427436-7427436 on Build GRCh38
5387286 c.1313_1314CT>GA p.P438R Chr.17: 7427304-7427305 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1009567488 Chr.17: 7427588-7427588 on Build GRCh38
rs771225208 Chr.17: 7427475-7427475 on Build GRCh38
rs749541208 Chr.17: 7427394-7427394 on Build GRCh38
rs766177360 Chr.17: 7427547-7427547 on Build GRCh38
rs770047620 Chr.17: 7427299-7427299 on Build GRCh38
rs757100482 Chr.17: 7427263-7427263 on Build GRCh38
rs571970694 Chr.17: 7427678-7427678 on Build GRCh38
rs113571740 Chr.17: 7427607-7427607 on Build GRCh38
rs990195692 Chr.17: 7427378-7427378 on Build GRCh38
rs751504429 Chr.17: 7427459-7427459 on Build GRCh38
rs948747487 Chr.17: 7427454-7427454 on Build GRCh38
rs762027982 Chr.17: 7427358-7427358 on Build GRCh38
rs763672128 Chr.17: 7427437-7427437 on Build GRCh38
rs781223493 Chr.17: 7427375-7427375 on Build GRCh38
rs774666797 Chr.17: 7427312-7427312 on Build GRCh38
rs779400015 Chr.17: 7427482-7427482 on Build GRCh38
rs201127764 Chr.17: 7427339-7427339 on Build GRCh38
rs201598699 Chr.17: 7427298-7427298 on Build GRCh38
rs976905687 Chr.17: 7427698-7427698 on Build GRCh38
rs774927123 Chr.17: 7427411-7427411 on Build GRCh38
rs1026717507 Chr.17: 7427677-7427677 on Build GRCh38
rs756092105 Chr.17: 7427463-7427463 on Build GRCh38
rs772879035 Chr.17: 7427534-7427534 on Build GRCh38
rs1027767300 Chr.17: 7427656-7427656 on Build GRCh38
rs771349956 Chr.17: 7427494-7427501 on Build GRCh38
rs200505211 Chr.17: 7427427-7427427 on Build GRCh38
rs536312334 Chr.17: 7427696-7427696 on Build GRCh38
rs201627896 Chr.17: 7427340-7427340 on Build GRCh38
rs373029891 Chr.17: 7427453-7427453 on Build GRCh38
rs907794658 Chr.17: 7427318-7427318 on Build GRCh38
rs749471824 Chr.17: 7427473-7427473 on Build GRCh38
rs1015922080 Chr.17: 7427704-7427704 on Build GRCh38
rs201902717 Chr.17: 7427400-7427400 on Build GRCh38
rs773266078 Chr.17: 7427693-7427693 on Build GRCh38
rs1023470007 Chr.17: 7427520-7427520 on Build GRCh38
rs750355933 Chr.17: 7427353-7427353 on Build GRCh38
rs180971352 Chr.17: 7427643-7427643 on Build GRCh38
rs958381873 Chr.17: 7427301-7427301 on Build GRCh38
rs768707771 Chr.17: 7427296-7427296 on Build GRCh38
rs76470256 Chr.17: 7427658-7427658 on Build GRCh38
rs931755863 Chr.17: 7427392-7427392 on Build GRCh38
rs762668341 Chr.17: 7427535-7427535 on Build GRCh38
rs771333790 Chr.17: 7427311-7427311 on Build GRCh38
rs763606341 Chr.17: 7427321-7427321 on Build GRCh38
rs200062575 Chr.17: 7427315-7427315 on Build GRCh38
rs747607927 Chr.17: 7427497-7427497 on Build GRCh38
rs565155443 Chr.17: 7427277-7427277 on Build GRCh38
rs776668655 Chr.17: 7427505-7427505 on Build GRCh38
rs748174437 Chr.17: 7427376-7427376 on Build GRCh38
rs774683450 Chr.17: 7427519-7427520 on Build GRCh38
rs569879387 Chr.17: 7427432-7427432 on Build GRCh38
rs970508648 Chr.17: 7427528-7427528 on Build GRCh38
rs760225345 Chr.17: 7427436-7427436 on Build GRCh38
rs199910812 Chr.17: 7427457-7427457 on Build GRCh38
rs777593959 Chr.17: 7427257-7427257 on Build GRCh38
rs776244852 Chr.17: 7427507-7427507 on Build GRCh38
rs371497094 Chr.17: 7427317-7427317 on Build GRCh38
rs376049395 Chr.17: 7427445-7427445 on Build GRCh38
rs1041022849 Chr.17: 7427271-7427271 on Build GRCh38
rs532496564 Chr.17: 7427316-7427316 on Build GRCh38
rs772755354 Chr.17: 7427428-7427428 on Build GRCh38
rs763277023 Chr.17: 7427348-7427348 on Build GRCh38
rs776080286 Chr.17: 7427489-7427489 on Build GRCh38
rs777970832 Chr.17: 7427466-7427466 on Build GRCh38
rs537230526 Chr.17: 7427448-7427448 on Build GRCh38
rs184219833 Chr.17: 7427393-7427393 on Build GRCh38
rs760382855 Chr.17: 7427320-7427320 on Build GRCh38
rs776868418 Chr.17: 7427355-7427355 on Build GRCh38
rs772597881 Chr.17: 7427483-7427483 on Build GRCh38
rs962295448 Chr.17: 7427620-7427620 on Build GRCh38
rs764910593 Chr.17: 7427446-7427446 on Build GRCh38
rs985805369 Chr.17: 7427666-7427666 on Build GRCh38
rs768046304 Chr.17: 7427506-7427506 on Build GRCh38
rs902776631 Chr.17: 7427479-7427479 on Build GRCh38
rs369230826 Chr.17: 7427388-7427388 on Build GRCh38
rs764744273 Chr.17: 7427625-7427625 on Build GRCh38
rs552378300 Chr.17: 7427525-7427525 on Build GRCh38
rs760182580 Chr.17: 7427313-7427313 on Build GRCh38

View Full Product Details