Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7445178-7445667 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNB1
Entrez Gene ID
1140
Gene Name
cholinergic receptor nicotinic beta 1 subunit
Gene Aliases
ACHRB, CHRNB, CMS1D, CMS2A, CMS2C, SCCMS
UniGene
Hs.330386
Gene Chromosome Location
Chr.17: 7445087-7457613 on Build GRCh38
Transcripts
NM_000747.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4651317 c.112T>C p.Y38H Chr.17: 7445323-7445323 on Build GRCh38
6628755 c.105C>A p.F35L Chr.17: 7445316-7445316 on Build GRCh38
7096952 c.155T>C p.V52A Chr.17: 7445366-7445366 on Build GRCh38
2978928 c.89T>A p.L30H Chr.17: 7445300-7445300 on Build GRCh38
4986492 c.113A>T p.Y38F Chr.17: 7445324-7445324 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1045028787 Chr.17: 7445641-7445641 on Build GRCh38
rs763061831 Chr.17: 7445222-7445222 on Build GRCh38
rs1054460489 Chr.17: 7445510-7445510 on Build GRCh38
rs765236873 Chr.17: 7445294-7445294 on Build GRCh38
rs372041531 Chr.17: 7445267-7445267 on Build GRCh38
rs764620030 Chr.17: 7445316-7445316 on Build GRCh38
rs775150297 Chr.17: 7445272-7445272 on Build GRCh38
rs768484065 Chr.17: 7445282-7445282 on Build GRCh38
rs779475593 Chr.17: 7445325-7445325 on Build GRCh38
rs753018299 Chr.17: 7445301-7445301 on Build GRCh38
rs968038340 Chr.17: 7445314-7445314 on Build GRCh38
rs757667532 Chr.17: 7445422-7445422 on Build GRCh38
rs760465598 Chr.17: 7445204-7445204 on Build GRCh38
rs745326511 Chr.17: 7445334-7445334 on Build GRCh38
rs746733626 Chr.17: 7445350-7445350 on Build GRCh38
rs962389622 Chr.17: 7445408-7445408 on Build GRCh38
rs886185054 Chr.17: 7445254-7445254 on Build GRCh38
rs772263997 Chr.17: 7445457-7445457 on Build GRCh38
rs780930151 Chr.17: 7445443-7445443 on Build GRCh38
rs1001271364 Chr.17: 7445330-7445330 on Build GRCh38
rs17856697 Chr.17: 7445306-7445306 on Build GRCh38
rs903202318 Chr.17: 7445216-7445216 on Build GRCh38
rs200245854 Chr.17: 7445421-7445421 on Build GRCh38
rs574225093 Chr.17: 7445243-7445243 on Build GRCh38
rs775653059 Chr.17: 7445461-7445461 on Build GRCh38
rs777542254 Chr.17: 7445449-7445449 on Build GRCh38
rs771885675 Chr.17: 7445197-7445197 on Build GRCh38
rs912711009 Chr.17: 7445419-7445419 on Build GRCh38
rs539992749 Chr.17: 7445567-7445567 on Build GRCh38
rs149024203 Chr.17: 7445625-7445625 on Build GRCh38
rs759668768 Chr.17: 7445378-7445378 on Build GRCh38
rs547772932 Chr.17: 7445229-7445229 on Build GRCh38
rs745392905 Chr.17: 7445266-7445266 on Build GRCh38
rs948817419 Chr.17: 7445534-7445534 on Build GRCh38
rs761035556 Chr.17: 7445398-7445398 on Build GRCh38
rs755101484 Chr.17: 7445231-7445231 on Build GRCh38
rs199665345 Chr.17: 7445424-7445424 on Build GRCh38
rs60038135 Chr.17: 7445445-7445445 on Build GRCh38
rs768429056 Chr.17: 7445355-7445355 on Build GRCh38
rs759694228 Chr.17: 7445296-7445296 on Build GRCh38
rs749304189 Chr.17: 7445453-7445453 on Build GRCh38
rs375737554 Chr.17: 7445529-7445529 on Build GRCh38
rs775849224 Chr.17: 7445394-7445394 on Build GRCh38
rs936030338 Chr.17: 7445494-7445494 on Build GRCh38
rs776631018 Chr.17: 7445289-7445289 on Build GRCh38
rs755694552 Chr.17: 7445312-7445313 on Build GRCh38
rs756569508 Chr.17: 7445242-7445242 on Build GRCh38
rs531280641 Chr.17: 7445528-7445528 on Build GRCh38
rs748919331 Chr.17: 7445421-7445422 on Build GRCh38
rs761717412 Chr.17: 7445293-7445293 on Build GRCh38
rs747983720 Chr.17: 7445375-7445375 on Build GRCh38
rs763975239 Chr.17: 7445218-7445218 on Build GRCh38
rs751068058 Chr.17: 7445328-7445328 on Build GRCh38
rs769579645 Chr.17: 7445376-7445376 on Build GRCh38
rs975656395 Chr.17: 7445430-7445430 on Build GRCh38
rs757646406 Chr.17: 7445321-7445321 on Build GRCh38
rs903761397 Chr.17: 7445642-7445642 on Build GRCh38
rs35872101 Chr.17: 7445343-7445343 on Build GRCh38
rs778342718 Chr.17: 7445263-7445263 on Build GRCh38
rs779654919 Chr.17: 7445348-7445348 on Build GRCh38
rs754354256 Chr.17: 7445318-7445318 on Build GRCh38
rs376933292 Chr.17: 7445437-7445437 on Build GRCh38
rs767799511 Chr.17: 7445297-7445297 on Build GRCh38
rs754080801 Chr.17: 7445418-7445418 on Build GRCh38
rs765610075 Chr.17: 7445423-7445423 on Build GRCh38
rs200985395 Chr.17: 7445223-7445223 on Build GRCh38
rs781570982 Chr.17: 7445232-7445232 on Build GRCh38
rs879255346 Chr.17: 7445377-7445377 on Build GRCh38
rs1056243202 Chr.17: 7445619-7445619 on Build GRCh38
rs749799086 Chr.17: 7445249-7445249 on Build GRCh38
rs746539526 Chr.17: 7445425-7445425 on Build GRCh38
rs138041351 Chr.17: 7445295-7445295 on Build GRCh38
rs753073504 Chr.17: 7445233-7445233 on Build GRCh38
rs1012581418 Chr.17: 7445629-7445629 on Build GRCh38
rs907656093 Chr.17: 7445530-7445530 on Build GRCh38
rs1015239523 Chr.17: 7445393-7445393 on Build GRCh38
rs777431654 Chr.17: 7445324-7445326 on Build GRCh38
rs746753981 Chr.17: 7445274-7445274 on Build GRCh38
rs924594452 Chr.17: 7445491-7445491 on Build GRCh38
rs767744371 Chr.17: 7445387-7445387 on Build GRCh38
rs776235550 Chr.17: 7445356-7445356 on Build GRCh38
rs371882653 Chr.17: 7445201-7445201 on Build GRCh38
rs764423909 Chr.17: 7445400-7445400 on Build GRCh38
rs921584841 Chr.17: 7445200-7445200 on Build GRCh38
rs770468832 Chr.17: 7445256-7445256 on Build GRCh38
rs944946128 Chr.17: 7445450-7445450 on Build GRCh38
rs781017018 Chr.17: 7445441-7445441 on Build GRCh38

View Full Product Details