Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7455179-7455674 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNB1
Entrez Gene ID
1140
Gene Name
cholinergic receptor nicotinic beta 1 subunit
Gene Aliases
ACHRB, CHRNB, CMS1D, CMS2A, CMS2C, SCCMS
UniGene
Hs.330386
Gene Chromosome Location
Chr.17: 7445087-7457613 on Build GRCh38
Transcripts
NM_000747.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7231786 c.1125C>A p.H375Q Chr.17: 7455364-7455364 on Build GRCh38
3890422 c.1173C>T p.F391F Chr.17: 7455412-7455412 on Build GRCh38
3820556 c.1143T>G p.S381R Chr.17: 7455382-7455382 on Build GRCh38
3932732 c.1096G>A p.E366K Chr.17: 7455335-7455335 on Build GRCh38
3890423 c.1203C>T p.F401F Chr.17: 7455442-7455442 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs979257638 Chr.17: 7455547-7455547 on Build GRCh38
rs201761770 Chr.17: 7455481-7455481 on Build GRCh38
rs1011735835 Chr.17: 7455211-7455211 on Build GRCh38
rs553816148 Chr.17: 7455531-7455531 on Build GRCh38
rs142367311 Chr.17: 7455588-7455588 on Build GRCh38
rs749002133 Chr.17: 7455451-7455451 on Build GRCh38
rs776417124 Chr.17: 7455244-7455244 on Build GRCh38
rs778609818 Chr.17: 7455457-7455457 on Build GRCh38
rs1018014699 Chr.17: 7455404-7455404 on Build GRCh38
rs201226380 Chr.17: 7455495-7455495 on Build GRCh38
rs776528675 Chr.17: 7455353-7455353 on Build GRCh38
rs768319367 Chr.17: 7455235-7455235 on Build GRCh38
rs190598236 Chr.17: 7455466-7455466 on Build GRCh38
rs1006544826 Chr.17: 7455606-7455606 on Build GRCh38
rs533577653 Chr.17: 7455324-7455324 on Build GRCh38
rs374308530 Chr.17: 7455268-7455268 on Build GRCh38
rs755454760 Chr.17: 7455252-7455253 on Build GRCh38
rs985214726 Chr.17: 7455394-7455394 on Build GRCh38
rs867944973 Chr.17: 7455362-7455362 on Build GRCh38
rs375829198 Chr.17: 7455273-7455273 on Build GRCh38
rs543581079 Chr.17: 7455557-7455557 on Build GRCh38
rs767883007 Chr.17: 7455492-7455492 on Build GRCh38
rs79209506 Chr.17: 7455278-7455278 on Build GRCh38
rs772969489 Chr.17: 7455334-7455334 on Build GRCh38
rs903450905 Chr.17: 7455564-7455564 on Build GRCh38
rs766285953 Chr.17: 7455262-7455262 on Build GRCh38
rs377094762 Chr.17: 7455234-7455234 on Build GRCh38
rs928660966 Chr.17: 7455518-7455518 on Build GRCh38
rs377089087 Chr.17: 7455493-7455493 on Build GRCh38
rs759408729 Chr.17: 7455349-7455349 on Build GRCh38
rs373665920 Chr.17: 7455308-7455308 on Build GRCh38
rs774261628 Chr.17: 7455343-7455343 on Build GRCh38
rs150290595 Chr.17: 7455365-7455365 on Build GRCh38
rs369840537 Chr.17: 7455263-7455263 on Build GRCh38
rs2302761 Chr.17: 7455201-7455201 on Build GRCh38
rs893330145 Chr.17: 7455209-7455209 on Build GRCh38
rs955666458 Chr.17: 7455530-7455530 on Build GRCh38
rs755501818 Chr.17: 7455397-7455397 on Build GRCh38
rs755568370 Chr.17: 7455494-7455494 on Build GRCh38
rs755557060 Chr.17: 7455305-7455305 on Build GRCh38
rs753963175 Chr.17: 7455274-7455274 on Build GRCh38
rs962890403 Chr.17: 7455438-7455438 on Build GRCh38
rs1017100339 Chr.17: 7455467-7455467 on Build GRCh38
rs772844442 Chr.17: 7455251-7455251 on Build GRCh38
rs548045427 Chr.17: 7455601-7455601 on Build GRCh38
rs750703601 Chr.17: 7455286-7455286 on Build GRCh38
rs760029039 Chr.17: 7455488-7455488 on Build GRCh38
rs752129718 Chr.17: 7455424-7455424 on Build GRCh38
rs913733564 Chr.17: 7455533-7455533 on Build GRCh38
rs1047438102 Chr.17: 7455634-7455634 on Build GRCh38
rs745689645 Chr.17: 7455464-7455464 on Build GRCh38
rs747724114 Chr.17: 7455318-7455318 on Build GRCh38
rs893204740 Chr.17: 7455541-7455541 on Build GRCh38
rs145242880 Chr.17: 7455465-7455465 on Build GRCh38
rs532600013 Chr.17: 7455593-7455593 on Build GRCh38
rs527273543 Chr.17: 7455650-7455650 on Build GRCh38
rs758883885 Chr.17: 7455295-7455295 on Build GRCh38
rs7214344 Chr.17: 7455484-7455484 on Build GRCh38
rs749009183 Chr.17: 7455341-7455341 on Build GRCh38
rs138345058 Chr.17: 7455437-7455437 on Build GRCh38
rs887277736 Chr.17: 7455638-7455638 on Build GRCh38
rs2302762 Chr.17: 7455542-7455542 on Build GRCh38
rs770580423 Chr.17: 7455454-7455454 on Build GRCh38
rs371323684 Chr.17: 7455532-7455532 on Build GRCh38
rs141174329 Chr.17: 7455303-7455303 on Build GRCh38
rs762833042 Chr.17: 7455258-7455258 on Build GRCh38
rs1029247916 Chr.17: 7455357-7455357 on Build GRCh38
rs962462725 Chr.17: 7455624-7455624 on Build GRCh38
rs765354765 Chr.17: 7455356-7455356 on Build GRCh38
rs981444851 Chr.17: 7455516-7455516 on Build GRCh38
rs886053403 Chr.17: 7455385-7455385 on Build GRCh38
rs781734050 Chr.17: 7455403-7455403 on Build GRCh38
rs937901466 Chr.17: 7455562-7455562 on Build GRCh38
rs993245017 Chr.17: 7455328-7455328 on Build GRCh38
rs375440711 Chr.17: 7455377-7455377 on Build GRCh38
rs747700318 Chr.17: 7455246-7455246 on Build GRCh38
rs886053402 Chr.17: 7455326-7455326 on Build GRCh38
rs746551945 Chr.17: 7455470-7455470 on Build GRCh38
rs542695847 Chr.17: 7455537-7455537 on Build GRCh38
rs561391876 Chr.17: 7455538-7455538 on Build GRCh38
rs572383677 Chr.17: 7455535-7455535 on Build GRCh38
rs761970777 Chr.17: 7455355-7455355 on Build GRCh38
rs769980666 Chr.17: 7455480-7455480 on Build GRCh38
rs997682687 Chr.17: 7455584-7455584 on Build GRCh38
rs371033292 Chr.17: 7455351-7455351 on Build GRCh38
rs780673811 Chr.17: 7455302-7455302 on Build GRCh38
rs56074046 Chr.17: 7455611-7455611 on Build GRCh38
rs755694034 Chr.17: 7455428-7455428 on Build GRCh38
rs1030536879 Chr.17: 7455585-7455585 on Build GRCh38
rs549061953 Chr.17: 7455391-7455391 on Build GRCh38
rs770780908 Chr.17: 7455342-7455342 on Build GRCh38
rs765524906 Chr.17: 7455284-7455284 on Build GRCh38
rs370637206 Chr.17: 7455265-7455265 on Build GRCh38

View Full Product Details