Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7577265-7577774 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF4A1
Entrez Gene ID
1973
Gene Name
eukaryotic translation initiation factor 4A1
Gene Aliases
DDX2A, EIF-4A, EIF4A, eIF-4A-I, eIF4A-I
UniGene
Hs.129673
Gene Chromosome Location
Chr.17: 7572706-7579006 on Build GRCh38
Transcripts
NM_001204510.1 NM_001416.3 NR_037926.1

Gene Symbol
SENP3-EIF4A1
Entrez Gene ID
100533955
Gene Name
SENP3-EIF4A1 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.129673
Gene Chromosome Location
Chr.17: 7561992-7579006 on Build GRCh38
Transcripts
NM_001204510.1 NM_001416.3 NR_037926.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4849866 c.638C>T p.S213L Chr.17: 7577357-7577357 on Build GRCh38
7237026 c.638C>A p.S213* Chr.17: 7577357-7577357 on Build GRCh38
6587611 c.699G>T p.R233R Chr.17: 7577418-7577418 on Build GRCh38
3692035 c.807C>A p.T269T Chr.17: 7577607-7577607 on Build GRCh38
3717612 c.788T>G p.L263R Chr.17: 7577588-7577588 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1049223219 Chr.17: 7577513-7577513 on Build GRCh38
rs767179329 Chr.17: 7577516-7577516 on Build GRCh38
rs761118765 Chr.17: 7577424-7577424 on Build GRCh38
rs748520404 Chr.17: 7577720-7577720 on Build GRCh38
rs780292809 Chr.17: 7577545-7577545 on Build GRCh38
rs565905669 Chr.17: 7577744-7577744 on Build GRCh38
rs754862238 Chr.17: 7577512-7577512 on Build GRCh38
rs768975515 Chr.17: 7577478-7577478 on Build GRCh38
rs750013227 Chr.17: 7577330-7577330 on Build GRCh38
rs776802273 Chr.17: 7577562-7577562 on Build GRCh38
rs184646196 Chr.17: 7577330-7577330 on Build GRCh38
rs978635014 Chr.17: 7577310-7577310 on Build GRCh38
rs368569884 Chr.17: 7577565-7577565 on Build GRCh38
rs779690652 Chr.17: 7577507-7577508 on Build GRCh38
rs189527959 Chr.17: 7577332-7577332 on Build GRCh38
rs759641276 Chr.17: 7577307-7577307 on Build GRCh38
rs751236029 Chr.17: 7577509-7577509 on Build GRCh38
rs545937171 Chr.17: 7577358-7577358 on Build GRCh38
rs758447237 Chr.17: 7577537-7577537 on Build GRCh38
rs865904208 Chr.17: 7577416-7577416 on Build GRCh38
rs373835260 Chr.17: 7577370-7577370 on Build GRCh38
rs373643163 Chr.17: 7577319-7577320 on Build GRCh38
rs575267818 Chr.17: 7577353-7577353 on Build GRCh38
rs759219316 Chr.17: 7577625-7577625 on Build GRCh38
rs747190718 Chr.17: 7577559-7577559 on Build GRCh38
rs147235946 Chr.17: 7577475-7577475 on Build GRCh38
rs780000726 Chr.17: 7577369-7577369 on Build GRCh38
rs557992096 Chr.17: 7577361-7577361 on Build GRCh38
rs761424714 Chr.17: 7577339-7577339 on Build GRCh38
rs755089234 Chr.17: 7577716-7577716 on Build GRCh38
rs772344319 Chr.17: 7577463-7577463 on Build GRCh38
rs755950697 Chr.17: 7577519-7577519 on Build GRCh38
rs948443811 Chr.17: 7577296-7577296 on Build GRCh38
rs757995543 Chr.17: 7577489-7577490 on Build GRCh38
rs772530085 Chr.17: 7577497-7577497 on Build GRCh38
rs772452235 Chr.17: 7577311-7577311 on Build GRCh38
rs754808974 Chr.17: 7577379-7577379 on Build GRCh38
rs775760123 Chr.17: 7577321-7577321 on Build GRCh38
rs764817232 Chr.17: 7577496-7577496 on Build GRCh38
rs777757461 Chr.17: 7577526-7577526 on Build GRCh38
rs766193151 Chr.17: 7577508-7577508 on Build GRCh38
rs375227176 Chr.17: 7577739-7577739 on Build GRCh38
rs370110194 Chr.17: 7577517-7577517 on Build GRCh38
rs879194979 Chr.17: 7577316-7577316 on Build GRCh38
rs764454917 Chr.17: 7577527-7577527 on Build GRCh38
rs1055411250 Chr.17: 7577518-7577518 on Build GRCh38
rs1051155483 Chr.17: 7577481-7577481 on Build GRCh38
rs758502373 Chr.17: 7577684-7577684 on Build GRCh38
rs139789270 Chr.17: 7577613-7577613 on Build GRCh38
rs780057605 Chr.17: 7577700-7577700 on Build GRCh38
rs751200894 Chr.17: 7577554-7577555 on Build GRCh38
rs373864323 Chr.17: 7577561-7577561 on Build GRCh38
rs375491044 Chr.17: 7577664-7577664 on Build GRCh38
rs774180617 Chr.17: 7577598-7577598 on Build GRCh38
rs761019570 Chr.17: 7577328-7577328 on Build GRCh38
rs754269073 Chr.17: 7577536-7577536 on Build GRCh38
rs368029902 Chr.17: 7577505-7577505 on Build GRCh38
rs143683973 Chr.17: 7577667-7577667 on Build GRCh38
rs113626456 Chr.17: 7577707-7577707 on Build GRCh38
rs764836748 Chr.17: 7577318-7577319 on Build GRCh38
rs112659619 Chr.17: 7577706-7577706 on Build GRCh38
rs774676775 Chr.17: 7577302-7577302 on Build GRCh38
rs778814483 Chr.17: 7577440-7577440 on Build GRCh38
rs921284645 Chr.17: 7577466-7577466 on Build GRCh38
rs945223741 Chr.17: 7577498-7577498 on Build GRCh38
rs753741425 Chr.17: 7577673-7577673 on Build GRCh38
rs372471773 Chr.17: 7577719-7577719 on Build GRCh38
rs369613503 Chr.17: 7577745-7577745 on Build GRCh38
rs767362546 Chr.17: 7577649-7577649 on Build GRCh38
rs762177448 Chr.17: 7577493-7577493 on Build GRCh38
rs770895700 Chr.17: 7577293-7577293 on Build GRCh38
rs746103175 Chr.17: 7577446-7577446 on Build GRCh38
rs748007558 Chr.17: 7577390-7577390 on Build GRCh38
rs778466223 Chr.17: 7577732-7577732 on Build GRCh38
rs554303388 Chr.17: 7577337-7577337 on Build GRCh38
rs540578884 Chr.17: 7577589-7577589 on Build GRCh38
rs781329958 Chr.17: 7577381-7577381 on Build GRCh38
rs775673905 Chr.17: 7577468-7577468 on Build GRCh38
rs11552639 Chr.17: 7577393-7577393 on Build GRCh38
rs777224973 Chr.17: 7577483-7577483 on Build GRCh38
rs770174559 Chr.17: 7577730-7577730 on Build GRCh38

View Full Product Details