Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7653282-7653782 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP1B2
Entrez Gene ID
482
Gene Name
ATPase Na+/K+ transporting subunit beta 2
Gene Aliases
AMOG
UniGene
Hs.643540
Gene Chromosome Location
Chr.17: 7646627-7657771 on Build GRCh38
Transcripts
NM_001303263.1 NM_001678.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2744413 c.227G>A p.R76Q Chr.17: 7653488-7653488 on Build GRCh38
984863 c.130T>G p.Y44D Chr.17: 7653391-7653391 on Build GRCh38
2744412 c.141T>A p.F47L Chr.17: 7653402-7653402 on Build GRCh38
4405369 c.138T>G p.V46V Chr.17: 7653399-7653399 on Build GRCh38
6298278 c.179G>T p.W60L Chr.17: 7653440-7653440 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749177856 Chr.17: 7653516-7653516 on Build GRCh38
rs764310229 Chr.17: 7653354-7653354 on Build GRCh38
rs770134651 Chr.17: 7653549-7653549 on Build GRCh38
rs777915470 Chr.17: 7653501-7653501 on Build GRCh38
rs937145780 Chr.17: 7653385-7653385 on Build GRCh38
rs751226790 Chr.17: 7653467-7653467 on Build GRCh38
rs141966039 Chr.17: 7653417-7653417 on Build GRCh38
rs960092356 Chr.17: 7653722-7653722 on Build GRCh38
rs928518935 Chr.17: 7653725-7653725 on Build GRCh38
rs763387316 Chr.17: 7653552-7653552 on Build GRCh38
rs981158707 Chr.17: 7653680-7653680 on Build GRCh38
rs569235346 Chr.17: 7653616-7653616 on Build GRCh38
rs951289943 Chr.17: 7653330-7653330 on Build GRCh38
rs746012048 Chr.17: 7653421-7653421 on Build GRCh38
rs370134148 Chr.17: 7653345-7653345 on Build GRCh38
rs747275097 Chr.17: 7653526-7653526 on Build GRCh38
rs1010869293 Chr.17: 7653563-7653563 on Build GRCh38
rs757958145 Chr.17: 7653462-7653462 on Build GRCh38
rs781061246 Chr.17: 7653480-7653480 on Build GRCh38
rs757868323 Chr.17: 7653392-7653392 on Build GRCh38
rs375583930 Chr.17: 7653493-7653493 on Build GRCh38
rs564913324 Chr.17: 7653411-7653411 on Build GRCh38
rs1001700981 Chr.17: 7653638-7653638 on Build GRCh38
rs904799170 Chr.17: 7653624-7653633 on Build GRCh38
rs765814772 Chr.17: 7653463-7653463 on Build GRCh38
rs199993423 Chr.17: 7653348-7653348 on Build GRCh38
rs776877483 Chr.17: 7653540-7653540 on Build GRCh38
rs749987809 Chr.17: 7653387-7653387 on Build GRCh38
rs768806571 Chr.17: 7653538-7653538 on Build GRCh38
rs759722465 Chr.17: 7653359-7653359 on Build GRCh38
rs889388451 Chr.17: 7653522-7653522 on Build GRCh38
rs111578256 Chr.17: 7653372-7653372 on Build GRCh38
rs962400354 Chr.17: 7653615-7653615 on Build GRCh38
rs1025430987 Chr.17: 7653644-7653644 on Build GRCh38
rs918254998 Chr.17: 7653307-7653307 on Build GRCh38
rs143282607 Chr.17: 7653554-7653554 on Build GRCh38
rs150658747 Chr.17: 7653418-7653418 on Build GRCh38
rs765777943 Chr.17: 7653444-7653444 on Build GRCh38
rs1034494530 Chr.17: 7653707-7653707 on Build GRCh38
rs757668749 Chr.17: 7653375-7653375 on Build GRCh38
rs548647471 Chr.17: 7653400-7653400 on Build GRCh38
rs370988866 Chr.17: 7653458-7653458 on Build GRCh38
rs371243052 Chr.17: 7653550-7653550 on Build GRCh38
rs997568142 Chr.17: 7653500-7653500 on Build GRCh38
rs112262799 Chr.17: 7653520-7653520 on Build GRCh38
rs35241187 Chr.17: 7653584-7653585 on Build GRCh38
rs760744091 Chr.17: 7653432-7653432 on Build GRCh38
rs747942023 Chr.17: 7653487-7653487 on Build GRCh38
rs550655804 Chr.17: 7653394-7653394 on Build GRCh38
rs754544873 Chr.17: 7653475-7653475 on Build GRCh38
rs762219437 Chr.17: 7653442-7653442 on Build GRCh38
rs536389520 Chr.17: 7653634-7653634 on Build GRCh38
rs755969286 Chr.17: 7653488-7653488 on Build GRCh38
rs951110450 Chr.17: 7653660-7653660 on Build GRCh38
rs762155123 Chr.17: 7653543-7653543 on Build GRCh38
rs553995552 Chr.17: 7653645-7653645 on Build GRCh38
rs928601983 Chr.17: 7653362-7653362 on Build GRCh38
rs866966044 Chr.17: 7653335-7653335 on Build GRCh38
rs772280682 Chr.17: 7653518-7653518 on Build GRCh38
rs376402102 Chr.17: 7653438-7653438 on Build GRCh38
rs74433844 Chr.17: 7653410-7653410 on Build GRCh38
rs548689117 Chr.17: 7653669-7653669 on Build GRCh38
rs780782316 Chr.17: 7653404-7653404 on Build GRCh38
rs922333387 Chr.17: 7653485-7653485 on Build GRCh38
rs41283405 Chr.17: 7653742-7653742 on Build GRCh38
rs376084687 Chr.17: 7653341-7653341 on Build GRCh38
rs775869812 Chr.17: 7653427-7653427 on Build GRCh38
rs764753710 Chr.17: 7653384-7653384 on Build GRCh38
rs776814725 Chr.17: 7653439-7653439 on Build GRCh38
rs183768284 Chr.17: 7653305-7653305 on Build GRCh38
rs267605074 Chr.17: 7653466-7653466 on Build GRCh38
rs771866868 Chr.17: 7653534-7653535 on Build GRCh38
rs754376564 Chr.17: 7653359-7653359 on Build GRCh38
rs1025567508 Chr.17: 7653736-7653736 on Build GRCh38
rs372597402 Chr.17: 7653397-7653397 on Build GRCh38
rs767892080 Chr.17: 7653338-7653338 on Build GRCh38
rs201156907 Chr.17: 7653422-7653422 on Build GRCh38

View Full Product Details