Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7653910-7654425 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP1B2
Entrez Gene ID
482
Gene Name
ATPase Na+/K+ transporting subunit beta 2
Gene Aliases
AMOG
UniGene
Hs.643540
Gene Chromosome Location
Chr.17: 7646627-7657771 on Build GRCh38
Transcripts
NM_001303263.1 NM_001678.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
984864 c.406G>A p.E136K Chr.17: 7654111-7654111 on Build GRCh38
6002456 c.484G>A p.G162S Chr.17: 7654189-7654189 on Build GRCh38
4130693 c.396A>G p.G132G Chr.17: 7654101-7654101 on Build GRCh38
88029 c.333C>T p.N111N Chr.17: 7653932-7653932 on Build GRCh38
5515609 c.353A>C p.N118T Chr.17: 7654058-7654058 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757566775 Chr.17: 7654093-7654093 on Build GRCh38
rs767518673 Chr.17: 7654059-7654059 on Build GRCh38
rs764893871 Chr.17: 7654275-7654276 on Build GRCh38
rs200669756 Chr.17: 7654209-7654209 on Build GRCh38
rs988009567 Chr.17: 7654326-7654326 on Build GRCh38
rs910671099 Chr.17: 7654397-7654397 on Build GRCh38
rs764000841 Chr.17: 7653938-7653938 on Build GRCh38
rs772681954 Chr.17: 7654099-7654099 on Build GRCh38
rs757450339 Chr.17: 7654157-7654157 on Build GRCh38
rs145660136 Chr.17: 7654147-7654147 on Build GRCh38
rs781759801 Chr.17: 7654280-7654280 on Build GRCh38
rs755496609 Chr.17: 7654276-7654276 on Build GRCh38
rs756282551 Chr.17: 7653991-7653991 on Build GRCh38
rs201606347 Chr.17: 7654188-7654188 on Build GRCh38
rs139562417 Chr.17: 7654111-7654111 on Build GRCh38
rs749693939 Chr.17: 7653994-7653994 on Build GRCh38
rs774286082 Chr.17: 7654219-7654219 on Build GRCh38
rs754089848 Chr.17: 7654149-7654149 on Build GRCh38
rs749499489 Chr.17: 7654080-7654080 on Build GRCh38
rs542414922 Chr.17: 7654177-7654177 on Build GRCh38
rs1052190576 Chr.17: 7654057-7654057 on Build GRCh38
rs904747577 Chr.17: 7654136-7654136 on Build GRCh38
rs750336902 Chr.17: 7654050-7654050 on Build GRCh38
rs781269923 Chr.17: 7653983-7653983 on Build GRCh38
rs987513139 Chr.17: 7654384-7654384 on Build GRCh38
rs747736011 Chr.17: 7654288-7654288 on Build GRCh38
rs761465658 Chr.17: 7654112-7654112 on Build GRCh38
rs899452039 Chr.17: 7654174-7654174 on Build GRCh38
rs576911066 Chr.17: 7654046-7654046 on Build GRCh38
rs770644298 Chr.17: 7653945-7653945 on Build GRCh38
rs144189143 Chr.17: 7654131-7654131 on Build GRCh38
rs753861402 Chr.17: 7653953-7653958 on Build GRCh38
rs765559894 Chr.17: 7654256-7654256 on Build GRCh38
rs1031509385 Chr.17: 7654368-7654368 on Build GRCh38
rs759813372 Chr.17: 7654116-7654116 on Build GRCh38
rs774734165 Chr.17: 7654005-7654005 on Build GRCh38
rs762635037 Chr.17: 7654143-7654143 on Build GRCh38
rs763876172 Chr.17: 7654022-7654022 on Build GRCh38
rs758319381 Chr.17: 7654056-7654056 on Build GRCh38
rs758800668 Chr.17: 7654267-7654267 on Build GRCh38
rs894146510 Chr.17: 7654273-7654273 on Build GRCh38
rs968681255 Chr.17: 7654329-7654329 on Build GRCh38
rs777540555 Chr.17: 7654297-7654297 on Build GRCh38
rs558550793 Chr.17: 7654011-7654011 on Build GRCh38
rs376672732 Chr.17: 7653966-7653966 on Build GRCh38
rs757083819 Chr.17: 7653960-7653960 on Build GRCh38
rs752825639 Chr.17: 7654060-7654060 on Build GRCh38
rs761362366 Chr.17: 7654273-7654274 on Build GRCh38
rs780503833 Chr.17: 7654269-7654269 on Build GRCh38
rs977627833 Chr.17: 7654340-7654340 on Build GRCh38
rs769184866 Chr.17: 7654294-7654294 on Build GRCh38
rs190796379 Chr.17: 7654360-7654360 on Build GRCh38
rs753732257 Chr.17: 7653946-7653946 on Build GRCh38
rs148893862 Chr.17: 7654148-7654148 on Build GRCh38
rs1642763 Chr.17: 7654101-7654101 on Build GRCh38
rs746382385 Chr.17: 7654096-7654096 on Build GRCh38
rs576295919 Chr.17: 7653972-7653972 on Build GRCh38
rs750365302 Chr.17: 7653969-7653969 on Build GRCh38
rs1002930865 Chr.17: 7654058-7654058 on Build GRCh38
rs773014135 Chr.17: 7654128-7654128 on Build GRCh38
rs772711589 Chr.17: 7654206-7654206 on Build GRCh38
rs36041343 Chr.17: 7654312-7654312 on Build GRCh38
rs779294665 Chr.17: 7654094-7654094 on Build GRCh38
rs367751526 Chr.17: 7654014-7654014 on Build GRCh38
rs943629286 Chr.17: 7653948-7653948 on Build GRCh38
rs753998361 Chr.17: 7654247-7654247 on Build GRCh38
rs768263993 Chr.17: 7654102-7654102 on Build GRCh38
rs1037909144 Chr.17: 7654169-7654169 on Build GRCh38
rs1017362714 Chr.17: 7654224-7654224 on Build GRCh38
rs764974809 Chr.17: 7654114-7654114 on Build GRCh38
rs758682764 Chr.17: 7654165-7654165 on Build GRCh38
rs141427807 Chr.17: 7653996-7653996 on Build GRCh38
rs764180280 Chr.17: 7654243-7654243 on Build GRCh38
rs913630842 Chr.17: 7654327-7654327 on Build GRCh38
rs372215828 Chr.17: 7654161-7654161 on Build GRCh38
rs761754748 Chr.17: 7654037-7654037 on Build GRCh38
rs199816243 Chr.17: 7654275-7654275 on Build GRCh38
rs748403475 Chr.17: 7653984-7653984 on Build GRCh38
rs776415142 Chr.17: 7654030-7654030 on Build GRCh38
rs150813046 Chr.17: 7654376-7654376 on Build GRCh38
rs776106709 Chr.17: 7654100-7654100 on Build GRCh38
rs376750521 Chr.17: 7654164-7654164 on Build GRCh38
rs780761413 Chr.17: 7654166-7654166 on Build GRCh38
rs142627831 Chr.17: 7654193-7654193 on Build GRCh38
rs759427489 Chr.17: 7654222-7654222 on Build GRCh38
rs34745087 Chr.17: 7654076-7654076 on Build GRCh38
rs777951083 Chr.17: 7653992-7653992 on Build GRCh38
rs775090279 Chr.17: 7654018-7654018 on Build GRCh38
rs955182701 Chr.17: 7654319-7654319 on Build GRCh38
rs761764774 Chr.17: 7654251-7654251 on Build GRCh38
rs750593994 Chr.17: 7654262-7654262 on Build GRCh38
rs747456774 Chr.17: 7654173-7654173 on Build GRCh38

View Full Product Details