Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7709457-7709962 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFNB3
Entrez Gene ID
1949
Gene Name
ephrin B3
Gene Aliases
EFL6, EPLG8, LERK8
UniGene
Hs.26988
Gene Chromosome Location
Chr.17: 7705202-7711375 on Build GRCh38
Transcripts
NM_001406.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6652099 c.972C>T p.I324I Chr.17: 7709525-7709525 on Build GRCh38
6652091 c.984_985insCC p.Q331fs*>11 Chr.17: 7709537-7709538 on Build GRCh38
6652092 c.919delT p.F307fs*8 Chr.17: 7709472-7709472 on Build GRCh38
4680344 c.968A>G p.Y323C Chr.17: 7709521-7709521 on Build GRCh38
1195307 c.995G>A p.S332N Chr.17: 7709548-7709548 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200403710 Chr.17: 7709611-7709611 on Build GRCh38
rs940142007 Chr.17: 7709918-7709918 on Build GRCh38
rs201191552 Chr.17: 7709590-7709590 on Build GRCh38
rs890488245 Chr.17: 7709898-7709898 on Build GRCh38
rs765897899 Chr.17: 7709591-7709591 on Build GRCh38
rs954220758 Chr.17: 7709752-7709752 on Build GRCh38
rs753413666 Chr.17: 7709502-7709502 on Build GRCh38
rs754767367 Chr.17: 7709601-7709601 on Build GRCh38
rs1009150307 Chr.17: 7709900-7709900 on Build GRCh38
rs758119966 Chr.17: 7709522-7709522 on Build GRCh38
rs750150083 Chr.17: 7709519-7709519 on Build GRCh38
rs965286746 Chr.17: 7709593-7709593 on Build GRCh38
rs548753118 Chr.17: 7709618-7709618 on Build GRCh38
rs541725555 Chr.17: 7709575-7709575 on Build GRCh38
rs1054815334 Chr.17: 7709670-7709672 on Build GRCh38
rs752684896 Chr.17: 7709608-7709608 on Build GRCh38
rs925043269 Chr.17: 7709655-7709655 on Build GRCh38
rs762414905 Chr.17: 7709624-7709624 on Build GRCh38
rs565882256 Chr.17: 7709704-7709704 on Build GRCh38
rs894738055 Chr.17: 7709480-7709480 on Build GRCh38
rs552194549 Chr.17: 7709771-7709771 on Build GRCh38
rs894825443 Chr.17: 7709670-7709670 on Build GRCh38
rs759736010 Chr.17: 7709477-7709477 on Build GRCh38
rs962663238 Chr.17: 7709550-7709550 on Build GRCh38
rs536550806 Chr.17: 7709717-7709717 on Build GRCh38
rs547278978 Chr.17: 7709683-7709683 on Build GRCh38
rs1026711448 Chr.17: 7709932-7709932 on Build GRCh38
rs1008143669 Chr.17: 7709770-7709770 on Build GRCh38
rs3744264 Chr.17: 7709929-7709929 on Build GRCh38
rs769239419 Chr.17: 7709570-7709570 on Build GRCh38
rs1013002598 Chr.17: 7709481-7709481 on Build GRCh38
rs372108937 Chr.17: 7709903-7709903 on Build GRCh38
rs754963786 Chr.17: 7709533-7709533 on Build GRCh38
rs531265694 Chr.17: 7709667-7709667 on Build GRCh38
rs747225977 Chr.17: 7709621-7709621 on Build GRCh38
rs761368685 Chr.17: 7709507-7709507 on Build GRCh38
rs916238924 Chr.17: 7709887-7709888 on Build GRCh38
rs752828539 Chr.17: 7709477-7709479 on Build GRCh38
rs746159356 Chr.17: 7709551-7709551 on Build GRCh38
rs375108616 Chr.17: 7709526-7709526 on Build GRCh38
rs760901575 Chr.17: 7709563-7709563 on Build GRCh38
rs549481492 Chr.17: 7709668-7709668 on Build GRCh38
rs1041409807 Chr.17: 7709787-7709787 on Build GRCh38
rs376721844 Chr.17: 7709622-7709622 on Build GRCh38
rs151125738 Chr.17: 7709496-7709496 on Build GRCh38
rs764898132 Chr.17: 7709583-7709583 on Build GRCh38
rs576397534 Chr.17: 7709799-7709799 on Build GRCh38
rs778973387 Chr.17: 7709610-7709610 on Build GRCh38
rs781015860 Chr.17: 7709605-7709605 on Build GRCh38
rs148494000 Chr.17: 7709680-7709680 on Build GRCh38
rs17854241 Chr.17: 7709543-7709543 on Build GRCh38
rs764810191 Chr.17: 7709517-7709517 on Build GRCh38
rs369584529 Chr.17: 7709880-7709880 on Build GRCh38
rs1015725440 Chr.17: 7709541-7709541 on Build GRCh38
rs775709769 Chr.17: 7709493-7709493 on Build GRCh38
rs375405308 Chr.17: 7709915-7709915 on Build GRCh38
rs780101009 Chr.17: 7709617-7709617 on Build GRCh38
rs900635559 Chr.17: 7709914-7709914 on Build GRCh38
rs756422375 Chr.17: 7709537-7709538 on Build GRCh38
rs767932268 Chr.17: 7709479-7709479 on Build GRCh38
rs182299356 Chr.17: 7709784-7709784 on Build GRCh38
rs952328044 Chr.17: 7709671-7709671 on Build GRCh38
rs779903044 Chr.17: 7709525-7709525 on Build GRCh38
rs1053037537 Chr.17: 7709811-7709811 on Build GRCh38
rs775940145 Chr.17: 7709554-7709554 on Build GRCh38
rs1004386410 Chr.17: 7709685-7709685 on Build GRCh38
rs751400261 Chr.17: 7709596-7709596 on Build GRCh38
rs376436975 Chr.17: 7709589-7709589 on Build GRCh38
rs532773147 Chr.17: 7709538-7709538 on Build GRCh38
rs957792101 Chr.17: 7709857-7709857 on Build GRCh38
rs991003038 Chr.17: 7709877-7709877 on Build GRCh38
rs537513542 Chr.17: 7709904-7709904 on Build GRCh38
rs567257903 Chr.17: 7709631-7709631 on Build GRCh38
rs1057428671 Chr.17: 7709716-7709716 on Build GRCh38
rs1043097128 Chr.17: 7709778-7709778 on Build GRCh38
rs925008672 Chr.17: 7709827-7709827 on Build GRCh38
rs780949918 Chr.17: 7709534-7709534 on Build GRCh38
rs762388499 Chr.17: 7709579-7709579 on Build GRCh38
rs1015651869 Chr.17: 7709687-7709687 on Build GRCh38
rs34423450 Chr.17: 7709497-7709498 on Build GRCh38
rs796071978 Chr.17: 7709536-7709536 on Build GRCh38

View Full Product Details