Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7936617-7937127 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTROB
Entrez Gene ID
116840
Gene Name
centrobin, centriole duplication and spindle assembly protein
Gene Aliases
LIP8, PP1221
UniGene
Hs.348012
Gene Chromosome Location
Chr.17: 7932054-7949919 on Build GRCh38
Transcripts
NM_001037144.5 NM_053051.3 XM_005256437.2 XM_005256438.2 XM_005256439.2 XM_017024128.1 XM_017024129.1 XM_017024130.1 XM_017024131.1 XM_017024132.1 XM_017024133.1 XM_017024134.1 XM_017024135.1 XM_017024136.1 XM_017024137.1 XM_017024138.1 XM_017024139.1 XM_017024140.1 XM_017024141.1 XM_017024142.1 XM_017024143.1 XM_017024144.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5895021 c.763G>A p.E255K Chr.17: 7936752-7936752 on Build GRCh38
1290778 c.793C>T p.Q265* Chr.17: 7936782-7936782 on Build GRCh38
6632055 c.722G>A p.R241H Chr.17: 7936711-7936711 on Build GRCh38
404433 c.758G>A p.R253Q Chr.17: 7936747-7936747 on Build GRCh38
1661211 c.754G>A p.E252K Chr.17: 7936743-7936743 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1043566220 Chr.17: 7936978-7936978 on Build GRCh38
rs761593379 Chr.17: 7937055-7937055 on Build GRCh38
rs935419492 Chr.17: 7937046-7937046 on Build GRCh38
rs765343755 Chr.17: 7936801-7936801 on Build GRCh38
rs1033687724 Chr.17: 7936993-7936993 on Build GRCh38
rs924045456 Chr.17: 7937006-7937006 on Build GRCh38
rs755666907 Chr.17: 7936657-7936658 on Build GRCh38
rs576711195 Chr.17: 7936711-7936711 on Build GRCh38
rs991147041 Chr.17: 7936917-7936917 on Build GRCh38
rs780505947 Chr.17: 7936737-7936737 on Build GRCh38
rs917869448 Chr.17: 7937074-7937074 on Build GRCh38
rs929189314 Chr.17: 7936718-7936718 on Build GRCh38
rs769508213 Chr.17: 7937068-7937068 on Build GRCh38
rs974206540 Chr.17: 7936646-7936646 on Build GRCh38
rs142384576 Chr.17: 7936927-7936927 on Build GRCh38
rs766121699 Chr.17: 7936687-7936687 on Build GRCh38
rs751371218 Chr.17: 7936692-7936692 on Build GRCh38
rs758606094 Chr.17: 7936824-7936824 on Build GRCh38
rs567020473 Chr.17: 7937040-7937040 on Build GRCh38
rs759220534 Chr.17: 7936770-7936770 on Build GRCh38
rs781636883 Chr.17: 7936848-7936848 on Build GRCh38
rs777376547 Chr.17: 7936693-7936694 on Build GRCh38
rs747883280 Chr.17: 7936760-7936760 on Build GRCh38
rs759436313 Chr.17: 7936694-7936694 on Build GRCh38
rs146753401 Chr.17: 7936732-7936732 on Build GRCh38
rs761221006 Chr.17: 7936668-7936668 on Build GRCh38
rs946566009 Chr.17: 7936956-7936956 on Build GRCh38
rs560112219 Chr.17: 7936958-7936958 on Build GRCh38
rs549067805 Chr.17: 7937013-7937013 on Build GRCh38
rs752144478 Chr.17: 7936839-7936839 on Build GRCh38
rs776399151 Chr.17: 7936653-7936653 on Build GRCh38
rs145425422 Chr.17: 7936779-7936779 on Build GRCh38
rs149757324 Chr.17: 7936731-7936731 on Build GRCh38
rs927634765 Chr.17: 7936780-7936780 on Build GRCh38
rs959405711 Chr.17: 7937030-7937030 on Build GRCh38
rs769333123 Chr.17: 7936682-7936682 on Build GRCh38
rs753961644 Chr.17: 7936788-7936788 on Build GRCh38
rs773110986 Chr.17: 7936684-7936684 on Build GRCh38
rs1027392714 Chr.17: 7936738-7936738 on Build GRCh38
rs764218484 Chr.17: 7936703-7936703 on Build GRCh38
rs370271101 Chr.17: 7936767-7936767 on Build GRCh38
rs765994475 Chr.17: 7936739-7936739 on Build GRCh38
rs532835734 Chr.17: 7936916-7936916 on Build GRCh38
rs866611668 Chr.17: 7937003-7937003 on Build GRCh38
rs544026241 Chr.17: 7936733-7936733 on Build GRCh38
rs762898145 Chr.17: 7936686-7936686 on Build GRCh38
rs375449180 Chr.17: 7936769-7936769 on Build GRCh38
rs116322830 Chr.17: 7936747-7936747 on Build GRCh38
rs769245057 Chr.17: 7936765-7936765 on Build GRCh38
rs754131227 Chr.17: 7936710-7936710 on Build GRCh38
rs780674173 Chr.17: 7936754-7936754 on Build GRCh38
rs186330280 Chr.17: 7936970-7936970 on Build GRCh38
rs571965958 Chr.17: 7936690-7936690 on Build GRCh38
rs368751901 Chr.17: 7936796-7936796 on Build GRCh38
rs750546865 Chr.17: 7936823-7936823 on Build GRCh38
rs747712837 Chr.17: 7936869-7936869 on Build GRCh38
rs997179755 Chr.17: 7936808-7936808 on Build GRCh38
rs992113253 Chr.17: 7937053-7937053 on Build GRCh38
rs375264706 Chr.17: 7936827-7936827 on Build GRCh38
rs968217923 Chr.17: 7936953-7936953 on Build GRCh38
rs769127193 Chr.17: 7936750-7936750 on Build GRCh38
rs755275298 Chr.17: 7936840-7936840 on Build GRCh38

View Full Product Details