Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 8038935-8039436 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALOX15B
Entrez Gene ID
247
Gene Name
arachidonate 15-lipoxygenase, type B
Gene Aliases
15-LOX-2
UniGene
Hs.111256
Gene Chromosome Location
Chr.17: 8039040-8049134 on Build GRCh38
Transcripts
NM_001039130.1 NM_001039131.1 NM_001141.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3523594 c.129G>A p.K43K Chr.17: 8039284-8039284 on Build GRCh38
3523593 c.16G>A p.V6I Chr.17: 8039171-8039171 on Build GRCh38
4564074 c.125_126GC>TT p.G42V Chr.17: 8039280-8039281 on Build GRCh38
5921084 c.92G>A p.G31E Chr.17: 8039247-8039247 on Build GRCh38
1235879 c.170T>C p.L57P Chr.17: 8039408-8039408 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1003212777 Chr.17: 8039096-8039096 on Build GRCh38
rs62065077 Chr.17: 8039107-8039107 on Build GRCh38
rs974608867 Chr.17: 8039322-8039322 on Build GRCh38
rs202018712 Chr.17: 8039111-8039111 on Build GRCh38
rs773211140 Chr.17: 8039271-8039271 on Build GRCh38
rs545455096 Chr.17: 8039296-8039296 on Build GRCh38
rs950334791 Chr.17: 8039088-8039088 on Build GRCh38
rs1025919809 Chr.17: 8039080-8039080 on Build GRCh38
rs975038804 Chr.17: 8039096-8039096 on Build GRCh38
rs754522077 Chr.17: 8039115-8039115 on Build GRCh38
rs983375574 Chr.17: 8038963-8038963 on Build GRCh38
rs771296412 Chr.17: 8039274-8039274 on Build GRCh38
rs190780534 Chr.17: 8038966-8038966 on Build GRCh38
rs1007015740 Chr.17: 8039028-8039028 on Build GRCh38
rs73972649 Chr.17: 8039116-8039116 on Build GRCh38
rs780972862 Chr.17: 8039247-8039248 on Build GRCh38
rs374598646 Chr.17: 8039209-8039209 on Build GRCh38
rs985140713 Chr.17: 8039342-8039342 on Build GRCh38
rs762517375 Chr.17: 8039293-8039294 on Build GRCh38
rs774901569 Chr.17: 8039286-8039286 on Build GRCh38
rs778939697 Chr.17: 8039227-8039227 on Build GRCh38
rs533198158 Chr.17: 8039098-8039098 on Build GRCh38
rs769860982 Chr.17: 8039162-8039162 on Build GRCh38
rs373549852 Chr.17: 8039021-8039021 on Build GRCh38
rs879338790 Chr.17: 8038969-8038969 on Build GRCh38
rs771016636 Chr.17: 8039364-8039364 on Build GRCh38
rs376917454 Chr.17: 8039126-8039126 on Build GRCh38
rs772714560 Chr.17: 8039371-8039371 on Build GRCh38
rs923919152 Chr.17: 8038999-8038999 on Build GRCh38
rs769805973 Chr.17: 8039253-8039254 on Build GRCh38
rs762070324 Chr.17: 8039158-8039158 on Build GRCh38
rs373389332 Chr.17: 8039129-8039129 on Build GRCh38
rs1040316227 Chr.17: 8039024-8039025 on Build GRCh38
rs956782910 Chr.17: 8038973-8038973 on Build GRCh38
rs1035084300 Chr.17: 8039355-8039355 on Build GRCh38
rs773338352 Chr.17: 8039177-8039177 on Build GRCh38
rs750342751 Chr.17: 8039344-8039344 on Build GRCh38
rs911711078 Chr.17: 8039061-8039061 on Build GRCh38
rs752370434 Chr.17: 8039201-8039201 on Build GRCh38
rs200119128 Chr.17: 8039381-8039381 on Build GRCh38
rs371780485 Chr.17: 8039324-8039324 on Build GRCh38
rs760343955 Chr.17: 8039317-8039317 on Build GRCh38
rs745678525 Chr.17: 8039146-8039146 on Build GRCh38
rs776861437 Chr.17: 8039157-8039157 on Build GRCh38
rs753399540 Chr.17: 8039214-8039214 on Build GRCh38
rs758265025 Chr.17: 8039410-8039410 on Build GRCh38
rs747925269 Chr.17: 8039254-8039254 on Build GRCh38
rs550742047 Chr.17: 8039048-8039048 on Build GRCh38
rs867086223 Chr.17: 8039205-8039205 on Build GRCh38
rs141930531 Chr.17: 8039244-8039244 on Build GRCh38
rs187841999 Chr.17: 8039050-8039050 on Build GRCh38
rs200798486 Chr.17: 8039353-8039353 on Build GRCh38
rs766337032 Chr.17: 8039411-8039411 on Build GRCh38
rs376534256 Chr.17: 8039152-8039152 on Build GRCh38
rs749916428 Chr.17: 8039406-8039406 on Build GRCh38
rs746369168 Chr.17: 8039368-8039368 on Build GRCh38
rs773956625 Chr.17: 8039134-8039134 on Build GRCh38
rs777404728 Chr.17: 8039211-8039211 on Build GRCh38
rs757072591 Chr.17: 8039219-8039219 on Build GRCh38
rs370310588 Chr.17: 8039185-8039185 on Build GRCh38
rs556299634 Chr.17: 8039087-8039087 on Build GRCh38
rs528887481 Chr.17: 8039032-8039032 on Build GRCh38
rs867172889 Chr.17: 8039275-8039275 on Build GRCh38
rs760005308 Chr.17: 8039300-8039300 on Build GRCh38
rs896632252 Chr.17: 8039151-8039151 on Build GRCh38
rs577729599 Chr.17: 8039264-8039264 on Build GRCh38
rs1042307493 Chr.17: 8038976-8038976 on Build GRCh38
rs747324375 Chr.17: 8039136-8039146 on Build GRCh38
rs748782371 Chr.17: 8039341-8039341 on Build GRCh38
rs763312932 Chr.17: 8039184-8039184 on Build GRCh38
rs763789688 Chr.17: 8039386-8039386 on Build GRCh38
rs746813261 Chr.17: 8039357-8039357 on Build GRCh38
rs386795064 Chr.17: 8038966-8038968 on Build GRCh38
rs751307987 Chr.17: 8039418-8039418 on Build GRCh38
rs145035264 Chr.17: 8039198-8039198 on Build GRCh38
rs149287751 Chr.17: 8039193-8039193 on Build GRCh38
rs1051096611 Chr.17: 8039011-8039015 on Build GRCh38
rs138830954 Chr.17: 8039241-8039241 on Build GRCh38
rs183507946 Chr.17: 8038968-8038968 on Build GRCh38
rs1030951062 Chr.17: 8039237-8039237 on Build GRCh38
rs368392738 Chr.17: 8039161-8039161 on Build GRCh38
rs922523809 Chr.17: 8039061-8039062 on Build GRCh38
rs776322613 Chr.17: 8039397-8039397 on Build GRCh38
rs368652352 Chr.17: 8039372-8039372 on Build GRCh38
rs774365211 Chr.17: 8039365-8039365 on Build GRCh38
rs367623091 Chr.17: 8039272-8039272 on Build GRCh38
rs768771995 Chr.17: 8039169-8039169 on Build GRCh38
rs934003961 Chr.17: 8039007-8039007 on Build GRCh38
rs967819297 Chr.17: 8039415-8039415 on Build GRCh38
rs769964785 Chr.17: 8039358-8039358 on Build GRCh38
rs773326725 Chr.17: 8039263-8039264 on Build GRCh38
rs370620348 Chr.17: 8039243-8039243 on Build GRCh38
rs867207144 Chr.17: 8039233-8039233 on Build GRCh38
rs758388153 Chr.17: 8039346-8039346 on Build GRCh38
rs763839484 Chr.17: 8039319-8039319 on Build GRCh38
rs761412865 Chr.17: 8039405-8039405 on Build GRCh38
rs767993975 Chr.17: 8039311-8039311 on Build GRCh38
rs755666587 Chr.17: 8039128-8039128 on Build GRCh38
rs915039621 Chr.17: 8039104-8039104 on Build GRCh38
rs756812079 Chr.17: 8039337-8039337 on Build GRCh38
rs1001561370 Chr.17: 8039217-8039217 on Build GRCh38
rs930268710 Chr.17: 8039099-8039099 on Build GRCh38
rs148000629 Chr.17: 8039200-8039200 on Build GRCh38
rs753167346 Chr.17: 8039314-8039314 on Build GRCh38
rs770957424 Chr.17: 8039130-8039130 on Build GRCh38

View Full Product Details