Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 8189718-8190226 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BORCS6
Entrez Gene ID
54785
Gene Name
BLOC-1 related complex subunit 6
Gene Aliases
C17orf59, PRO2472
UniGene
Hs.129563
Gene Chromosome Location
Chr.17: 8188333-8190246 on Build GRCh38
Transcripts
NM_017622.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
381147 c.389G>A p.G130E Chr.17: 8189752-8189752 on Build GRCh38
6573512 c.101C>G p.S34C Chr.17: 8190040-8190040 on Build GRCh38
6573511 c.174C>G p.P58P Chr.17: 8189967-8189967 on Build GRCh38
4070976 c.421G>C p.E141Q Chr.17: 8189720-8189720 on Build GRCh38
4070977 c.19G>C p.E7Q Chr.17: 8189720-8189720 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773853354 Chr.17: 8189811-8189811 on Build GRCh38
rs971201397 Chr.17: 8190012-8190012 on Build GRCh38
rs370859187 Chr.17: 8189742-8189742 on Build GRCh38
rs150061068 Chr.17: 8190006-8190006 on Build GRCh38
rs981784327 Chr.17: 8190015-8190015 on Build GRCh38
rs1006528441 Chr.17: 8190000-8190000 on Build GRCh38
rs751749586 Chr.17: 8189831-8189831 on Build GRCh38
rs927205332 Chr.17: 8190019-8190019 on Build GRCh38
rs1022227342 Chr.17: 8190133-8190133 on Build GRCh38
rs758504849 Chr.17: 8189775-8189775 on Build GRCh38
rs759498783 Chr.17: 8189755-8189755 on Build GRCh38
rs745448908 Chr.17: 8189929-8189929 on Build GRCh38
rs749708322 Chr.17: 8190166-8190166 on Build GRCh38
rs533276759 Chr.17: 8189854-8189854 on Build GRCh38
rs750447631 Chr.17: 8189774-8189774 on Build GRCh38
rs994589786 Chr.17: 8189996-8189996 on Build GRCh38
rs781400698 Chr.17: 8189797-8189797 on Build GRCh38
rs966338039 Chr.17: 8190137-8190137 on Build GRCh38
rs772963022 Chr.17: 8189803-8189803 on Build GRCh38
rs775449715 Chr.17: 8189949-8189949 on Build GRCh38
rs374315817 Chr.17: 8189745-8189745 on Build GRCh38
rs911460700 Chr.17: 8189952-8189952 on Build GRCh38
rs748434754 Chr.17: 8189851-8189851 on Build GRCh38
rs907306609 Chr.17: 8190056-8190056 on Build GRCh38
rs748615756 Chr.17: 8189800-8189800 on Build GRCh38
rs1033962336 Chr.17: 8190099-8190099 on Build GRCh38
rs748869780 Chr.17: 8189886-8189886 on Build GRCh38
rs778027585 Chr.17: 8189856-8189856 on Build GRCh38
rs115424454 Chr.17: 8189834-8189834 on Build GRCh38
rs776305336 Chr.17: 8190053-8190053 on Build GRCh38
rs779261793 Chr.17: 8189772-8189772 on Build GRCh38
rs918454116 Chr.17: 8190011-8190011 on Build GRCh38
rs759408918 Chr.17: 8189813-8189813 on Build GRCh38
rs922357133 Chr.17: 8189900-8189900 on Build GRCh38
rs766776685 Chr.17: 8190067-8190067 on Build GRCh38
rs528851609 Chr.17: 8189818-8189818 on Build GRCh38
rs183207665 Chr.17: 8189922-8189922 on Build GRCh38
rs763883871 Chr.17: 8190026-8190026 on Build GRCh38
rs537803711 Chr.17: 8190105-8190105 on Build GRCh38
rs747455049 Chr.17: 8189784-8189784 on Build GRCh38
rs978178702 Chr.17: 8190145-8190145 on Build GRCh38
rs752429991 Chr.17: 8189816-8189816 on Build GRCh38
rs750619625 Chr.17: 8189820-8189820 on Build GRCh38
rs959307403 Chr.17: 8189838-8189838 on Build GRCh38
rs202045414 Chr.17: 8190110-8190110 on Build GRCh38
rs755540795 Chr.17: 8189832-8189832 on Build GRCh38
rs902041308 Chr.17: 8190116-8190116 on Build GRCh38
rs962873911 Chr.17: 8189799-8189799 on Build GRCh38
rs1041770600 Chr.17: 8190104-8190104 on Build GRCh38
rs756897380 Chr.17: 8190180-8190180 on Build GRCh38
rs893187065 Chr.17: 8189879-8189879 on Build GRCh38
rs769552118 Chr.17: 8189744-8189744 on Build GRCh38
rs902011955 Chr.17: 8189916-8189916 on Build GRCh38
rs1038592879 Chr.17: 8189976-8189976 on Build GRCh38
rs562595644 Chr.17: 8189853-8189853 on Build GRCh38
rs777142396 Chr.17: 8189741-8189741 on Build GRCh38
rs778576787 Chr.17: 8190187-8190187 on Build GRCh38
rs937334521 Chr.17: 8189850-8189850 on Build GRCh38
rs757406881 Chr.17: 8189762-8189762 on Build GRCh38
rs972470809 Chr.17: 8190008-8190008 on Build GRCh38
rs755161699 Chr.17: 8190098-8190098 on Build GRCh38
rs895058967 Chr.17: 8190109-8190109 on Build GRCh38
rs780313893 Chr.17: 8189783-8189783 on Build GRCh38
rs779850055 Chr.17: 8190111-8190111 on Build GRCh38
rs955328605 Chr.17: 8190169-8190169 on Build GRCh38
rs751446913 Chr.17: 8189749-8189749 on Build GRCh38
rs941655972 Chr.17: 8189964-8189964 on Build GRCh38
rs1010788257 Chr.17: 8190125-8190125 on Build GRCh38
rs561853521 Chr.17: 8189815-8189815 on Build GRCh38
rs753205201 Chr.17: 8190108-8190108 on Build GRCh38
rs1049980190 Chr.17: 8189792-8189792 on Build GRCh38
rs989136980 Chr.17: 8189849-8189849 on Build GRCh38
rs911350774 Chr.17: 8190186-8190186 on Build GRCh38
rs753966049 Chr.17: 8189748-8189748 on Build GRCh38
rs752549079 Chr.17: 8189758-8189758 on Build GRCh38
rs1050371374 Chr.17: 8189930-8189930 on Build GRCh38
rs937347980 Chr.17: 8190049-8190049 on Build GRCh38
rs765064766 Chr.17: 8189858-8189858 on Build GRCh38
rs946169180 Chr.17: 8190102-8190102 on Build GRCh38
rs770582281 Chr.17: 8189898-8189898 on Build GRCh38
rs867022836 Chr.17: 8190035-8190035 on Build GRCh38
rs760558175 Chr.17: 8189817-8189817 on Build GRCh38
rs551741051 Chr.17: 8189870-8189870 on Build GRCh38
rs767230907 Chr.17: 8189756-8189756 on Build GRCh38
rs763257512 Chr.17: 8190059-8190059 on Build GRCh38
rs758700053 Chr.17: 8189821-8189821 on Build GRCh38
rs527516176 Chr.17: 8189939-8189939 on Build GRCh38
rs966367446 Chr.17: 8189868-8189868 on Build GRCh38
rs538941740 Chr.17: 8190202-8190202 on Build GRCh38
rs567768810 Chr.17: 8190074-8190074 on Build GRCh38
rs780227387 Chr.17: 8189827-8189827 on Build GRCh38
rs770384390 Chr.17: 8189805-8189805 on Build GRCh38
rs769302085 Chr.17: 8189788-8189788 on Build GRCh38
rs977833038 Chr.17: 8189897-8189897 on Build GRCh38
rs897129943 Chr.17: 8189991-8189991 on Build GRCh38
rs188696819 Chr.17: 8190161-8190161 on Build GRCh38

View Full Product Details