Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 8312321-8312831 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF15
Entrez Gene ID
22899
Gene Name
Rho guanine nucleotide exchange factor 15
Gene Aliases
ARGEF15, E5, Ephexin5, Vsm-RhoGEF
UniGene
Hs.443109
Gene Chromosome Location
Chr.17: 8310238-8322516 on Build GRCh38
Transcripts
NM_025014.1 NM_173728.3 XM_011523734.1 XM_011523735.1 XM_011523736.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5925904 c.406G>A p.G136R Chr.17: 8312445-8312445 on Build GRCh38
3523969 c.328C>T p.P110S Chr.17: 8312367-8312367 on Build GRCh38
6496139 c.451C>T p.R151W Chr.17: 8312490-8312490 on Build GRCh38
5574397 c.478G>T p.G160C Chr.17: 8312517-8312517 on Build GRCh38
4440486 c.326delC p.P110fs*72 Chr.17: 8312365-8312365 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs890547379 Chr.17: 8312536-8312536 on Build GRCh38
rs375648145 Chr.17: 8312352-8312352 on Build GRCh38
rs2313147 Chr.17: 8312643-8312643 on Build GRCh38
rs746183719 Chr.17: 8312641-8312641 on Build GRCh38
rs757098576 Chr.17: 8312414-8312414 on Build GRCh38
rs759261231 Chr.17: 8312794-8312794 on Build GRCh38
rs752615298 Chr.17: 8312406-8312406 on Build GRCh38
rs774127729 Chr.17: 8312791-8312791 on Build GRCh38
rs752849043 Chr.17: 8312669-8312669 on Build GRCh38
rs1041089036 Chr.17: 8312689-8312689 on Build GRCh38
rs1016145702 Chr.17: 8312785-8312785 on Build GRCh38
rs776793274 Chr.17: 8312586-8312586 on Build GRCh38
rs753845921 Chr.17: 8312502-8312502 on Build GRCh38
rs773310104 Chr.17: 8312662-8312662 on Build GRCh38
rs938432141 Chr.17: 8312627-8312627 on Build GRCh38
rs146482304 Chr.17: 8312356-8312356 on Build GRCh38
rs17857129 Chr.17: 8312503-8312503 on Build GRCh38
rs147736312 Chr.17: 8312551-8312551 on Build GRCh38
rs760064314 Chr.17: 8312568-8312568 on Build GRCh38
rs753858130 Chr.17: 8312687-8312687 on Build GRCh38
rs1027304853 Chr.17: 8312600-8312600 on Build GRCh38
rs751244420 Chr.17: 8312398-8312398 on Build GRCh38
rs753519754 Chr.17: 8312412-8312412 on Build GRCh38
rs550687567 Chr.17: 8312543-8312543 on Build GRCh38
rs372888591 Chr.17: 8312552-8312552 on Build GRCh38
rs761415894 Chr.17: 8312656-8312656 on Build GRCh38
rs775232310 Chr.17: 8312557-8312557 on Build GRCh38
rs777250745 Chr.17: 8312379-8312379 on Build GRCh38
rs768290924 Chr.17: 8312736-8312736 on Build GRCh38
rs62637603 Chr.17: 8312609-8312609 on Build GRCh38
rs747278375 Chr.17: 8312425-8312425 on Build GRCh38
rs774975398 Chr.17: 8312645-8312645 on Build GRCh38
rs771324452 Chr.17: 8312465-8312465 on Build GRCh38
rs368035234 Chr.17: 8312538-8312538 on Build GRCh38
rs773787456 Chr.17: 8312439-8312439 on Build GRCh38
rs779354039 Chr.17: 8312694-8312694 on Build GRCh38
rs755181959 Chr.17: 8312517-8312517 on Build GRCh38
rs934898137 Chr.17: 8312676-8312676 on Build GRCh38
rs374195875 Chr.17: 8312491-8312491 on Build GRCh38
rs367942685 Chr.17: 8312630-8312630 on Build GRCh38
rs773380012 Chr.17: 8312382-8312382 on Build GRCh38
rs756075166 Chr.17: 8312407-8312407 on Build GRCh38
rs559241646 Chr.17: 8312615-8312615 on Build GRCh38
rs571400409 Chr.17: 8312405-8312405 on Build GRCh38
rs752082895 Chr.17: 8312487-8312487 on Build GRCh38
rs768892841 Chr.17: 8312375-8312375 on Build GRCh38
rs1031646825 Chr.17: 8312529-8312529 on Build GRCh38
rs372262256 Chr.17: 8312652-8312652 on Build GRCh38
rs769465417 Chr.17: 8312755-8312755 on Build GRCh38
rs750161380 Chr.17: 8312731-8312733 on Build GRCh38
rs759243254 Chr.17: 8312476-8312476 on Build GRCh38
rs758324428 Chr.17: 8312795-8312797 on Build GRCh38
rs758287768 Chr.17: 8312610-8312610 on Build GRCh38
rs756974695 Chr.17: 8312509-8312509 on Build GRCh38
rs114491707 Chr.17: 8312640-8312640 on Build GRCh38
rs375713883 Chr.17: 8312569-8312569 on Build GRCh38
rs763386020 Chr.17: 8312383-8312383 on Build GRCh38
rs777484923 Chr.17: 8312353-8312353 on Build GRCh38
rs148813316 Chr.17: 8312481-8312481 on Build GRCh38
rs748124101 Chr.17: 8312531-8312531 on Build GRCh38
rs545136585 Chr.17: 8312693-8312693 on Build GRCh38
rs200188980 Chr.17: 8312454-8312454 on Build GRCh38
rs746728875 Chr.17: 8312651-8312651 on Build GRCh38
rs746640088 Chr.17: 8312724-8312724 on Build GRCh38
rs776506172 Chr.17: 8312731-8312732 on Build GRCh38
rs372533490 Chr.17: 8312358-8312358 on Build GRCh38
rs777771857 Chr.17: 8312408-8312408 on Build GRCh38
rs770188779 Chr.17: 8312380-8312380 on Build GRCh38
rs757692544 Chr.17: 8312635-8312635 on Build GRCh38
rs536796205 Chr.17: 8312518-8312518 on Build GRCh38
rs112241851 Chr.17: 8312703-8312703 on Build GRCh38
rs748261919 Chr.17: 8312435-8312435 on Build GRCh38
rs139532485 Chr.17: 8312361-8312361 on Build GRCh38
rs568352928 Chr.17: 8312772-8312772 on Build GRCh38
rs149692144 Chr.17: 8312426-8312426 on Build GRCh38
rs116075794 Chr.17: 8312515-8312515 on Build GRCh38
rs761901404 Chr.17: 8312666-8312667 on Build GRCh38
rs781575113 Chr.17: 8312527-8312527 on Build GRCh38
rs780387549 Chr.17: 8312419-8312419 on Build GRCh38
rs200846099 Chr.17: 8312606-8312606 on Build GRCh38
rs77503849 Chr.17: 8312754-8312754 on Build GRCh38
rs780440722 Chr.17: 8312360-8312360 on Build GRCh38
rs368543027 Chr.17: 8312427-8312427 on Build GRCh38
rs761174975 Chr.17: 8312459-8312459 on Build GRCh38
rs541684718 Chr.17: 8312702-8312702 on Build GRCh38
rs764103901 Chr.17: 8312527-8312529 on Build GRCh38
rs144080590 Chr.17: 8312355-8312355 on Build GRCh38
rs764224578 Chr.17: 8312671-8312671 on Build GRCh38
rs926440781 Chr.17: 8312394-8312394 on Build GRCh38
rs776275342 Chr.17: 8312655-8312655 on Build GRCh38
rs1005649572 Chr.17: 8312486-8312486 on Build GRCh38
rs760247454 Chr.17: 8312490-8312490 on Build GRCh38
rs200099841 Chr.17: 8312614-8312614 on Build GRCh38
rs115065009 Chr.17: 8312437-8312437 on Build GRCh38
rs763042060 Chr.17: 8312773-8312773 on Build GRCh38
rs780727510 Chr.17: 8312753-8312753 on Build GRCh38
rs926331333 Chr.17: 8312570-8312570 on Build GRCh38
rs536515839 Chr.17: 8312544-8312544 on Build GRCh38
rs185403108 Chr.17: 8312396-8312396 on Build GRCh38
rs773969766 Chr.17: 8312475-8312475 on Build GRCh38
rs756124923 Chr.17: 8312618-8312618 on Build GRCh38
rs750968437 Chr.17: 8312700-8312700 on Build GRCh38
rs765962698 Chr.17: 8312390-8312390 on Build GRCh38
rs766244736 Chr.17: 8312666-8312666 on Build GRCh38
rs776414313 Chr.17: 8312576-8312576 on Build GRCh38
rs756244846 Chr.17: 8312532-8312532 on Build GRCh38
rs765214731 Chr.17: 8312660-8312660 on Build GRCh38
rs142622501 Chr.17: 8312602-8312602 on Build GRCh38

View Full Product Details