Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 8735248-8735767 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC42
Entrez Gene ID
146849
Gene Name
coiled-coil domain containing 42
Gene Aliases
CCDC42A
UniGene
Hs.121438
Gene Chromosome Location
Chr.17: 8729928-8744836 on Build GRCh38
Transcripts
NM_001158261.1 NM_144681.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1387677 c.611C>T p.A204V Chr.17: 8735493-8735493 on Build GRCh38
6622218 c.545G>A p.R182H Chr.17: 8735559-8735559 on Build GRCh38
5916572 c.665C>A p.A222E Chr.17: 8735439-8735439 on Build GRCh38
2882597 c.664G>C p.A222P Chr.17: 8735440-8735440 on Build GRCh38
1247519 c.527A>T p.K176M Chr.17: 8735577-8735577 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1000046352 Chr.17: 8735315-8735316 on Build GRCh38
rs774174598 Chr.17: 8735294-8735295 on Build GRCh38
rs781125276 Chr.17: 8735482-8735482 on Build GRCh38
rs374987425 Chr.17: 8735369-8735369 on Build GRCh38
rs141777168 Chr.17: 8735513-8735513 on Build GRCh38
rs895510386 Chr.17: 8735291-8735295 on Build GRCh38
rs777362328 Chr.17: 8735343-8735343 on Build GRCh38
rs1008726041 Chr.17: 8735493-8735493 on Build GRCh38
rs781085439 Chr.17: 8735517-8735517 on Build GRCh38
rs778525153 Chr.17: 8735416-8735416 on Build GRCh38
rs753089209 Chr.17: 8735394-8735394 on Build GRCh38
rs777608376 Chr.17: 8735620-8735620 on Build GRCh38
rs371274592 Chr.17: 8735484-8735484 on Build GRCh38
rs369861323 Chr.17: 8735295-8735295 on Build GRCh38
rs1031478925 Chr.17: 8735341-8735341 on Build GRCh38
rs370642730 Chr.17: 8735350-8735350 on Build GRCh38
rs145668973 Chr.17: 8735500-8735500 on Build GRCh38
rs149912256 Chr.17: 8735491-8735491 on Build GRCh38
rs904451571 Chr.17: 8735309-8735309 on Build GRCh38
rs772570264 Chr.17: 8735496-8735496 on Build GRCh38
rs371328672 Chr.17: 8735559-8735559 on Build GRCh38
rs560557076 Chr.17: 8735342-8735342 on Build GRCh38
rs879482716 Chr.17: 8735733-8735733 on Build GRCh38
rs781487670 Chr.17: 8735672-8735672 on Build GRCh38
rs868631145 Chr.17: 8735276-8735276 on Build GRCh38
rs773366051 Chr.17: 8735316-8735317 on Build GRCh38
rs776146375 Chr.17: 8735503-8735503 on Build GRCh38
rs752050583 Chr.17: 8735301-8735301 on Build GRCh38
rs768516897 Chr.17: 8735480-8735480 on Build GRCh38
rs557440923 Chr.17: 8735424-8735424 on Build GRCh38
rs375583938 Chr.17: 8735560-8735560 on Build GRCh38
rs761492172 Chr.17: 8735571-8735571 on Build GRCh38
rs375806043 Chr.17: 8735274-8735274 on Build GRCh38
rs138218720 Chr.17: 8735507-8735507 on Build GRCh38
rs918815968 Chr.17: 8735469-8735469 on Build GRCh38
rs749482733 Chr.17: 8735295-8735296 on Build GRCh38
rs141192837 Chr.17: 8735509-8735509 on Build GRCh38
rs372539503 Chr.17: 8735377-8735377 on Build GRCh38
rs1025905124 Chr.17: 8735674-8735674 on Build GRCh38
rs767314074 Chr.17: 8735608-8735608 on Build GRCh38
rs368917591 Chr.17: 8735401-8735401 on Build GRCh38
rs937342803 Chr.17: 8735329-8735329 on Build GRCh38
rs368546763 Chr.17: 8735617-8735617 on Build GRCh38
rs745712450 Chr.17: 8735299-8735300 on Build GRCh38
rs200479173 Chr.17: 8735355-8735355 on Build GRCh38
rs982052641 Chr.17: 8735700-8735700 on Build GRCh38
rs762044749 Chr.17: 8735571-8735571 on Build GRCh38
rs755430900 Chr.17: 8735303-8735303 on Build GRCh38
rs542691621 Chr.17: 8735682-8735682 on Build GRCh38
rs199724755 Chr.17: 8735374-8735374 on Build GRCh38
rs763748335 Chr.17: 8735405-8735405 on Build GRCh38
rs375820682 Chr.17: 8735379-8735379 on Build GRCh38
rs752677099 Chr.17: 8735512-8735512 on Build GRCh38
rs751086836 Chr.17: 8735585-8735585 on Build GRCh38
rs776937755 Chr.17: 8735639-8735639 on Build GRCh38
rs766057119 Chr.17: 8735584-8735584 on Build GRCh38
rs778272908 Chr.17: 8735363-8735363 on Build GRCh38
rs116841782 Chr.17: 8735273-8735273 on Build GRCh38
rs150575602 Chr.17: 8735532-8735532 on Build GRCh38
rs757353433 Chr.17: 8735604-8735604 on Build GRCh38
rs778933103 Chr.17: 8735539-8735539 on Build GRCh38
rs149274270 Chr.17: 8735554-8735554 on Build GRCh38
rs370804265 Chr.17: 8735499-8735499 on Build GRCh38
rs113726847 Chr.17: 8735492-8735492 on Build GRCh38
rs935667406 Chr.17: 8735697-8735704 on Build GRCh38
rs959072207 Chr.17: 8735728-8735728 on Build GRCh38
rs1053000970 Chr.17: 8735705-8735705 on Build GRCh38
rs761885044 Chr.17: 8735317-8735318 on Build GRCh38
rs566514810 Chr.17: 8735487-8735487 on Build GRCh38
rs1006624837 Chr.17: 8735574-8735574 on Build GRCh38
rs758587213 Chr.17: 8735626-8735626 on Build GRCh38
rs549982767 Chr.17: 8735378-8735378 on Build GRCh38
rs764949930 Chr.17: 8735573-8735573 on Build GRCh38
rs373057677 Chr.17: 8735344-8735344 on Build GRCh38
rs200503858 Chr.17: 8735423-8735423 on Build GRCh38
rs767905662 Chr.17: 8735340-8735340 on Build GRCh38
rs549310973 Chr.17: 8735364-8735364 on Build GRCh38
rs778024427 Chr.17: 8735278-8735279 on Build GRCh38
rs754983758 Chr.17: 8735588-8735588 on Build GRCh38
rs780101445 Chr.17: 8735435-8735435 on Build GRCh38
rs763542623 Chr.17: 8735451-8735451 on Build GRCh38
rs760134781 Chr.17: 8735381-8735381 on Build GRCh38
rs374114436 Chr.17: 8735633-8735633 on Build GRCh38
rs367796885 Chr.17: 8735537-8735537 on Build GRCh38
rs367923929 Chr.17: 8735742-8735742 on Build GRCh38
rs928714651 Chr.17: 8735278-8735278 on Build GRCh38
rs1035844091 Chr.17: 8735703-8735703 on Build GRCh38
rs138584468 Chr.17: 8735547-8735547 on Build GRCh38
rs974310068 Chr.17: 8735442-8735442 on Build GRCh38
rs375234897 Chr.17: 8735504-8735504 on Build GRCh38
rs771099985 Chr.17: 8735295-8735308 on Build GRCh38
rs770173173 Chr.17: 8735373-8735373 on Build GRCh38
rs748560553 Chr.17: 8735646-8735646 on Build GRCh38
rs769237596 Chr.17: 8735634-8735634 on Build GRCh38
rs914634831 Chr.17: 8735743-8735743 on Build GRCh38
rs571535127 Chr.17: 8735409-8735409 on Build GRCh38
rs951628477 Chr.17: 8735679-8735679 on Build GRCh38
rs777077616 Chr.17: 8735565-8735565 on Build GRCh38
rs767959126 Chr.17: 8735382-8735382 on Build GRCh38
rs764030052 Chr.17: 8735514-8735514 on Build GRCh38
rs147370742 Chr.17: 8735555-8735555 on Build GRCh38
rs772045155 Chr.17: 8735300-8735307 on Build GRCh38
rs1055765647 Chr.17: 8735330-8735330 on Build GRCh38
rs185250830 Chr.17: 8735632-8735632 on Build GRCh38
rs772685088 Chr.17: 8735582-8735582 on Build GRCh38
rs770116952 Chr.17: 8735655-8735655 on Build GRCh38
rs138157856 Chr.17: 8735511-8735511 on Build GRCh38
rs139469533 Chr.17: 8735583-8735583 on Build GRCh38
rs746808021 Chr.17: 8735462-8735462 on Build GRCh38
rs145580826 Chr.17: 8735510-8735510 on Build GRCh38
rs756855861 Chr.17: 8735415-8735415 on Build GRCh38
rs939496600 Chr.17: 8735400-8735400 on Build GRCh38
rs775342880 Chr.17: 8735369-8735392 on Build GRCh38
rs760621171 Chr.17: 8735413-8735413 on Build GRCh38
rs752538761 Chr.17: 8735609-8735609 on Build GRCh38
rs746154960 Chr.17: 8735622-8735622 on Build GRCh38
rs965108897 Chr.17: 8735368-8735368 on Build GRCh38
rs772831416 Chr.17: 8735281-8735281 on Build GRCh38

View Full Product Details