Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 9861936-9862436 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLP2R
Entrez Gene ID
9340
Gene Name
glucagon like peptide 2 receptor
Gene Aliases
-
UniGene
Hs.248202
Gene Chromosome Location
Chr.17: 9825508-9894163 on Build GRCh38
Transcripts
NM_004246.2 XM_005256861.3 XM_011524077.2 XM_017025339.1 XM_017025340.1 XM_017025341.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3524156 c.1026C>T p.I342I Chr.17: 9862060-9862060 on Build GRCh38
6043044 c.1021T>G p.W341G Chr.17: 9862055-9862055 on Build GRCh38
6671692 c.1033G>T p.G345* Chr.17: 9862067-9862067 on Build GRCh38
5500661 c.1056+10A>T p.? Chr.17: 9862100-9862100 on Build GRCh38
244712 c.1030C>T p.R344* Chr.17: 9862064-9862064 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1009544603 Chr.17: 9862166-9862166 on Build GRCh38
rs371544939 Chr.17: 9862150-9862150 on Build GRCh38
rs778163715 Chr.17: 9862028-9862028 on Build GRCh38
rs375063769 Chr.17: 9862127-9862127 on Build GRCh38
rs1041828740 Chr.17: 9862202-9862202 on Build GRCh38
rs755161590 Chr.17: 9862338-9862338 on Build GRCh38
rs749454238 Chr.17: 9862030-9862030 on Build GRCh38
rs766265172 Chr.17: 9861993-9861993 on Build GRCh38
rs1030157819 Chr.17: 9862310-9862310 on Build GRCh38
rs565298769 Chr.17: 9862291-9862291 on Build GRCh38
rs775412453 Chr.17: 9862115-9862115 on Build GRCh38
rs986231021 Chr.17: 9862061-9862061 on Build GRCh38
rs767362323 Chr.17: 9862128-9862128 on Build GRCh38
rs563544551 Chr.17: 9862002-9862002 on Build GRCh38
rs930147236 Chr.17: 9862140-9862140 on Build GRCh38
rs750248536 Chr.17: 9862151-9862151 on Build GRCh38
rs1023516310 Chr.17: 9862365-9862365 on Build GRCh38
rs572336806 Chr.17: 9862195-9862195 on Build GRCh38
rs907571871 Chr.17: 9861999-9861999 on Build GRCh38
rs983523547 Chr.17: 9861974-9861974 on Build GRCh38
rs951772308 Chr.17: 9862267-9862267 on Build GRCh38
rs1047642856 Chr.17: 9862144-9862144 on Build GRCh38
rs747353309 Chr.17: 9862071-9862071 on Build GRCh38
rs2293101 Chr.17: 9862281-9862281 on Build GRCh38
rs79952508 Chr.17: 9862122-9862122 on Build GRCh38
rs376142802 Chr.17: 9862015-9862015 on Build GRCh38
rs979232841 Chr.17: 9862394-9862394 on Build GRCh38
rs564780030 Chr.17: 9862217-9862217 on Build GRCh38
rs992113993 Chr.17: 9862048-9862048 on Build GRCh38
rs969155083 Chr.17: 9862370-9862370 on Build GRCh38
rs996351813 Chr.17: 9862207-9862207 on Build GRCh38
rs779122087 Chr.17: 9862036-9862036 on Build GRCh38
rs928892987 Chr.17: 9862149-9862149 on Build GRCh38
rs752485448 Chr.17: 9862026-9862027 on Build GRCh38
rs776273872 Chr.17: 9862074-9862074 on Build GRCh38
rs751282452 Chr.17: 9862010-9862010 on Build GRCh38
rs761399385 Chr.17: 9862090-9862090 on Build GRCh38
rs926043015 Chr.17: 9862145-9862145 on Build GRCh38
rs891822588 Chr.17: 9862160-9862160 on Build GRCh38
rs530124872 Chr.17: 9862121-9862121 on Build GRCh38
rs764787387 Chr.17: 9862100-9862100 on Build GRCh38
rs757547728 Chr.17: 9862139-9862139 on Build GRCh38
rs768965075 Chr.17: 9862073-9862073 on Build GRCh38
rs181933856 Chr.17: 9862333-9862333 on Build GRCh38
rs754169112 Chr.17: 9862134-9862134 on Build GRCh38
rs201983334 Chr.17: 9862080-9862080 on Build GRCh38
rs908860421 Chr.17: 9862063-9862063 on Build GRCh38
rs367669929 Chr.17: 9862146-9862146 on Build GRCh38
rs1027366297 Chr.17: 9862236-9862236 on Build GRCh38
rs373329217 Chr.17: 9862052-9862052 on Build GRCh38
rs772425874 Chr.17: 9862056-9862056 on Build GRCh38
rs200291078 Chr.17: 9862113-9862113 on Build GRCh38
rs12150273 Chr.17: 9862197-9862197 on Build GRCh38
rs532076935 Chr.17: 9862258-9862258 on Build GRCh38
rs548575418 Chr.17: 9862375-9862375 on Build GRCh38
rs992458343 Chr.17: 9862324-9862324 on Build GRCh38
rs775969089 Chr.17: 9862064-9862064 on Build GRCh38
rs760679080 Chr.17: 9862142-9862142 on Build GRCh38
rs757424868 Chr.17: 9862035-9862035 on Build GRCh38

View Full Product Details