Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 32489083-32489593 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK5R1
Entrez Gene ID
8851
Gene Name
cyclin dependent kinase 5 regulatory subunit 1
Gene Aliases
CDK5P35, CDK5R, NCK5A, p23, p25, p35, p35nck5a
UniGene
Hs.500015
Gene Chromosome Location
Chr.17: 32485281-32491256 on Build GRCh38
Transcripts
NM_003885.2 XM_011525438.2 XM_017025281.1

SNP Detail

SNP ID Chromosome Location
rs568323776 Chr.17: 32489453-32489453 on Build GRCh38
rs778154210 Chr.17: 32489230-32489230 on Build GRCh38
rs142764418 Chr.17: 32489510-32489510 on Build GRCh38
rs186551397 Chr.17: 32489471-32489471 on Build GRCh38
rs890280444 Chr.17: 32489422-32489422 on Build GRCh38
rs893576867 Chr.17: 32489451-32489451 on Build GRCh38
rs554464402 Chr.17: 32489396-32489396 on Build GRCh38
rs974976081 Chr.17: 32489273-32489273 on Build GRCh38
rs529784212 Chr.17: 32489234-32489234 on Build GRCh38
rs571782226 Chr.17: 32489227-32489227 on Build GRCh38
rs761735963 Chr.17: 32489555-32489555 on Build GRCh38
rs560864203 Chr.17: 32489231-32489231 on Build GRCh38
rs1030744543 Chr.17: 32489232-32489232 on Build GRCh38
rs577041865 Chr.17: 32489482-32489482 on Build GRCh38
rs923455646 Chr.17: 32489276-32489276 on Build GRCh38
rs982704081 Chr.17: 32489115-32489115 on Build GRCh38
rs1037587323 Chr.17: 32489164-32489164 on Build GRCh38
rs774148041 Chr.17: 32489283-32489283 on Build GRCh38
rs1030670924 Chr.17: 32489447-32489447 on Build GRCh38
rs951492951 Chr.17: 32489534-32489534 on Build GRCh38
rs552514471 Chr.17: 32489285-32489285 on Build GRCh38
rs1044180217 Chr.17: 32489181-32489181 on Build GRCh38
rs989418313 Chr.17: 32489346-32489346 on Build GRCh38
rs1015964112 Chr.17: 32489512-32489512 on Build GRCh38
rs893017349 Chr.17: 32489151-32489151 on Build GRCh38
rs995996060 Chr.17: 32489203-32489203 on Build GRCh38
rs1051724446 Chr.17: 32489421-32489421 on Build GRCh38
rs768592857 Chr.17: 32489158-32489159 on Build GRCh38
rs907064403 Chr.17: 32489439-32489439 on Build GRCh38
rs982767177 Chr.17: 32489517-32489517 on Build GRCh38
rs936672480 Chr.17: 32489342-32489342 on Build GRCh38
rs780893977 Chr.17: 32489217-32489219 on Build GRCh38
rs931873993 Chr.17: 32489420-32489420 on Build GRCh38
rs899861030 Chr.17: 32489401-32489401 on Build GRCh38
rs1047200997 Chr.17: 32489134-32489134 on Build GRCh38
rs544171092 Chr.17: 32489179-32489179 on Build GRCh38
rs569151995 Chr.17: 32489340-32489340 on Build GRCh38
rs9333305 Chr.17: 32489519-32489519 on Build GRCh38
rs561461921 Chr.17: 32489180-32489180 on Build GRCh38
rs988884907 Chr.17: 32489408-32489408 on Build GRCh38
rs1005850704 Chr.17: 32489247-32489247 on Build GRCh38
rs762383006 Chr.17: 32489322-32489324 on Build GRCh38
rs184096796 Chr.17: 32489257-32489257 on Build GRCh38
rs982856145 Chr.17: 32489361-32489361 on Build GRCh38
rs1010497669 Chr.17: 32489455-32489455 on Build GRCh38
rs1012722557 Chr.17: 32489153-32489153 on Build GRCh38
rs779464492 Chr.17: 32489174-32489174 on Build GRCh38
rs553824605 Chr.17: 32489473-32489473 on Build GRCh38
rs9333304 Chr.17: 32489392-32489392 on Build GRCh38
rs746754197 Chr.17: 32489134-32489134 on Build GRCh38
rs539631470 Chr.17: 32489487-32489487 on Build GRCh38
rs1047937665 Chr.17: 32489444-32489444 on Build GRCh38
rs560945275 Chr.17: 32489523-32489523 on Build GRCh38
rs938494036 Chr.17: 32489440-32489440 on Build GRCh38
rs776818247 Chr.17: 32489251-32489251 on Build GRCh38
rs928040949 Chr.17: 32489133-32489134 on Build GRCh38
rs1016542602 Chr.17: 32489248-32489248 on Build GRCh38
rs924016632 Chr.17: 32489317-32489317 on Build GRCh38
rs914504029 Chr.17: 32489363-32489363 on Build GRCh38
rs556466463 Chr.17: 32489498-32489498 on Build GRCh38
rs560392464 Chr.17: 32489246-32489246 on Build GRCh38
rs538924341 Chr.17: 32489244-32489244 on Build GRCh38
rs768408428 Chr.17: 32489277-32489277 on Build GRCh38
rs941301715 Chr.17: 32489394-32489394 on Build GRCh38

View Full Product Details