Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 32489879-32490378 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK5R1
Entrez Gene ID
8851
Gene Name
cyclin dependent kinase 5 regulatory subunit 1
Gene Aliases
CDK5P35, CDK5R, NCK5A, p23, p25, p35, p35nck5a
UniGene
Hs.500015
Gene Chromosome Location
Chr.17: 32485281-32491256 on Build GRCh38
Transcripts
NM_003885.2 XM_011525438.2 XM_017025281.1

SNP Detail

SNP ID Chromosome Location
rs1037466445 Chr.17: 32490242-32490242 on Build GRCh38
rs921642830 Chr.17: 32490278-32490278 on Build GRCh38
rs931655569 Chr.17: 32490279-32490279 on Build GRCh38
rs955519659 Chr.17: 32489975-32489975 on Build GRCh38
rs941837090 Chr.17: 32490241-32490241 on Build GRCh38
rs958181299 Chr.17: 32490022-32490022 on Build GRCh38
rs149535076 Chr.17: 32490281-32490281 on Build GRCh38
rs921367718 Chr.17: 32489954-32489954 on Build GRCh38
rs890691368 Chr.17: 32489935-32489935 on Build GRCh38
rs987321406 Chr.17: 32490225-32490225 on Build GRCh38
rs766814393 Chr.17: 32490017-32490020 on Build GRCh38
rs1003597210 Chr.17: 32490125-32490125 on Build GRCh38
rs886766747 Chr.17: 32490321-32490321 on Build GRCh38
rs533908625 Chr.17: 32490169-32490169 on Build GRCh38
rs1056154220 Chr.17: 32490130-32490130 on Build GRCh38
rs1023693043 Chr.17: 32490345-32490345 on Build GRCh38
rs1023640834 Chr.17: 32490108-32490108 on Build GRCh38
rs767484304 Chr.17: 32490121-32490121 on Build GRCh38
rs769582003 Chr.17: 32490033-32490033 on Build GRCh38
rs138054348 Chr.17: 32490069-32490069 on Build GRCh38
rs1045178949 Chr.17: 32490123-32490123 on Build GRCh38
rs371187446 Chr.17: 32490072-32490072 on Build GRCh38
rs935234405 Chr.17: 32490087-32490087 on Build GRCh38
rs969363424 Chr.17: 32490112-32490112 on Build GRCh38
rs547521140 Chr.17: 32490179-32490179 on Build GRCh38
rs986838681 Chr.17: 32490055-32490055 on Build GRCh38
rs946502477 Chr.17: 32490280-32490280 on Build GRCh38
rs1028804031 Chr.17: 32490213-32490213 on Build GRCh38
rs1053113840 Chr.17: 32490298-32490298 on Build GRCh38
rs1031004695 Chr.17: 32489909-32489909 on Build GRCh38
rs988009725 Chr.17: 32490271-32490271 on Build GRCh38
rs1004466305 Chr.17: 32490187-32490187 on Build GRCh38
rs956076881 Chr.17: 32490265-32490265 on Build GRCh38
rs907747231 Chr.17: 32490234-32490234 on Build GRCh38
rs773173904 Chr.17: 32490203-32490203 on Build GRCh38
rs928554040 Chr.17: 32490188-32490188 on Build GRCh38
rs960011718 Chr.17: 32490204-32490204 on Build GRCh38
rs982111188 Chr.17: 32490116-32490116 on Build GRCh38
rs144091317 Chr.17: 32490290-32490290 on Build GRCh38
rs995810519 Chr.17: 32489903-32489903 on Build GRCh38
rs886798052 Chr.17: 32490146-32490146 on Build GRCh38
rs570588520 Chr.17: 32490217-32490217 on Build GRCh38
rs536148917 Chr.17: 32490338-32490338 on Build GRCh38
rs962838086 Chr.17: 32490195-32490195 on Build GRCh38
rs548141431 Chr.17: 32489970-32489970 on Build GRCh38
rs183036703 Chr.17: 32489939-32489939 on Build GRCh38
rs969415848 Chr.17: 32490354-32490354 on Build GRCh38
rs878927766 Chr.17: 32490034-32490034 on Build GRCh38
rs893984063 Chr.17: 32490304-32490304 on Build GRCh38
rs1031612868 Chr.17: 32490252-32490252 on Build GRCh38
rs1000115964 Chr.17: 32489966-32489966 on Build GRCh38
rs1010990113 Chr.17: 32490337-32490337 on Build GRCh38
rs556326266 Chr.17: 32490285-32490285 on Build GRCh38

View Full Product Details