Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 33013001-33013501 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASIC2
Entrez Gene ID
40
Gene Name
acid sensing ion channel subunit 2
Gene Aliases
ACCN, ACCN1, ASIC2a, BNC1, BNaC1, MDEG, hBNaC1
UniGene
Hs.368417
Gene Chromosome Location
Chr.17: 33013087-34156806 on Build GRCh38
Transcripts
NM_001094.4 NM_183377.1

SNP Detail

SNP ID Chromosome Location
rs1038446285 Chr.17: 33013038-33013038 on Build GRCh38
rs762956265 Chr.17: 33013260-33013260 on Build GRCh38
rs375636776 Chr.17: 33013418-33013418 on Build GRCh38
rs887619531 Chr.17: 33013254-33013254 on Build GRCh38
rs961675544 Chr.17: 33013338-33013338 on Build GRCh38
rs963290256 Chr.17: 33013449-33013449 on Build GRCh38
rs898707047 Chr.17: 33013040-33013040 on Build GRCh38
rs998164557 Chr.17: 33013311-33013311 on Build GRCh38
rs912692572 Chr.17: 33013368-33013368 on Build GRCh38
rs3025255 Chr.17: 33013425-33013425 on Build GRCh38
rs974187190 Chr.17: 33013350-33013350 on Build GRCh38
rs562387447 Chr.17: 33013287-33013287 on Build GRCh38
rs1035464952 Chr.17: 33013253-33013253 on Build GRCh38
rs1043604 Chr.17: 33013273-33013273 on Build GRCh38
rs28935 Chr.17: 33013426-33013426 on Build GRCh38
rs141990016 Chr.17: 33013043-33013046 on Build GRCh38
rs981170894 Chr.17: 33013186-33013186 on Build GRCh38
rs527941611 Chr.17: 33013349-33013349 on Build GRCh38
rs1035518726 Chr.17: 33013404-33013404 on Build GRCh38
rs943245561 Chr.17: 33013267-33013267 on Build GRCh38
rs115971485 Chr.17: 33013053-33013053 on Build GRCh38
rs1031347637 Chr.17: 33013135-33013135 on Build GRCh38
rs575622351 Chr.17: 33013108-33013108 on Build GRCh38
rs919039915 Chr.17: 33013194-33013194 on Build GRCh38
rs969745359 Chr.17: 33013232-33013232 on Build GRCh38
rs940150651 Chr.17: 33013037-33013037 on Build GRCh38
rs7217893 Chr.17: 33013382-33013382 on Build GRCh38
rs891442895 Chr.17: 33013146-33013146 on Build GRCh38
rs919009062 Chr.17: 33013414-33013414 on Build GRCh38
rs540995659 Chr.17: 33013417-33013417 on Build GRCh38
rs1001211350 Chr.17: 33013401-33013401 on Build GRCh38
rs554110511 Chr.17: 33013399-33013399 on Build GRCh38
rs3025256 Chr.17: 33013380-33013380 on Build GRCh38
rs915535484 Chr.17: 33013079-33013079 on Build GRCh38
rs371500609 Chr.17: 33013451-33013451 on Build GRCh38
rs562959568 Chr.17: 33013205-33013205 on Build GRCh38
rs891378728 Chr.17: 33013071-33013071 on Build GRCh38
rs979909926 Chr.17: 33013234-33013234 on Build GRCh38
rs367841139 Chr.17: 33013103-33013103 on Build GRCh38
rs1011348658 Chr.17: 33013171-33013171 on Build GRCh38
rs71377465 Chr.17: 33013029-33013029 on Build GRCh38
rs28936 Chr.17: 33013372-33013372 on Build GRCh38
rs1053761181 Chr.17: 33013376-33013376 on Build GRCh38
rs534101991 Chr.17: 33013400-33013400 on Build GRCh38
rs760750801 Chr.17: 33013365-33013365 on Build GRCh38
rs368595029 Chr.17: 33013367-33013367 on Build GRCh38
rs1033562888 Chr.17: 33013380-33013380 on Build GRCh38
rs908566629 Chr.17: 33013321-33013321 on Build GRCh38
rs542499865 Chr.17: 33013021-33013021 on Build GRCh38
rs531772483 Chr.17: 33013225-33013225 on Build GRCh38
rs571007080 Chr.17: 33013381-33013381 on Build GRCh38
rs1039342352 Chr.17: 33013449-33013450 on Build GRCh38
rs188674975 Chr.17: 33013384-33013384 on Build GRCh38
rs62068265 Chr.17: 33013446-33013446 on Build GRCh38
rs765122996 Chr.17: 33013235-33013235 on Build GRCh38
rs149574914 Chr.17: 33013398-33013398 on Build GRCh38
rs878942615 Chr.17: 33013383-33013383 on Build GRCh38
rs774567359 Chr.17: 33013310-33013310 on Build GRCh38
rs138754112 Chr.17: 33013069-33013069 on Build GRCh38
rs920014060 Chr.17: 33013364-33013364 on Build GRCh38
rs184835491 Chr.17: 33013271-33013271 on Build GRCh38
rs769942513 Chr.17: 33013054-33013054 on Build GRCh38

View Full Product Details