Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 34287007-34287519 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCL11
Entrez Gene ID
6356
Gene Name
C-C motif chemokine ligand 11
Gene Aliases
SCYA11
UniGene
Hs.54460
Gene Chromosome Location
Chr.17: 34285668-34288180 on Build GRCh38
Transcripts
NM_002986.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3180807 c.188T>A p.I63N Chr.17: 34287207-34287207 on Build GRCh38
5017554 c.91delA p.T31fs*16 Chr.17: 34287110-34287110 on Build GRCh38
3180808 c.188+1G>T p.? Chr.17: 34287208-34287208 on Build GRCh38
5055374 c.127C>A p.L43I Chr.17: 34287146-34287146 on Build GRCh38
319301 c.173C>G p.P58R Chr.17: 34287192-34287192 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201112720 Chr.17: 34287129-34287129 on Build GRCh38
rs77512792 Chr.17: 34287369-34287369 on Build GRCh38
rs749416164 Chr.17: 34287204-34287204 on Build GRCh38
rs764321223 Chr.17: 34287117-34287117 on Build GRCh38
rs373131160 Chr.17: 34287245-34287246 on Build GRCh38
rs765496025 Chr.17: 34287309-34287309 on Build GRCh38
rs948894180 Chr.17: 34287048-34287048 on Build GRCh38
rs993461865 Chr.17: 34287296-34287297 on Build GRCh38
rs1011647583 Chr.17: 34287339-34287339 on Build GRCh38
rs1007817347 Chr.17: 34287365-34287365 on Build GRCh38
rs200431917 Chr.17: 34287184-34287184 on Build GRCh38
rs3815341 Chr.17: 34287406-34287406 on Build GRCh38
rs201485727 Chr.17: 34287233-34287233 on Build GRCh38
rs779946609 Chr.17: 34287201-34287201 on Build GRCh38
rs771528461 Chr.17: 34287094-34287094 on Build GRCh38
rs200955262 Chr.17: 34287050-34287050 on Build GRCh38
rs749068344 Chr.17: 34287299-34287299 on Build GRCh38
rs200119205 Chr.17: 34287082-34287082 on Build GRCh38
rs747402916 Chr.17: 34287085-34287085 on Build GRCh38
rs200920749 Chr.17: 34287183-34287183 on Build GRCh38
rs866266504 Chr.17: 34287191-34287191 on Build GRCh38
rs1038117969 Chr.17: 34287419-34287419 on Build GRCh38
rs928836122 Chr.17: 34287362-34287362 on Build GRCh38
rs766246869 Chr.17: 34287132-34287132 on Build GRCh38
rs200597949 Chr.17: 34287247-34287247 on Build GRCh38
rs766334928 Chr.17: 34287059-34287059 on Build GRCh38
rs140141523 Chr.17: 34287167-34287167 on Build GRCh38
rs769892704 Chr.17: 34287194-34287194 on Build GRCh38
rs757750206 Chr.17: 34287174-34287174 on Build GRCh38
rs746191397 Chr.17: 34287058-34287059 on Build GRCh38
rs760449538 Chr.17: 34287228-34287228 on Build GRCh38
rs770101786 Chr.17: 34287239-34287239 on Build GRCh38
rs551220221 Chr.17: 34287396-34287396 on Build GRCh38
rs908185409 Chr.17: 34287386-34287386 on Build GRCh38
rs759569752 Chr.17: 34287046-34287046 on Build GRCh38
rs201940576 Chr.17: 34287113-34287113 on Build GRCh38
rs777040446 Chr.17: 34287234-34287234 on Build GRCh38
rs143782300 Chr.17: 34287205-34287205 on Build GRCh38
rs202159929 Chr.17: 34287143-34287143 on Build GRCh38
rs772466882 Chr.17: 34287075-34287075 on Build GRCh38
rs867743076 Chr.17: 34287093-34287093 on Build GRCh38
rs202220701 Chr.17: 34287221-34287221 on Build GRCh38
rs199528980 Chr.17: 34287047-34287047 on Build GRCh38
rs891045704 Chr.17: 34287265-34287265 on Build GRCh38
rs200153494 Chr.17: 34287245-34287245 on Build GRCh38
rs377267955 Chr.17: 34287068-34287068 on Build GRCh38
rs938958368 Chr.17: 34287079-34287079 on Build GRCh38
rs199758114 Chr.17: 34287057-34287057 on Build GRCh38
rs941135808 Chr.17: 34287166-34287166 on Build GRCh38
rs913551614 Chr.17: 34287059-34287060 on Build GRCh38
rs781762601 Chr.17: 34287181-34287181 on Build GRCh38
rs764694346 Chr.17: 34287138-34287138 on Build GRCh38
rs566855964 Chr.17: 34287467-34287467 on Build GRCh38
rs201374252 Chr.17: 34287073-34287073 on Build GRCh38
rs747824817 Chr.17: 34287072-34287072 on Build GRCh38
rs200381052 Chr.17: 34287230-34287230 on Build GRCh38
rs200955863 Chr.17: 34287153-34287153 on Build GRCh38
rs982829615 Chr.17: 34287375-34287375 on Build GRCh38
rs990695330 Chr.17: 34287034-34287034 on Build GRCh38
rs762098197 Chr.17: 34287238-34287239 on Build GRCh38
rs768925966 Chr.17: 34287413-34287413 on Build GRCh38
rs1050893922 Chr.17: 34287462-34287462 on Build GRCh38
rs4795898 Chr.17: 34287423-34287423 on Build GRCh38
rs957662891 Chr.17: 34287314-34287314 on Build GRCh38
rs199822520 Chr.17: 34287116-34287116 on Build GRCh38
rs754507318 Chr.17: 34287137-34287137 on Build GRCh38
rs372627424 Chr.17: 34287494-34287494 on Build GRCh38
rs536735619 Chr.17: 34287422-34287422 on Build GRCh38
rs899664664 Chr.17: 34287180-34287180 on Build GRCh38
rs200721308 Chr.17: 34287101-34287101 on Build GRCh38
rs201590633 Chr.17: 34287051-34287051 on Build GRCh38
rs774475356 Chr.17: 34287404-34287404 on Build GRCh38
rs200581050 Chr.17: 34287065-34287065 on Build GRCh38
rs367601754 Chr.17: 34287086-34287086 on Build GRCh38
rs753277441 Chr.17: 34287064-34287064 on Build GRCh38
rs775748052 Chr.17: 34287112-34287112 on Build GRCh38
rs768841988 Chr.17: 34287202-34287202 on Build GRCh38
rs758759953 Chr.17: 34287075-34287075 on Build GRCh38
rs966358071 Chr.17: 34287432-34287432 on Build GRCh38
rs747124405 Chr.17: 34287185-34287185 on Build GRCh38
rs1027574724 Chr.17: 34287326-34287326 on Build GRCh38
rs753778448 Chr.17: 34287136-34287136 on Build GRCh38
rs894566628 Chr.17: 34287157-34287157 on Build GRCh38
rs777556986 Chr.17: 34287279-34287279 on Build GRCh38
rs200085641 Chr.17: 34287152-34287152 on Build GRCh38
rs1023087954 Chr.17: 34287342-34287342 on Build GRCh38
rs538656320 Chr.17: 34287495-34287495 on Build GRCh38
rs527555142 Chr.17: 34287481-34287481 on Build GRCh38
rs981610100 Chr.17: 34287358-34287358 on Build GRCh38
rs769827637 Chr.17: 34287104-34287104 on Build GRCh38
rs762040254 Chr.17: 34287461-34287461 on Build GRCh38
rs1017424617 Chr.17: 34287391-34287391 on Build GRCh38
rs774569655 Chr.17: 34287217-34287217 on Build GRCh38
rs80326212 Chr.17: 34287351-34287351 on Build GRCh38
rs190489401 Chr.17: 34287271-34287271 on Build GRCh38
rs375131716 Chr.17: 34287103-34287103 on Build GRCh38
rs552096945 Chr.17: 34287350-34287350 on Build GRCh38
rs780543744 Chr.17: 34287074-34287075 on Build GRCh38
rs772949504 Chr.17: 34287125-34287125 on Build GRCh38
rs559276662 Chr.17: 34287249-34287249 on Build GRCh38

View Full Product Details