Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 35587185-35587693 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP2B1
Entrez Gene ID
163
Gene Name
adaptor related protein complex 2 beta 1 subunit
Gene Aliases
ADTB2, AP105B, AP2-BETA, CLAPB1
UniGene
Hs.514819
Gene Chromosome Location
Chr.17: 35586899-35726417 on Build GRCh38
Transcripts
NM_001030006.1 NM_001282.2 XM_005257937.3 XM_005257938.2 XM_005257941.2 XM_011524448.1 XM_011524449.2 XM_011524450.1 XM_011524451.1 XM_011524452.1 XM_011524453.1 XM_011524454.1 XM_011524455.2 XM_017024284.1 XM_017024285.1 XM_017024286.1 XM_017024287.1

SNP Detail

SNP ID Chromosome Location
rs907984994 Chr.17: 35587544-35587544 on Build GRCh38
rs940739337 Chr.17: 35587294-35587294 on Build GRCh38
rs745352858 Chr.17: 35587217-35587217 on Build GRCh38
rs1035312119 Chr.17: 35587496-35587496 on Build GRCh38
rs968934945 Chr.17: 35587470-35587470 on Build GRCh38
rs957381160 Chr.17: 35587233-35587233 on Build GRCh38
rs1022681301 Chr.17: 35587428-35587428 on Build GRCh38
rs1051010378 Chr.17: 35587349-35587349 on Build GRCh38
rs764513091 Chr.17: 35587241-35587241 on Build GRCh38
rs914861798 Chr.17: 35587216-35587216 on Build GRCh38
rs761017423 Chr.17: 35587239-35587239 on Build GRCh38
rs886420376 Chr.17: 35587279-35587279 on Build GRCh38
rs981113458 Chr.17: 35587477-35587477 on Build GRCh38
rs920611500 Chr.17: 35587595-35587595 on Build GRCh38
rs1057244609 Chr.17: 35587261-35587261 on Build GRCh38
rs983619383 Chr.17: 35587319-35587319 on Build GRCh38
rs989901051 Chr.17: 35587240-35587240 on Build GRCh38
rs929559912 Chr.17: 35587290-35587290 on Build GRCh38
rs898381693 Chr.17: 35587304-35587304 on Build GRCh38
rs981668929 Chr.17: 35587269-35587269 on Build GRCh38
rs1031499581 Chr.17: 35587229-35587229 on Build GRCh38
rs993317500 Chr.17: 35587503-35587503 on Build GRCh38
rs531558525 Chr.17: 35587221-35587221 on Build GRCh38
rs938885656 Chr.17: 35587478-35587478 on Build GRCh38
rs1021303067 Chr.17: 35587459-35587459 on Build GRCh38
rs891086350 Chr.17: 35587363-35587363 on Build GRCh38
rs1031949206 Chr.17: 35587383-35587383 on Build GRCh38
rs927462899 Chr.17: 35587247-35587247 on Build GRCh38
rs115695941 Chr.17: 35587472-35587472 on Build GRCh38
rs928853453 Chr.17: 35587285-35587285 on Build GRCh38
rs955199107 Chr.17: 35587218-35587218 on Build GRCh38
rs113321432 Chr.17: 35587600-35587600 on Build GRCh38
rs148383271 Chr.17: 35587219-35587219 on Build GRCh38
rs942182443 Chr.17: 35587557-35587557 on Build GRCh38
rs556653555 Chr.17: 35587384-35587384 on Build GRCh38
rs373492487 Chr.17: 35587402-35587407 on Build GRCh38
rs538792888 Chr.17: 35587441-35587441 on Build GRCh38
rs955891171 Chr.17: 35587431-35587431 on Build GRCh38
rs1037625151 Chr.17: 35587582-35587582 on Build GRCh38
rs150888368 Chr.17: 35587250-35587250 on Build GRCh38
rs757850617 Chr.17: 35587631-35587631 on Build GRCh38
rs892950131 Chr.17: 35587413-35587413 on Build GRCh38
rs77790967 Chr.17: 35587338-35587338 on Build GRCh38
rs749909491 Chr.17: 35587592-35587592 on Build GRCh38
rs909389917 Chr.17: 35587543-35587543 on Build GRCh38
rs900515244 Chr.17: 35587343-35587343 on Build GRCh38
rs950997739 Chr.17: 35587504-35587504 on Build GRCh38
rs547396858 Chr.17: 35587462-35587462 on Build GRCh38
rs931219513 Chr.17: 35587606-35587606 on Build GRCh38
rs933273873 Chr.17: 35587344-35587344 on Build GRCh38
rs763891742 Chr.17: 35587527-35587527 on Build GRCh38
rs1036900480 Chr.17: 35587301-35587301 on Build GRCh38
rs889658155 Chr.17: 35587391-35587391 on Build GRCh38
rs761504050 Chr.17: 35587446-35587446 on Build GRCh38
rs998762618 Chr.17: 35587377-35587377 on Build GRCh38
rs891504855 Chr.17: 35587661-35587661 on Build GRCh38
rs761796467 Chr.17: 35587274-35587275 on Build GRCh38
rs566849438 Chr.17: 35587418-35587418 on Build GRCh38
rs1039094417 Chr.17: 35587339-35587339 on Build GRCh38
rs766281521 Chr.17: 35587634-35587634 on Build GRCh38
rs1008376420 Chr.17: 35587407-35587407 on Build GRCh38
rs1049657413 Chr.17: 35587618-35587618 on Build GRCh38
rs141518898 Chr.17: 35587244-35587244 on Build GRCh38
rs760391368 Chr.17: 35587420-35587420 on Build GRCh38
rs547064742 Chr.17: 35587401-35587401 on Build GRCh38
rs764820663 Chr.17: 35587522-35587522 on Build GRCh38
rs909369619 Chr.17: 35587320-35587320 on Build GRCh38
rs575624993 Chr.17: 35587484-35587484 on Build GRCh38
rs191418126 Chr.17: 35587644-35587644 on Build GRCh38

View Full Product Details