Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 35624303-35624843 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP2B1
Entrez Gene ID
163
Gene Name
adaptor related protein complex 2 beta 1 subunit
Gene Aliases
ADTB2, AP105B, AP2-BETA, CLAPB1
UniGene
Hs.514819
Gene Chromosome Location
Chr.17: 35586899-35726417 on Build GRCh38
Transcripts
NM_001030006.1 NM_001282.2 XM_005257937.3 XM_005257938.2 XM_005257941.2 XM_011524448.1 XM_011524449.2 XM_011524450.1 XM_011524451.1 XM_011524452.1 XM_011524453.1 XM_011524454.1 XM_011524455.2 XM_017024284.1 XM_017024285.1 XM_017024286.1 XM_017024287.1

SNP Detail

SNP ID Chromosome Location
rs746870411 Chr.17: 35624599-35624599 on Build GRCh38
rs776560708 Chr.17: 35624612-35624612 on Build GRCh38
rs201990674 Chr.17: 35624372-35624372 on Build GRCh38
rs377038661 Chr.17: 35624786-35624786 on Build GRCh38
rs9908809 Chr.17: 35624800-35624800 on Build GRCh38
rs771928156 Chr.17: 35624451-35624451 on Build GRCh38
rs750391593 Chr.17: 35624362-35624362 on Build GRCh38
rs375608434 Chr.17: 35624587-35624587 on Build GRCh38
rs763770429 Chr.17: 35624528-35624528 on Build GRCh38
rs960821942 Chr.17: 35624774-35624774 on Build GRCh38
rs980904273 Chr.17: 35624748-35624748 on Build GRCh38
rs376148009 Chr.17: 35624552-35624552 on Build GRCh38
rs939342935 Chr.17: 35624573-35624573 on Build GRCh38
rs1056140513 Chr.17: 35624732-35624732 on Build GRCh38
rs369289788 Chr.17: 35624353-35624353 on Build GRCh38
rs568576436 Chr.17: 35624795-35624795 on Build GRCh38
rs369824252 Chr.17: 35624381-35624381 on Build GRCh38
rs752702096 Chr.17: 35624452-35624452 on Build GRCh38
rs570816164 Chr.17: 35624677-35624677 on Build GRCh38
rs897536756 Chr.17: 35624771-35624771 on Build GRCh38
rs766407729 Chr.17: 35624380-35624381 on Build GRCh38
rs886326154 Chr.17: 35624340-35624340 on Build GRCh38
rs554058730 Chr.17: 35624337-35624337 on Build GRCh38
rs180860333 Chr.17: 35624412-35624412 on Build GRCh38
rs576576568 Chr.17: 35624739-35624739 on Build GRCh38
rs892110789 Chr.17: 35624700-35624700 on Build GRCh38
rs994595083 Chr.17: 35624782-35624782 on Build GRCh38
rs772846719 Chr.17: 35624454-35624454 on Build GRCh38
rs781781708 Chr.17: 35624543-35624543 on Build GRCh38
rs913888297 Chr.17: 35624334-35624335 on Build GRCh38
rs141661445 Chr.17: 35624468-35624468 on Build GRCh38
rs768262640 Chr.17: 35624788-35624788 on Build GRCh38
rs760436832 Chr.17: 35624455-35624455 on Build GRCh38
rs369015632 Chr.17: 35624561-35624561 on Build GRCh38
rs377410712 Chr.17: 35624591-35624591 on Build GRCh38
rs763712597 Chr.17: 35624633-35624633 on Build GRCh38
rs563134418 Chr.17: 35624387-35624387 on Build GRCh38
rs767152693 Chr.17: 35624355-35624355 on Build GRCh38
rs779952468 Chr.17: 35624575-35624575 on Build GRCh38
rs990119747 Chr.17: 35624418-35624418 on Build GRCh38
rs776542260 Chr.17: 35624471-35624471 on Build GRCh38
rs746003974 Chr.17: 35624411-35624411 on Build GRCh38
rs751918992 Chr.17: 35624529-35624529 on Build GRCh38
rs940642706 Chr.17: 35624608-35624608 on Build GRCh38
rs187416301 Chr.17: 35624692-35624692 on Build GRCh38
rs898975894 Chr.17: 35624370-35624370 on Build GRCh38
rs746234965 Chr.17: 35624684-35624684 on Build GRCh38
rs897486380 Chr.17: 35624417-35624417 on Build GRCh38
rs758802832 Chr.17: 35624384-35624384 on Build GRCh38
rs201493061 Chr.17: 35624379-35624379 on Build GRCh38
rs375404651 Chr.17: 35624616-35624616 on Build GRCh38
rs111831737 Chr.17: 35624737-35624737 on Build GRCh38
rs74437884 Chr.17: 35624731-35624731 on Build GRCh38
rs760485450 Chr.17: 35624367-35624367 on Build GRCh38
rs890122295 Chr.17: 35624385-35624385 on Build GRCh38
rs757782289 Chr.17: 35624630-35624630 on Build GRCh38
rs561847181 Chr.17: 35624493-35624493 on Build GRCh38
rs757946926 Chr.17: 35624525-35624525 on Build GRCh38
rs775788814 Chr.17: 35624415-35624415 on Build GRCh38
rs890236740 Chr.17: 35624663-35624663 on Build GRCh38
rs774902099 Chr.17: 35624629-35624629 on Build GRCh38
rs927997306 Chr.17: 35624572-35624572 on Build GRCh38
rs569608858 Chr.17: 35624712-35624712 on Build GRCh38
rs547432797 Chr.17: 35624627-35624627 on Build GRCh38
rs74670369 Chr.17: 35624653-35624653 on Build GRCh38
rs749754793 Chr.17: 35624574-35624574 on Build GRCh38
rs532913025 Chr.17: 35624679-35624679 on Build GRCh38
rs200199804 Chr.17: 35624382-35624382 on Build GRCh38
rs757684565 Chr.17: 35624535-35624535 on Build GRCh38
rs763030515 Chr.17: 35624416-35624416 on Build GRCh38
rs749559407 Chr.17: 35624578-35624578 on Build GRCh38
rs113108836 Chr.17: 35624605-35624605 on Build GRCh38
rs752414543 Chr.17: 35624519-35624519 on Build GRCh38
rs1035180337 Chr.17: 35624762-35624762 on Build GRCh38
rs775562694 Chr.17: 35624347-35624347 on Build GRCh38
rs931776167 Chr.17: 35624637-35624637 on Build GRCh38
rs973346459 Chr.17: 35624798-35624798 on Build GRCh38
rs781759240 Chr.17: 35624397-35624397 on Build GRCh38
rs774575981 Chr.17: 35624799-35624805 on Build GRCh38
rs764666800 Chr.17: 35624500-35624500 on Build GRCh38
rs926638973 Chr.17: 35624716-35624716 on Build GRCh38

View Full Product Details