Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 35746104-35746604 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAS2L2
Entrez Gene ID
246176
Gene Name
growth arrest specific 2 like 2
Gene Aliases
GAR17
UniGene
Hs.591194
Gene Chromosome Location
Chr.17: 35744511-35752878 on Build GRCh38
Transcripts
NM_139285.3

SNP Detail

SNP ID Chromosome Location
rs376441430 Chr.17: 35746541-35746541 on Build GRCh38
rs782226940 Chr.17: 35746331-35746331 on Build GRCh38
rs201533229 Chr.17: 35746419-35746419 on Build GRCh38
rs781995607 Chr.17: 35746192-35746192 on Build GRCh38
rs782668524 Chr.17: 35746425-35746425 on Build GRCh38
rs782447823 Chr.17: 35746424-35746424 on Build GRCh38
rs147724556 Chr.17: 35746272-35746272 on Build GRCh38
rs267604815 Chr.17: 35746257-35746257 on Build GRCh38
rs782162209 Chr.17: 35746296-35746296 on Build GRCh38
rs958169147 Chr.17: 35746124-35746124 on Build GRCh38
rs782767432 Chr.17: 35746304-35746304 on Build GRCh38
rs143661487 Chr.17: 35746373-35746373 on Build GRCh38
rs782275638 Chr.17: 35746276-35746276 on Build GRCh38
rs909116845 Chr.17: 35746565-35746565 on Build GRCh38
rs782648157 Chr.17: 35746252-35746252 on Build GRCh38
rs782730530 Chr.17: 35746136-35746136 on Build GRCh38
rs587757863 Chr.17: 35746269-35746269 on Build GRCh38
rs1025662935 Chr.17: 35746552-35746552 on Build GRCh38
rs781966545 Chr.17: 35746280-35746280 on Build GRCh38
rs782771713 Chr.17: 35746362-35746362 on Build GRCh38
rs782531734 Chr.17: 35746538-35746538 on Build GRCh38
rs375527898 Chr.17: 35746322-35746322 on Build GRCh38
rs782276437 Chr.17: 35746431-35746431 on Build GRCh38
rs782212939 Chr.17: 35746271-35746271 on Build GRCh38
rs781824574 Chr.17: 35746313-35746313 on Build GRCh38
rs782010097 Chr.17: 35746357-35746357 on Build GRCh38
rs782292177 Chr.17: 35746517-35746517 on Build GRCh38
rs781938274 Chr.17: 35746344-35746344 on Build GRCh38
rs587702439 Chr.17: 35746233-35746233 on Build GRCh38
rs782695784 Chr.17: 35746231-35746231 on Build GRCh38
rs73990296 Chr.17: 35746384-35746384 on Build GRCh38
rs983410861 Chr.17: 35746556-35746556 on Build GRCh38
rs781791810 Chr.17: 35746139-35746139 on Build GRCh38
rs782459490 Chr.17: 35746315-35746315 on Build GRCh38
rs139833672 Chr.17: 35746203-35746203 on Build GRCh38
rs201387695 Chr.17: 35746278-35746278 on Build GRCh38
rs781843411 Chr.17: 35746126-35746126 on Build GRCh38
rs587765443 Chr.17: 35746330-35746330 on Build GRCh38
rs587742522 Chr.17: 35746518-35746518 on Build GRCh38
rs587750122 Chr.17: 35746135-35746135 on Build GRCh38
rs782645395 Chr.17: 35746270-35746271 on Build GRCh38
rs900198063 Chr.17: 35746236-35746236 on Build GRCh38
rs144191508 Chr.17: 35746319-35746319 on Build GRCh38
rs140607972 Chr.17: 35746223-35746223 on Build GRCh38
rs374540202 Chr.17: 35746448-35746448 on Build GRCh38
rs782334491 Chr.17: 35746337-35746337 on Build GRCh38
rs150473184 Chr.17: 35746228-35746228 on Build GRCh38
rs139277329 Chr.17: 35746175-35746175 on Build GRCh38
rs1012821921 Chr.17: 35746474-35746474 on Build GRCh38
rs887583205 Chr.17: 35746219-35746219 on Build GRCh38
rs200932373 Chr.17: 35746300-35746300 on Build GRCh38
rs115955572 Chr.17: 35746200-35746200 on Build GRCh38
rs782302554 Chr.17: 35746174-35746174 on Build GRCh38
rs782494175 Chr.17: 35746261-35746261 on Build GRCh38
rs782745076 Chr.17: 35746220-35746220 on Build GRCh38
rs371509072 Chr.17: 35746125-35746125 on Build GRCh38
rs587630648 Chr.17: 35746271-35746271 on Build GRCh38
rs781792991 Chr.17: 35746383-35746383 on Build GRCh38
rs782539282 Chr.17: 35746328-35746328 on Build GRCh38
rs933036189 Chr.17: 35746418-35746418 on Build GRCh38
rs142059348 Chr.17: 35746204-35746204 on Build GRCh38
rs782445349 Chr.17: 35746352-35746352 on Build GRCh38
rs144556689 Chr.17: 35746172-35746172 on Build GRCh38
rs781804325 Chr.17: 35746420-35746420 on Build GRCh38
rs781893994 Chr.17: 35746232-35746232 on Build GRCh38
rs75608951 Chr.17: 35746207-35746207 on Build GRCh38
rs781975317 Chr.17: 35746365-35746365 on Build GRCh38
rs782438002 Chr.17: 35746452-35746452 on Build GRCh38
rs782752408 Chr.17: 35746358-35746358 on Build GRCh38
rs782383891 Chr.17: 35746432-35746432 on Build GRCh38
rs2013766 Chr.17: 35746472-35746472 on Build GRCh38
rs781872060 Chr.17: 35746395-35746395 on Build GRCh38
rs782628971 Chr.17: 35746435-35746435 on Build GRCh38
rs782487976 Chr.17: 35746397-35746397 on Build GRCh38
rs587632671 Chr.17: 35746569-35746569 on Build GRCh38
rs587734975 Chr.17: 35746568-35746568 on Build GRCh38
rs782804531 Chr.17: 35746471-35746471 on Build GRCh38
rs782401822 Chr.17: 35746190-35746190 on Build GRCh38
rs372670402 Chr.17: 35746386-35746386 on Build GRCh38
rs149365305 Chr.17: 35746372-35746372 on Build GRCh38
rs76566031 Chr.17: 35746555-35746555 on Build GRCh38
rs982117634 Chr.17: 35746325-35746325 on Build GRCh38
rs782485123 Chr.17: 35746157-35746157 on Build GRCh38
rs782134797 Chr.17: 35746457-35746457 on Build GRCh38
rs202100272 Chr.17: 35746206-35746206 on Build GRCh38
rs587668501 Chr.17: 35746493-35746493 on Build GRCh38
rs782564406 Chr.17: 35746142-35746142 on Build GRCh38
rs782237363 Chr.17: 35746442-35746442 on Build GRCh38
rs146645172 Chr.17: 35746293-35746293 on Build GRCh38
rs1003604300 Chr.17: 35746539-35746539 on Build GRCh38
rs782503326 Chr.17: 35746302-35746302 on Build GRCh38
rs970752574 Chr.17: 35746324-35746324 on Build GRCh38
rs782606707 Chr.17: 35746262-35746262 on Build GRCh38
rs782650881 Chr.17: 35746144-35746144 on Build GRCh38
rs377124082 Chr.17: 35746456-35746456 on Build GRCh38
rs560220278 Chr.17: 35746246-35746246 on Build GRCh38
rs148661453 Chr.17: 35746510-35746510 on Build GRCh38
rs782765550 Chr.17: 35746176-35746176 on Build GRCh38
rs56135703 Chr.17: 35746282-35746282 on Build GRCh38
rs139141646 Chr.17: 35746301-35746301 on Build GRCh38
rs1012356107 Chr.17: 35746320-35746320 on Build GRCh38
rs200152054 Chr.17: 35746147-35746147 on Build GRCh38

View Full Product Details