Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 35977445-35977955 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCL16
Entrez Gene ID
6360
Gene Name
C-C motif chemokine ligand 16
Gene Aliases
CKb12, HCC-4, ILINCK, LCC-1, LEC, LMC, Mtn-1, NCC-4, NCC4, SCYA16, SCYL4
UniGene
Hs.10458
Gene Chromosome Location
Chr.17: 35976493-35983620 on Build GRCh38
Transcripts
NM_004590.3 XM_005258020.4

SNP Detail

SNP ID Chromosome Location
rs751547434 Chr.17: 35977734-35977734 on Build GRCh38
rs536870292 Chr.17: 35977621-35977621 on Build GRCh38
rs745570882 Chr.17: 35977669-35977669 on Build GRCh38
rs757119773 Chr.17: 35977740-35977740 on Build GRCh38
rs906249340 Chr.17: 35977803-35977803 on Build GRCh38
rs755425321 Chr.17: 35977583-35977602 on Build GRCh38
rs551409831 Chr.17: 35977472-35977472 on Build GRCh38
rs553061107 Chr.17: 35977632-35977632 on Build GRCh38
rs79254649 Chr.17: 35977569-35977569 on Build GRCh38
rs770479253 Chr.17: 35977655-35977655 on Build GRCh38
rs199673019 Chr.17: 35977753-35977753 on Build GRCh38
rs770546149 Chr.17: 35977549-35977549 on Build GRCh38
rs868104903 Chr.17: 35977651-35977651 on Build GRCh38
rs954799699 Chr.17: 35977709-35977709 on Build GRCh38
rs952927455 Chr.17: 35977905-35977905 on Build GRCh38
rs775901091 Chr.17: 35977685-35977685 on Build GRCh38
rs776916140 Chr.17: 35977779-35977779 on Build GRCh38
rs74842203 Chr.17: 35977644-35977644 on Build GRCh38
rs886961509 Chr.17: 35977856-35977856 on Build GRCh38
rs200456787 Chr.17: 35977620-35977620 on Build GRCh38
rs878937062 Chr.17: 35977518-35977518 on Build GRCh38
rs200922005 Chr.17: 35977728-35977728 on Build GRCh38
rs780722080 Chr.17: 35977744-35977744 on Build GRCh38
rs565165679 Chr.17: 35977879-35977879 on Build GRCh38
rs200446584 Chr.17: 35977759-35977759 on Build GRCh38
rs1005861881 Chr.17: 35977895-35977895 on Build GRCh38
rs1000906777 Chr.17: 35977505-35977505 on Build GRCh38
rs796158003 Chr.17: 35977716-35977716 on Build GRCh38
rs141731897 Chr.17: 35977686-35977686 on Build GRCh38
rs935552389 Chr.17: 35977721-35977721 on Build GRCh38
rs988005468 Chr.17: 35977711-35977711 on Build GRCh38
rs769406651 Chr.17: 35977677-35977677 on Build GRCh38
rs767347374 Chr.17: 35977791-35977791 on Build GRCh38
rs748734275 Chr.17: 35977768-35977768 on Build GRCh38
rs754829277 Chr.17: 35977515-35977515 on Build GRCh38
rs558624004 Chr.17: 35977723-35977723 on Build GRCh38
rs199727921 Chr.17: 35977597-35977597 on Build GRCh38
rs199882984 Chr.17: 35977659-35977659 on Build GRCh38
rs920219119 Chr.17: 35977596-35977596 on Build GRCh38
rs768847780 Chr.17: 35977690-35977690 on Build GRCh38
rs544986098 Chr.17: 35977840-35977840 on Build GRCh38
rs758333118 Chr.17: 35977648-35977648 on Build GRCh38
rs573042033 Chr.17: 35977782-35977782 on Build GRCh38
rs192887586 Chr.17: 35977712-35977712 on Build GRCh38
rs267604820 Chr.17: 35977573-35977573 on Build GRCh38
rs1003413896 Chr.17: 35977516-35977516 on Build GRCh38
rs571599143 Chr.17: 35977483-35977483 on Build GRCh38
rs567732899 Chr.17: 35977611-35977611 on Build GRCh38
rs899160903 Chr.17: 35977919-35977919 on Build GRCh38
rs202162865 Chr.17: 35977771-35977771 on Build GRCh38
rs537140016 Chr.17: 35977653-35977653 on Build GRCh38
rs772020465 Chr.17: 35977542-35977542 on Build GRCh38
rs774930177 Chr.17: 35977695-35977695 on Build GRCh38
rs778807323 Chr.17: 35977535-35977535 on Build GRCh38
rs777751361 Chr.17: 35977650-35977650 on Build GRCh38
rs772634344 Chr.17: 35977545-35977545 on Build GRCh38
rs550960986 Chr.17: 35977900-35977900 on Build GRCh38
rs116832677 Chr.17: 35977752-35977752 on Build GRCh38
rs756352545 Chr.17: 35977625-35977625 on Build GRCh38
rs1047314374 Chr.17: 35977823-35977823 on Build GRCh38
rs58515146 Chr.17: 35977849-35977849 on Build GRCh38
rs1014055932 Chr.17: 35977808-35977808 on Build GRCh38
rs751517629 Chr.17: 35977594-35977594 on Build GRCh38
rs1033639120 Chr.17: 35977777-35977777 on Build GRCh38
rs748703665 Chr.17: 35977725-35977727 on Build GRCh38
rs759122517 Chr.17: 35977560-35977560 on Build GRCh38
rs1033755299 Chr.17: 35977544-35977544 on Build GRCh38
rs762191626 Chr.17: 35977707-35977707 on Build GRCh38
rs530959964 Chr.17: 35977881-35977881 on Build GRCh38
rs371738984 Chr.17: 35977667-35977667 on Build GRCh38
rs372164332 Chr.17: 35977558-35977558 on Build GRCh38
rs748114373 Chr.17: 35977540-35977540 on Build GRCh38
rs554869390 Chr.17: 35977735-35977735 on Build GRCh38
rs774525937 Chr.17: 35977760-35977760 on Build GRCh38
rs763998359 Chr.17: 35977783-35977783 on Build GRCh38
rs369787226 Chr.17: 35977764-35977764 on Build GRCh38
rs772673314 Chr.17: 35977770-35977770 on Build GRCh38
rs544207368 Chr.17: 35977731-35977731 on Build GRCh38
rs939120076 Chr.17: 35977809-35977809 on Build GRCh38
rs780632362 Chr.17: 35977665-35977665 on Build GRCh38
rs147072785 Chr.17: 35977726-35977726 on Build GRCh38
rs781345741 Chr.17: 35977622-35977633 on Build GRCh38
rs752595378 Chr.17: 35977727-35977727 on Build GRCh38

View Full Product Details