Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 36013652-36014144 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCL23
Entrez Gene ID
6368
Gene Name
C-C motif chemokine ligand 23
Gene Aliases
CK-BETA-8, CKb8, Ckb-8, Ckb-8-1, MIP-3, MIP3, MPIF-1, SCYA23, hmrp-2a
UniGene
Hs.169191
Gene Chromosome Location
Chr.17: 36013056-36018179 on Build GRCh38
Transcripts
NM_005064.5 NM_145898.3

SNP Detail

SNP ID Chromosome Location
rs755696186 Chr.17: 36013931-36013931 on Build GRCh38
rs1719203 Chr.17: 36013939-36013939 on Build GRCh38
rs142372148 Chr.17: 36013810-36013810 on Build GRCh38
rs753105212 Chr.17: 36013702-36013702 on Build GRCh38
rs751391277 Chr.17: 36013896-36013896 on Build GRCh38
rs759333711 Chr.17: 36013770-36013770 on Build GRCh38
rs780325362 Chr.17: 36013807-36013807 on Build GRCh38
rs767649970 Chr.17: 36013803-36013803 on Build GRCh38
rs386352289 Chr.17: 36013814-36013814 on Build GRCh38
rs902308615 Chr.17: 36013705-36013707 on Build GRCh38
rs142522197 Chr.17: 36013804-36013804 on Build GRCh38
rs377578794 Chr.17: 36013695-36013695 on Build GRCh38
rs769365155 Chr.17: 36013824-36013824 on Build GRCh38
rs922095369 Chr.17: 36014065-36014065 on Build GRCh38
rs749032415 Chr.17: 36013952-36013952 on Build GRCh38
rs1044294268 Chr.17: 36014123-36014123 on Build GRCh38
rs974976945 Chr.17: 36014058-36014058 on Build GRCh38
rs763431465 Chr.17: 36013710-36013710 on Build GRCh38
rs764314357 Chr.17: 36013785-36013785 on Build GRCh38
rs932976529 Chr.17: 36013838-36013838 on Build GRCh38
rs762365056 Chr.17: 36013852-36013852 on Build GRCh38
rs556089186 Chr.17: 36014051-36014051 on Build GRCh38
rs760774067 Chr.17: 36013868-36013868 on Build GRCh38
rs963528541 Chr.17: 36014054-36014054 on Build GRCh38
rs11657605 Chr.17: 36013794-36013794 on Build GRCh38
rs933556286 Chr.17: 36014072-36014072 on Build GRCh38
rs974333044 Chr.17: 36014052-36014052 on Build GRCh38
rs773934905 Chr.17: 36013958-36013958 on Build GRCh38
rs368207475 Chr.17: 36013891-36013891 on Build GRCh38
rs892946430 Chr.17: 36013886-36013886 on Build GRCh38
rs947188773 Chr.17: 36014119-36014119 on Build GRCh38
rs1002121881 Chr.17: 36013966-36013966 on Build GRCh38
rs374320959 Chr.17: 36013767-36013767 on Build GRCh38
rs569676764 Chr.17: 36013766-36013766 on Build GRCh38
rs534551723 Chr.17: 36013936-36013936 on Build GRCh38
rs746101314 Chr.17: 36013846-36013846 on Build GRCh38
rs769560097 Chr.17: 36013773-36013773 on Build GRCh38
rs947326166 Chr.17: 36013908-36013908 on Build GRCh38
rs577239457 Chr.17: 36014035-36014035 on Build GRCh38
rs73990625 Chr.17: 36014108-36014108 on Build GRCh38
rs186959176 Chr.17: 36013927-36013927 on Build GRCh38
rs1035666048 Chr.17: 36014003-36014003 on Build GRCh38
rs988144770 Chr.17: 36014078-36014078 on Build GRCh38
rs893622599 Chr.17: 36013932-36013932 on Build GRCh38
rs538427986 Chr.17: 36013788-36013788 on Build GRCh38
rs778736599 Chr.17: 36013745-36013745 on Build GRCh38
rs553238648 Chr.17: 36013703-36013703 on Build GRCh38
rs554219075 Chr.17: 36013998-36013998 on Build GRCh38
rs373678141 Chr.17: 36013866-36013866 on Build GRCh38
rs896605088 Chr.17: 36013988-36013988 on Build GRCh38
rs113634985 Chr.17: 36013737-36013737 on Build GRCh38
rs1028543772 Chr.17: 36014053-36014053 on Build GRCh38
rs766323126 Chr.17: 36013881-36013881 on Build GRCh38
rs201178323 Chr.17: 36013748-36013748 on Build GRCh38
rs950713335 Chr.17: 36014018-36014018 on Build GRCh38
rs764677052 Chr.17: 36013716-36013716 on Build GRCh38
rs745387175 Chr.17: 36013823-36013823 on Build GRCh38
rs780819762 Chr.17: 36013887-36013887 on Build GRCh38
rs774950997 Chr.17: 36013774-36013774 on Build GRCh38
rs370259219 Chr.17: 36013959-36013959 on Build GRCh38
rs905211120 Chr.17: 36013955-36013955 on Build GRCh38
rs765666000 Chr.17: 36013688-36013688 on Build GRCh38
rs756323655 Chr.17: 36013738-36013738 on Build GRCh38
rs533067929 Chr.17: 36013719-36013719 on Build GRCh38
rs774570865 Chr.17: 36013851-36013851 on Build GRCh38
rs768179410 Chr.17: 36013957-36013957 on Build GRCh38
rs747987827 Chr.17: 36013752-36013752 on Build GRCh38
rs199911286 Chr.17: 36013820-36013820 on Build GRCh38
rs768024464 Chr.17: 36013854-36013854 on Build GRCh38
rs1015788401 Chr.17: 36014039-36014039 on Build GRCh38
rs1035432146 Chr.17: 36013977-36013977 on Build GRCh38
rs777800056 Chr.17: 36013753-36013753 on Build GRCh38
rs140475420 Chr.17: 36013849-36013849 on Build GRCh38
rs182352077 Chr.17: 36013806-36013806 on Build GRCh38
rs751968168 Chr.17: 36013717-36013717 on Build GRCh38
rs190518785 Chr.17: 36013697-36013697 on Build GRCh38
rs986522055 Chr.17: 36014088-36014088 on Build GRCh38
rs779395886 Chr.17: 36013945-36013945 on Build GRCh38
rs768975427 Chr.17: 36013675-36013675 on Build GRCh38
rs1719204 Chr.17: 36014042-36014042 on Build GRCh38
rs757063662 Chr.17: 36013899-36013899 on Build GRCh38
rs151273610 Chr.17: 36013826-36013826 on Build GRCh38
rs763383748 Chr.17: 36013782-36013782 on Build GRCh38
rs571152375 Chr.17: 36013857-36013857 on Build GRCh38
rs758235883 Chr.17: 36013813-36013813 on Build GRCh38
rs988943814 Chr.17: 36014117-36014117 on Build GRCh38
rs572587529 Chr.17: 36014118-36014118 on Build GRCh38
rs145624767 Chr.17: 36013764-36013764 on Build GRCh38
rs139428167 Chr.17: 36014090-36014090 on Build GRCh38
rs781223375 Chr.17: 36013721-36013721 on Build GRCh38
rs754139698 Chr.17: 36013889-36013889 on Build GRCh38
rs752154492 Chr.17: 36013787-36013787 on Build GRCh38
rs778893945 Chr.17: 36013822-36013823 on Build GRCh38
rs576034537 Chr.17: 36014066-36014066 on Build GRCh38
rs745762807 Chr.17: 36013928-36013928 on Build GRCh38

View Full Product Details