Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 37625754-37626262 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX52
Entrez Gene ID
11056
Gene Name
DEAD-box helicase 52
Gene Aliases
HUSSY19, ROK1
UniGene
Hs.590937
Gene Chromosome Location
Chr.17: 37609739-37643464 on Build GRCh38
Transcripts
NM_001291476.1 NM_007010.4 XM_011524232.2 XM_011524233.2

SNP Detail

SNP ID Chromosome Location
rs61749870 Chr.17: 37625937-37625937 on Build GRCh38
rs765447309 Chr.17: 37626180-37626180 on Build GRCh38
rs748184353 Chr.17: 37626057-37626057 on Build GRCh38
rs765607708 Chr.17: 37625896-37625896 on Build GRCh38
rs765610060 Chr.17: 37626000-37626000 on Build GRCh38
rs139689560 Chr.17: 37626229-37626229 on Build GRCh38
rs576442612 Chr.17: 37625989-37625989 on Build GRCh38
rs767522787 Chr.17: 37626214-37626214 on Build GRCh38
rs568352709 Chr.17: 37625794-37625794 on Build GRCh38
rs771983986 Chr.17: 37626065-37626065 on Build GRCh38
rs376605490 Chr.17: 37625939-37625939 on Build GRCh38
rs890839032 Chr.17: 37625874-37625874 on Build GRCh38
rs760466599 Chr.17: 37625971-37625971 on Build GRCh38
rs1012252953 Chr.17: 37625808-37625808 on Build GRCh38
rs71135752 Chr.17: 37626229-37626229 on Build GRCh38
rs764419421 Chr.17: 37626119-37626119 on Build GRCh38
rs755489849 Chr.17: 37625888-37625888 on Build GRCh38
rs765968039 Chr.17: 37625974-37625974 on Build GRCh38
rs767030051 Chr.17: 37625847-37625847 on Build GRCh38
rs79312838 Chr.17: 37626085-37626085 on Build GRCh38
rs772998805 Chr.17: 37625970-37625970 on Build GRCh38
rs893898731 Chr.17: 37626112-37626112 on Build GRCh38
rs375109456 Chr.17: 37626071-37626071 on Build GRCh38
rs767385481 Chr.17: 37625852-37625852 on Build GRCh38
rs758619601 Chr.17: 37626014-37626014 on Build GRCh38
rs571950577 Chr.17: 37626157-37626157 on Build GRCh38
rs534347171 Chr.17: 37625797-37625797 on Build GRCh38
rs770670029 Chr.17: 37626091-37626091 on Build GRCh38
rs771905966 Chr.17: 37625968-37625968 on Build GRCh38
rs555389399 Chr.17: 37626049-37626049 on Build GRCh38
rs201448621 Chr.17: 37625851-37625852 on Build GRCh38
rs774370402 Chr.17: 37625947-37625947 on Build GRCh38
rs147670905 Chr.17: 37626184-37626184 on Build GRCh38
rs887061938 Chr.17: 37625806-37625806 on Build GRCh38
rs546014925 Chr.17: 37626225-37626225 on Build GRCh38
rs776324127 Chr.17: 37625988-37625988 on Build GRCh38
rs773013105 Chr.17: 37625852-37625852 on Build GRCh38
rs750554657 Chr.17: 37626183-37626183 on Build GRCh38
rs754387825 Chr.17: 37625887-37625887 on Build GRCh38
rs768440587 Chr.17: 37625940-37625940 on Build GRCh38
rs751958676 Chr.17: 37626124-37626124 on Build GRCh38
rs113192748 Chr.17: 37626041-37626041 on Build GRCh38
rs546721148 Chr.17: 37626037-37626037 on Build GRCh38
rs922233573 Chr.17: 37625925-37625925 on Build GRCh38
rs199835531 Chr.17: 37625975-37625975 on Build GRCh38
rs777910042 Chr.17: 37625917-37625917 on Build GRCh38
rs758810819 Chr.17: 37625903-37625903 on Build GRCh38
rs763572097 Chr.17: 37626110-37626110 on Build GRCh38
rs908518555 Chr.17: 37626034-37626034 on Build GRCh38
rs770126415 Chr.17: 37625818-37625818 on Build GRCh38
rs890910678 Chr.17: 37626121-37626121 on Build GRCh38
rs143352139 Chr.17: 37626054-37626054 on Build GRCh38
rs369898002 Chr.17: 37626097-37626097 on Build GRCh38
rs775852924 Chr.17: 37626103-37626103 on Build GRCh38
rs557966203 Chr.17: 37626205-37626205 on Build GRCh38
rs200696375 Chr.17: 37626025-37626025 on Build GRCh38
rs764509073 Chr.17: 37626019-37626019 on Build GRCh38
rs1012258966 Chr.17: 37626166-37626166 on Build GRCh38
rs140006660 Chr.17: 37625859-37625859 on Build GRCh38
rs924182313 Chr.17: 37626042-37626042 on Build GRCh38
rs1003390468 Chr.17: 37625846-37625846 on Build GRCh38
rs137989328 Chr.17: 37625922-37625922 on Build GRCh38
rs747841191 Chr.17: 37625955-37625955 on Build GRCh38
rs547576759 Chr.17: 37625838-37625838 on Build GRCh38
rs1054324957 Chr.17: 37626086-37626086 on Build GRCh38
rs200253582 Chr.17: 37625883-37625883 on Build GRCh38
rs1052812193 Chr.17: 37626203-37626203 on Build GRCh38
rs138636479 Chr.17: 37626006-37626006 on Build GRCh38
rs755634170 Chr.17: 37626024-37626024 on Build GRCh38
rs752961463 Chr.17: 37625898-37625898 on Build GRCh38
rs761337604 Chr.17: 37626043-37626044 on Build GRCh38
rs778649073 Chr.17: 37626055-37626055 on Build GRCh38
rs769347732 Chr.17: 37626098-37626098 on Build GRCh38
rs145418414 Chr.17: 37626171-37626171 on Build GRCh38
rs147610462 Chr.17: 37625851-37625851 on Build GRCh38

View Full Product Details