Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 38733513-38734016 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CISD3
Entrez Gene ID
284106
Gene Name
CDGSH iron sulfur domain 3
Gene Aliases
Miner2
UniGene
Hs.713595
Gene Chromosome Location
Chr.17: 38730257-38735605 on Build GRCh38
Transcripts
NM_001136498.1 NM_007144.2 XM_005257640.2 XM_005257641.4 XM_005257642.3 XM_017025016.1

Gene Symbol
PCGF2
Entrez Gene ID
7703
Gene Name
polycomb group ring finger 2
Gene Aliases
MEL-18, RNF110, ZNF144
UniGene
Hs.371617
Gene Chromosome Location
Chr.17: 38733897-38749817 on Build GRCh38
Transcripts
NM_001136498.1 NM_007144.2 XM_005257640.2 XM_005257641.4 XM_005257642.3 XM_017025016.1

SNP Detail

SNP ID Chromosome Location
rs553565584 Chr.17: 38733756-38733756 on Build GRCh38
rs958015816 Chr.17: 38733690-38733690 on Build GRCh38
rs1023716662 Chr.17: 38733778-38733778 on Build GRCh38
rs966946026 Chr.17: 38733716-38733716 on Build GRCh38
rs34857493 Chr.17: 38733726-38733726 on Build GRCh38
rs1004471876 Chr.17: 38733620-38733620 on Build GRCh38
rs958302871 Chr.17: 38733858-38733858 on Build GRCh38
rs561563842 Chr.17: 38733988-38733988 on Build GRCh38
rs563725112 Chr.17: 38733878-38733878 on Build GRCh38
rs1002286801 Chr.17: 38733658-38733658 on Build GRCh38
rs1004296184 Chr.17: 38733646-38733646 on Build GRCh38
rs770407449 Chr.17: 38733945-38733945 on Build GRCh38
rs943714249 Chr.17: 38733557-38733557 on Build GRCh38
rs1025065096 Chr.17: 38733695-38733695 on Build GRCh38
rs554773170 Chr.17: 38733684-38733684 on Build GRCh38
rs545386128 Chr.17: 38733847-38733847 on Build GRCh38
rs1019102862 Chr.17: 38733783-38733783 on Build GRCh38
rs977106991 Chr.17: 38733946-38733946 on Build GRCh38
rs538093990 Chr.17: 38733626-38733626 on Build GRCh38
rs531130384 Chr.17: 38733972-38733972 on Build GRCh38
rs146587185 Chr.17: 38733986-38733986 on Build GRCh38
rs886973314 Chr.17: 38733627-38733627 on Build GRCh38
rs975030319 Chr.17: 38733977-38733977 on Build GRCh38
rs931241434 Chr.17: 38733552-38733552 on Build GRCh38
rs930980015 Chr.17: 38733954-38733954 on Build GRCh38
rs990173858 Chr.17: 38733905-38733905 on Build GRCh38
rs887000962 Chr.17: 38733612-38733612 on Build GRCh38
rs1033682822 Chr.17: 38733660-38733660 on Build GRCh38
rs1048328373 Chr.17: 38733608-38733608 on Build GRCh38
rs1012285948 Chr.17: 38733752-38733752 on Build GRCh38
rs562104601 Chr.17: 38733881-38733881 on Build GRCh38
rs965540734 Chr.17: 38733812-38733812 on Build GRCh38
rs76161689 Chr.17: 38733995-38733995 on Build GRCh38
rs576477429 Chr.17: 38733813-38733813 on Build GRCh38
rs931246136 Chr.17: 38733600-38733600 on Build GRCh38
rs569180772 Chr.17: 38733623-38733623 on Build GRCh38
rs967283559 Chr.17: 38733940-38733940 on Build GRCh38
rs1039565719 Chr.17: 38733561-38733561 on Build GRCh38
rs539029233 Chr.17: 38733548-38733548 on Build GRCh38
rs1021122018 Chr.17: 38733934-38733934 on Build GRCh38
rs558931700 Chr.17: 38733555-38733555 on Build GRCh38
rs188173076 Chr.17: 38733693-38733693 on Build GRCh38
rs866980465 Chr.17: 38733824-38733824 on Build GRCh38
rs776738159 Chr.17: 38733590-38733590 on Build GRCh38
rs201692804 Chr.17: 38733986-38733986 on Build GRCh38
rs912324081 Chr.17: 38733961-38733961 on Build GRCh38
rs1002226020 Chr.17: 38733806-38733806 on Build GRCh38

View Full Product Details