Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 38733919-38734422 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CISD3
Entrez Gene ID
284106
Gene Name
CDGSH iron sulfur domain 3
Gene Aliases
Miner2
UniGene
Hs.713595
Gene Chromosome Location
Chr.17: 38730257-38735605 on Build GRCh38
Transcripts
NM_001136498.1 NM_007144.2 XM_005257640.2 XM_005257641.4 XM_005257642.3 XM_017025016.1

Gene Symbol
PCGF2
Entrez Gene ID
7703
Gene Name
polycomb group ring finger 2
Gene Aliases
MEL-18, RNF110, ZNF144
UniGene
Hs.371617
Gene Chromosome Location
Chr.17: 38733897-38749817 on Build GRCh38
Transcripts
NM_001136498.1 NM_007144.2 XM_005257640.2 XM_005257641.4 XM_005257642.3 XM_017025016.1

SNP Detail

SNP ID Chromosome Location
rs952400880 Chr.17: 38734154-38734154 on Build GRCh38
rs1011119604 Chr.17: 38734087-38734087 on Build GRCh38
rs547115531 Chr.17: 38734061-38734061 on Build GRCh38
rs561563842 Chr.17: 38733988-38733988 on Build GRCh38
rs893897546 Chr.17: 38734223-38734223 on Build GRCh38
rs533745133 Chr.17: 38734395-38734395 on Build GRCh38
rs141292317 Chr.17: 38734084-38734084 on Build GRCh38
rs1009590543 Chr.17: 38734294-38734294 on Build GRCh38
rs531130384 Chr.17: 38733972-38733972 on Build GRCh38
rs146587185 Chr.17: 38733986-38733986 on Build GRCh38
rs1002319370 Chr.17: 38734071-38734071 on Build GRCh38
rs975030319 Chr.17: 38733977-38733977 on Build GRCh38
rs554553725 Chr.17: 38734350-38734350 on Build GRCh38
rs569248571 Chr.17: 38734267-38734267 on Build GRCh38
rs982034403 Chr.17: 38734181-38734181 on Build GRCh38
rs984019601 Chr.17: 38734253-38734253 on Build GRCh38
rs1042820407 Chr.17: 38734234-38734234 on Build GRCh38
rs76161689 Chr.17: 38733995-38733995 on Build GRCh38
rs1054843608 Chr.17: 38734221-38734221 on Build GRCh38
rs552605655 Chr.17: 38734162-38734162 on Build GRCh38
rs144991968 Chr.17: 38734160-38734160 on Build GRCh38
rs952483959 Chr.17: 38734220-38734220 on Build GRCh38
rs1019205984 Chr.17: 38734332-38734332 on Build GRCh38
rs964893623 Chr.17: 38734105-38734105 on Build GRCh38
rs570914364 Chr.17: 38734258-38734258 on Build GRCh38
rs538156057 Chr.17: 38734335-38734335 on Build GRCh38
rs912324081 Chr.17: 38733961-38733961 on Build GRCh38
rs1010862410 Chr.17: 38734054-38734055 on Build GRCh38
rs927818153 Chr.17: 38734198-38734200 on Build GRCh38
rs975600091 Chr.17: 38734175-38734175 on Build GRCh38
rs1028095378 Chr.17: 38734150-38734150 on Build GRCh38
rs775732069 Chr.17: 38734281-38734284 on Build GRCh38
rs745511175 Chr.17: 38734058-38734058 on Build GRCh38
rs796997805 Chr.17: 38734206-38734206 on Build GRCh38
rs1046494877 Chr.17: 38734055-38734055 on Build GRCh38
rs772549367 Chr.17: 38734239-38734240 on Build GRCh38
rs966821056 Chr.17: 38734116-38734116 on Build GRCh38
rs908329787 Chr.17: 38733997-38733997 on Build GRCh38
rs770407449 Chr.17: 38733945-38733945 on Build GRCh38
rs946792907 Chr.17: 38734228-38734228 on Build GRCh38
rs920934343 Chr.17: 38734215-38734215 on Build GRCh38
rs977106991 Chr.17: 38733946-38733946 on Build GRCh38
rs930980015 Chr.17: 38733954-38733954 on Build GRCh38
rs1055123650 Chr.17: 38734012-38734012 on Build GRCh38
rs111988561 Chr.17: 38734370-38734370 on Build GRCh38
rs773148114 Chr.17: 38734113-38734113 on Build GRCh38
rs974879241 Chr.17: 38734106-38734106 on Build GRCh38
rs1021634410 Chr.17: 38734115-38734115 on Build GRCh38
rs11653368 Chr.17: 38734168-38734168 on Build GRCh38
rs370251783 Chr.17: 38734244-38734244 on Build GRCh38
rs769183102 Chr.17: 38734085-38734085 on Build GRCh38
rs939945857 Chr.17: 38734271-38734271 on Build GRCh38
rs938123604 Chr.17: 38734002-38734002 on Build GRCh38
rs939850570 Chr.17: 38734042-38734042 on Build GRCh38
rs201692804 Chr.17: 38733986-38733986 on Build GRCh38
rs772678561 Chr.17: 38734288-38734288 on Build GRCh38

View Full Product Details