Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 38954575-38955009 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO47
Entrez Gene ID
494188
Gene Name
F-box protein 47
Gene Aliases
-
UniGene
Hs.549536
Gene Chromosome Location
Chr.17: 38936278-38967475 on Build GRCh38
Transcripts
NM_001008777.2 XM_011524865.2 XM_011524866.2 XM_011524867.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4410443 c.430-1G>T p.? Chr.17: 38954934-38954934 on Build GRCh38
5456609 c.506A>G p.Q169R Chr.17: 38954857-38954857 on Build GRCh38
978516 c.507+2T>C p.? Chr.17: 38954854-38954854 on Build GRCh38
5386193 c.494G>A p.G165D Chr.17: 38954869-38954869 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs554557125 Chr.17: 38954899-38954899 on Build GRCh38
rs747915639 Chr.17: 38954851-38954851 on Build GRCh38
rs979178913 Chr.17: 38954739-38954739 on Build GRCh38
rs778860503 Chr.17: 38954849-38954849 on Build GRCh38
rs777342528 Chr.17: 38954881-38954881 on Build GRCh38
rs765430568 Chr.17: 38954842-38954843 on Build GRCh38
rs1639929 Chr.17: 38954689-38954689 on Build GRCh38
rs78522966 Chr.17: 38954649-38954649 on Build GRCh38
rs925025100 Chr.17: 38954952-38954952 on Build GRCh38
rs774476028 Chr.17: 38954960-38954960 on Build GRCh38
rs997024505 Chr.17: 38954658-38954658 on Build GRCh38
rs762088432 Chr.17: 38954963-38954963 on Build GRCh38
rs868157981 Chr.17: 38954929-38954929 on Build GRCh38
rs1040789133 Chr.17: 38954623-38954623 on Build GRCh38
rs771097450 Chr.17: 38954938-38954938 on Build GRCh38
rs553336064 Chr.17: 38954670-38954670 on Build GRCh38
rs552277876 Chr.17: 38954627-38954627 on Build GRCh38
rs754967119 Chr.17: 38954984-38954984 on Build GRCh38
rs757327318 Chr.17: 38954825-38954825 on Build GRCh38
rs750469048 Chr.17: 38954968-38954968 on Build GRCh38
rs568724763 Chr.17: 38954682-38954682 on Build GRCh38
rs375223374 Chr.17: 38954959-38954959 on Build GRCh38
rs763177657 Chr.17: 38954949-38954949 on Build GRCh38
rs113385999 Chr.17: 38954981-38954981 on Build GRCh38
rs771781481 Chr.17: 38954879-38954879 on Build GRCh38
rs888623621 Chr.17: 38954667-38954667 on Build GRCh38
rs867639414 Chr.17: 38954710-38954710 on Build GRCh38
rs770220457 Chr.17: 38954914-38954914 on Build GRCh38
rs79488852 Chr.17: 38954720-38954720 on Build GRCh38
rs145872520 Chr.17: 38954663-38954663 on Build GRCh38
rs776107374 Chr.17: 38954908-38954908 on Build GRCh38
rs961349346 Chr.17: 38954860-38954860 on Build GRCh38
rs987728282 Chr.17: 38954961-38954961 on Build GRCh38
rs767548996 Chr.17: 38954965-38954965 on Build GRCh38
rs754948958 Chr.17: 38954847-38954847 on Build GRCh38
rs1015870783 Chr.17: 38954820-38954820 on Build GRCh38
rs746644991 Chr.17: 38954893-38954893 on Build GRCh38
rs761956756 Chr.17: 38954843-38954843 on Build GRCh38
rs760050684 Chr.17: 38954910-38954910 on Build GRCh38
rs781297799 Chr.17: 38954826-38954826 on Build GRCh38
rs866625205 Chr.17: 38954941-38954941 on Build GRCh38

View Full Product Details