Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 39183914-39184450 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNB1
Entrez Gene ID
782
Gene Name
calcium voltage-gated channel auxiliary subunit beta 1
Gene Aliases
CAB1, CACNLB1, CCHLB1
UniGene
Hs.635
Gene Chromosome Location
Chr.17: 39173456-39197703 on Build GRCh38
Transcripts
NM_000723.4 NM_199247.2 NM_199248.2 XM_005257645.2 XM_005257646.2 XM_006722072.2 XM_017025024.1 XM_017025025.1 XM_017025026.1 XM_017025027.1 XM_017025028.1 XM_017025029.1 XM_017025030.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6146707 c.1005T>A p.I335I Chr.17: 39184059-39184059 on Build GRCh38
4979723 c.878C>G p.S293C Chr.17: 39184051-39184051 on Build GRCh38
4668740 c.809C>T p.T270M Chr.17: 39184120-39184120 on Build GRCh38
1382790 c.987C>G p.P329P Chr.17: 39184077-39184077 on Build GRCh38
1382792 c.937C>T p.R313C Chr.17: 39184127-39184127 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11867859 Chr.17: 39184255-39184255 on Build GRCh38
rs202030955 Chr.17: 39184080-39184080 on Build GRCh38
rs759717309 Chr.17: 39184000-39184000 on Build GRCh38
rs777109375 Chr.17: 39184131-39184131 on Build GRCh38
rs1033757480 Chr.17: 39183972-39183972 on Build GRCh38
rs1038968188 Chr.17: 39184235-39184235 on Build GRCh38
rs559307773 Chr.17: 39184400-39184400 on Build GRCh38
rs765061590 Chr.17: 39184299-39184299 on Build GRCh38
rs747575318 Chr.17: 39184137-39184137 on Build GRCh38
rs1048058317 Chr.17: 39184244-39184244 on Build GRCh38
rs17853739 Chr.17: 39184138-39184138 on Build GRCh38
rs377703005 Chr.17: 39184091-39184091 on Build GRCh38
rs370346231 Chr.17: 39184133-39184133 on Build GRCh38
rs1051629758 Chr.17: 39184175-39184175 on Build GRCh38
rs369335412 Chr.17: 39184188-39184188 on Build GRCh38
rs747731549 Chr.17: 39184027-39184027 on Build GRCh38
rs779192417 Chr.17: 39184420-39184420 on Build GRCh38
rs780346179 Chr.17: 39184395-39184395 on Build GRCh38
rs200175077 Chr.17: 39184143-39184143 on Build GRCh38
rs749639772 Chr.17: 39184399-39184399 on Build GRCh38
rs762467500 Chr.17: 39184304-39184304 on Build GRCh38
rs778931435 Chr.17: 39184001-39184001 on Build GRCh38
rs931063773 Chr.17: 39184219-39184219 on Build GRCh38
rs768802644 Chr.17: 39184418-39184418 on Build GRCh38
rs879094157 Chr.17: 39184002-39184002 on Build GRCh38
rs534874332 Chr.17: 39184323-39184323 on Build GRCh38
rs757968830 Chr.17: 39184037-39184037 on Build GRCh38
rs570141603 Chr.17: 39183992-39183992 on Build GRCh38
rs147889821 Chr.17: 39184120-39184120 on Build GRCh38
rs78894589 Chr.17: 39184083-39184083 on Build GRCh38
rs531185253 Chr.17: 39184003-39184003 on Build GRCh38
rs759421843 Chr.17: 39184298-39184298 on Build GRCh38
rs770842288 Chr.17: 39184285-39184285 on Build GRCh38
rs571087887 Chr.17: 39184082-39184082 on Build GRCh38
rs994493912 Chr.17: 39184243-39184243 on Build GRCh38
rs749470987 Chr.17: 39184179-39184179 on Build GRCh38
rs756501317 Chr.17: 39184394-39184394 on Build GRCh38
rs746198242 Chr.17: 39184043-39184043 on Build GRCh38
rs374004024 Chr.17: 39184314-39184314 on Build GRCh38
rs994546034 Chr.17: 39183946-39183946 on Build GRCh38
rs376120125 Chr.17: 39184319-39184319 on Build GRCh38
rs933993710 Chr.17: 39184423-39184423 on Build GRCh38
rs373015201 Chr.17: 39184274-39184274 on Build GRCh38
rs766045356 Chr.17: 39184015-39184015 on Build GRCh38
rs1003923108 Chr.17: 39184251-39184251 on Build GRCh38
rs760477594 Chr.17: 39184092-39184092 on Build GRCh38
rs113407357 Chr.17: 39184248-39184248 on Build GRCh38
rs373568239 Chr.17: 39184145-39184145 on Build GRCh38
rs781670933 Chr.17: 39184338-39184338 on Build GRCh38
rs576153053 Chr.17: 39184236-39184236 on Build GRCh38
rs370438039 Chr.17: 39184018-39184018 on Build GRCh38
rs770085936 Chr.17: 39184177-39184177 on Build GRCh38
rs369245854 Chr.17: 39184262-39184262 on Build GRCh38
rs750862091 Chr.17: 39184356-39184356 on Build GRCh38
rs763030691 Chr.17: 39184075-39184075 on Build GRCh38
rs75090495 Chr.17: 39184119-39184119 on Build GRCh38
rs750265808 Chr.17: 39184005-39184005 on Build GRCh38
rs765045106 Chr.17: 39184113-39184113 on Build GRCh38
rs970159464 Chr.17: 39184283-39184283 on Build GRCh38
rs775690339 Chr.17: 39184178-39184178 on Build GRCh38
rs752431863 Chr.17: 39184301-39184301 on Build GRCh38
rs144168844 Chr.17: 39184216-39184216 on Build GRCh38
rs552500952 Chr.17: 39184067-39184067 on Build GRCh38
rs111370424 Chr.17: 39183990-39183990 on Build GRCh38
rs778680819 Chr.17: 39184025-39184025 on Build GRCh38
rs775676025 Chr.17: 39184070-39184070 on Build GRCh38
rs759446272 Chr.17: 39184292-39184297 on Build GRCh38
rs776918261 Chr.17: 39183994-39183994 on Build GRCh38
rs1026476836 Chr.17: 39183949-39183949 on Build GRCh38
rs761159262 Chr.17: 39183989-39183989 on Build GRCh38
rs369891566 Chr.17: 39184291-39184291 on Build GRCh38
rs1039966261 Chr.17: 39184210-39184210 on Build GRCh38
rs776696700 Chr.17: 39184292-39184292 on Build GRCh38
rs747408791 Chr.17: 39184039-39184039 on Build GRCh38
rs771360174 Chr.17: 39184041-39184041 on Build GRCh38
rs376850728 Chr.17: 39184042-39184042 on Build GRCh38
rs774694063 Chr.17: 39184143-39184143 on Build GRCh38
rs374609331 Chr.17: 39184090-39184090 on Build GRCh38
rs987905518 Chr.17: 39184337-39184337 on Build GRCh38
rs774345146 Chr.17: 39184189-39184189 on Build GRCh38
rs753729987 Chr.17: 39184096-39184096 on Build GRCh38
rs778357476 Chr.17: 39184038-39184038 on Build GRCh38
rs766320294 Chr.17: 39184094-39184094 on Build GRCh38
rs755939533 Chr.17: 39184008-39184008 on Build GRCh38

View Full Product Details