Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 39462208-39462718 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK12
Entrez Gene ID
51755
Gene Name
cyclin dependent kinase 12
Gene Aliases
CRK7, CRKR, CRKRS
UniGene
Hs.345028
Gene Chromosome Location
Chr.17: 39461486-39567560 on Build GRCh38
Transcripts
NM_015083.2 NM_016507.3 XM_005257456.3 XM_005257458.4 XM_011524892.2 XM_011524893.2 XM_011524894.2 XM_011524895.2 XM_011524896.2 XM_011524897.2 XM_011524898.2 XM_011524899.2 XM_011524900.2 XM_011524901.2 XM_011524902.2 XM_011524903.2 XM_011524905.2 XM_011524906.2 XM_011524907.2 XM_017024744.1 XM_017024745.1 XM_017024746.1 XM_017024747.1 XM_017024748.1 XM_017024749.1 XM_017024750.1 XM_017024751.1 XM_017024752.1 XM_017024753.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1302739 c.565G>A p.E189K Chr.17: 39462636-39462636 on Build GRCh38
6947222 c.555G>C p.R185S Chr.17: 39462626-39462626 on Build GRCh38
6909838 c.453_468del16 p.K152fs*10 Chr.17: 39462524-39462539 on Build GRCh38
4990974 c.619C>T p.P207S Chr.17: 39462690-39462690 on Build GRCh38
6948175 c.295C>T p.R99C Chr.17: 39462366-39462366 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201864067 Chr.17: 39462495-39462495 on Build GRCh38
rs908457937 Chr.17: 39462614-39462614 on Build GRCh38
rs771462646 Chr.17: 39462639-39462639 on Build GRCh38
rs1021200845 Chr.17: 39462423-39462423 on Build GRCh38
rs760641257 Chr.17: 39462692-39462692 on Build GRCh38
rs769915324 Chr.17: 39462656-39462656 on Build GRCh38
rs149034064 Chr.17: 39462342-39462342 on Build GRCh38
rs777158914 Chr.17: 39462644-39462644 on Build GRCh38
rs778567890 Chr.17: 39462603-39462603 on Build GRCh38
rs780116042 Chr.17: 39462660-39462660 on Build GRCh38
rs558704332 Chr.17: 39462483-39462483 on Build GRCh38
rs373315719 Chr.17: 39462450-39462450 on Build GRCh38
rs142040871 Chr.17: 39462484-39462484 on Build GRCh38
rs757116953 Chr.17: 39462324-39462324 on Build GRCh38
rs753976596 Chr.17: 39462272-39462272 on Build GRCh38
rs774974902 Chr.17: 39462383-39462383 on Build GRCh38
rs1008908878 Chr.17: 39462499-39462499 on Build GRCh38
rs543840788 Chr.17: 39462317-39462317 on Build GRCh38
rs541561206 Chr.17: 39462367-39462367 on Build GRCh38
rs1055721209 Chr.17: 39462349-39462349 on Build GRCh38
rs370663761 Chr.17: 39462533-39462533 on Build GRCh38
rs778313862 Chr.17: 39462470-39462470 on Build GRCh38
rs9896487 Chr.17: 39462554-39462554 on Build GRCh38
rs200703770 Chr.17: 39462418-39462418 on Build GRCh38
rs755902248 Chr.17: 39462451-39462451 on Build GRCh38
rs768025531 Chr.17: 39462393-39462393 on Build GRCh38
rs201160144 Chr.17: 39462588-39462588 on Build GRCh38
rs964807686 Chr.17: 39462541-39462541 on Build GRCh38
rs752782484 Chr.17: 39462429-39462429 on Build GRCh38
rs548797163 Chr.17: 39462596-39462596 on Build GRCh38
rs759887608 Chr.17: 39462646-39462646 on Build GRCh38
rs758471913 Chr.17: 39462542-39462542 on Build GRCh38
rs753585320 Chr.17: 39462694-39462694 on Build GRCh38
rs747496820 Chr.17: 39462634-39462634 on Build GRCh38
rs1046007764 Chr.17: 39462337-39462337 on Build GRCh38
rs773729401 Chr.17: 39462397-39462397 on Build GRCh38
rs765207917 Chr.17: 39462420-39462420 on Build GRCh38
rs151330887 Chr.17: 39462608-39462608 on Build GRCh38
rs1019981847 Chr.17: 39462585-39462585 on Build GRCh38
rs377586655 Chr.17: 39462441-39462441 on Build GRCh38
rs984483662 Chr.17: 39462604-39462604 on Build GRCh38
rs188667797 Chr.17: 39462469-39462469 on Build GRCh38
rs761059862 Chr.17: 39462398-39462398 on Build GRCh38
rs771407477 Chr.17: 39462524-39462524 on Build GRCh38
rs750404979 Chr.17: 39462691-39462691 on Build GRCh38
rs79630893 Chr.17: 39462500-39462500 on Build GRCh38
rs752734792 Chr.17: 39462550-39462550 on Build GRCh38
rs1048293300 Chr.17: 39462266-39462266 on Build GRCh38
rs779797685 Chr.17: 39462593-39462593 on Build GRCh38
rs754105952 Chr.17: 39462409-39462409 on Build GRCh38
rs748874194 Chr.17: 39462464-39462464 on Build GRCh38
rs1014209082 Chr.17: 39462424-39462424 on Build GRCh38
rs957118817 Chr.17: 39462514-39462514 on Build GRCh38
rs955200004 Chr.17: 39462613-39462615 on Build GRCh38
rs561243282 Chr.17: 39462256-39462256 on Build GRCh38
rs764139189 Chr.17: 39462680-39462680 on Build GRCh38
rs773784590 Chr.17: 39462494-39462494 on Build GRCh38
rs145123537 Chr.17: 39462331-39462331 on Build GRCh38
rs140590436 Chr.17: 39462693-39462693 on Build GRCh38
rs763025111 Chr.17: 39462553-39462553 on Build GRCh38
rs368803953 Chr.17: 39462527-39462527 on Build GRCh38
rs753625450 Chr.17: 39462594-39462594 on Build GRCh38
rs374544508 Chr.17: 39462626-39462626 on Build GRCh38
rs779742493 Chr.17: 39462455-39462455 on Build GRCh38
rs747640647 Chr.17: 39462471-39462471 on Build GRCh38
rs921081528 Chr.17: 39462254-39462254 on Build GRCh38
rs766655918 Chr.17: 39462258-39462258 on Build GRCh38
rs767753760 Chr.17: 39462233-39462233 on Build GRCh38
rs763003770 Chr.17: 39462677-39462677 on Build GRCh38
rs748867645 Chr.17: 39462359-39462359 on Build GRCh38
rs905721300 Chr.17: 39462490-39462490 on Build GRCh38
rs755955758 Chr.17: 39462590-39462590 on Build GRCh38
rs781051442 Chr.17: 39462446-39462446 on Build GRCh38
rs772467240 Chr.17: 39462475-39462475 on Build GRCh38
rs893957608 Chr.17: 39462389-39462389 on Build GRCh38
rs150124402 Chr.17: 39462305-39462305 on Build GRCh38
rs147398865 Chr.17: 39462237-39462237 on Build GRCh38
rs767512934 Chr.17: 39462555-39462555 on Build GRCh38
rs147574732 Chr.17: 39462338-39462338 on Build GRCh38
rs780882570 Chr.17: 39462330-39462330 on Build GRCh38
rs771702282 Chr.17: 39462620-39462620 on Build GRCh38
rs759709034 Chr.17: 39462532-39462532 on Build GRCh38
rs927611525 Chr.17: 39462669-39462669 on Build GRCh38
rs34303030 Chr.17: 39462246-39462247 on Build GRCh38
rs762191969 Chr.17: 39462385-39462385 on Build GRCh38

View Full Product Details